Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1703539-1703782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLBP
Entrez Gene ID
7884
Gene Name
stem-loop binding protein
Gene Aliases
HBP
UniGene
Hs.298345
Gene Chromosome Location
Chr.4: 1692731-1712741 on Build GRCh38
Transcripts
NM_001306074.1 NM_001306075.1 NM_006527.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1225839 c.271G>T p.E91* Chr.4: 1703606-1703606 on Build GRCh38
5948154 c.181A>G p.T61A Chr.4: 1703696-1703696 on Build GRCh38
1053180 c.206G>A p.R69H Chr.4: 1703671-1703671 on Build GRCh38
5866061 c.197C>T p.P66L Chr.4: 1703680-1703680 on Build GRCh38
6779170 c.263T>C p.V88A Chr.4: 1703614-1703614 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566930816 Chr.4: 1703718-1703718 on Build GRCh38
rs771608554 Chr.4: 1703649-1703649 on Build GRCh38
rs2854911 Chr.4: 1703720-1703720 on Build GRCh38
rs752644515 Chr.4: 1703597-1703597 on Build GRCh38
rs771665114 Chr.4: 1703741-1703745 on Build GRCh38
rs770486797 Chr.4: 1703634-1703634 on Build GRCh38
rs752838621 Chr.4: 1703675-1703675 on Build GRCh38
rs888965118 Chr.4: 1703660-1703660 on Build GRCh38
rs781246659 Chr.4: 1703591-1703591 on Build GRCh38
rs141111228 Chr.4: 1703647-1703647 on Build GRCh38
rs969964816 Chr.4: 1703744-1703744 on Build GRCh38
rs749113394 Chr.4: 1703633-1703633 on Build GRCh38
rs755161422 Chr.4: 1703580-1703580 on Build GRCh38
rs766491458 Chr.4: 1703669-1703669 on Build GRCh38
rs369980820 Chr.4: 1703714-1703714 on Build GRCh38
rs777375245 Chr.4: 1703630-1703630 on Build GRCh38
rs762256523 Chr.4: 1703651-1703651 on Build GRCh38
rs778593556 Chr.4: 1703638-1703638 on Build GRCh38
rs554781467 Chr.4: 1703688-1703688 on Build GRCh38
rs746530753 Chr.4: 1703724-1703724 on Build GRCh38
rs915202413 Chr.4: 1703637-1703637 on Build GRCh38
rs757123258 Chr.4: 1703713-1703713 on Build GRCh38
rs981878416 Chr.4: 1703581-1703581 on Build GRCh38
rs41286667 Chr.4: 1703672-1703672 on Build GRCh38
rs759564710 Chr.4: 1703707-1703707 on Build GRCh38
rs770960111 Chr.4: 1703577-1703577 on Build GRCh38
rs1050332699 Chr.4: 1703653-1703653 on Build GRCh38
rs986142249 Chr.4: 1703578-1703578 on Build GRCh38
rs751873933 Chr.4: 1703671-1703671 on Build GRCh38
rs775011181 Chr.4: 1703650-1703650 on Build GRCh38
rs745481375 Chr.4: 1703716-1703716 on Build GRCh38
rs774479287 Chr.4: 1703632-1703634 on Build GRCh38
rs140082881 Chr.4: 1703676-1703676 on Build GRCh38
rs150237030 Chr.4: 1703655-1703655 on Build GRCh38
rs202009267 Chr.4: 1703728-1703728 on Build GRCh38
rs866760917 Chr.4: 1703690-1703690 on Build GRCh38
rs41286669 Chr.4: 1703738-1703738 on Build GRCh38
rs770192707 Chr.4: 1703725-1703725 on Build GRCh38
rs756101521 Chr.4: 1703611-1703611 on Build GRCh38
rs771288881 Chr.4: 1703746-1703746 on Build GRCh38
rs753562913 Chr.4: 1703702-1703702 on Build GRCh38

View Full Product Details