Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86022358-86022593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PICALM
Entrez Gene ID
8301
Gene Name
phosphatidylinositol binding clathrin assembly protein
Gene Aliases
CALM, CLTH, LAP
UniGene
Hs.163893
Gene Chromosome Location
Chr.11: 85957171-86069881 on Build GRCh38
Transcripts
NM_001008660.2 NM_001206946.1 NM_001206947.1 NM_007166.3 XM_005274322.2 XM_005274323.2 XM_005274324.2 XM_005274325.2 XM_005274326.2 XM_005274327.2 XM_005274328.2 XM_005274329.3 XM_005274330.2 XM_005274331.2 XM_005274332.2 XM_005274333.2 XM_005274334.2 XM_005274335.2 XM_005274336.2 XM_005274337.2 XM_005274340.2 XM_006718699.2 XM_006718700.2 XM_006718701.3 XM_011545293.1 XM_017018381.1 XM_017018382.1 XM_017018383.1 XM_017018384.1 XM_017018385.1 XM_017018386.1 XM_017018387.1 XM_017018388.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7203692 c.426A>G p.A142A Chr.11: 86022393-86022393 on Build GRCh38
1492717 c.405A>G p.A135A Chr.11: 86022414-86022414 on Build GRCh38
4545823 c.386G>C p.R129T Chr.11: 86022433-86022433 on Build GRCh38
1561941 c.375G>A p.R125R Chr.11: 86022444-86022444 on Build GRCh38
6238556 c.435C>A p.F145L Chr.11: 86022384-86022384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769811550 Chr.11: 86022499-86022500 on Build GRCh38
rs746886990 Chr.11: 86022488-86022489 on Build GRCh38
rs745573027 Chr.11: 86022476-86022487 on Build GRCh38
rs749688028 Chr.11: 86022487-86022487 on Build GRCh38
rs760656374 Chr.11: 86022513-86022513 on Build GRCh38
rs775862399 Chr.11: 86022482-86022482 on Build GRCh38
rs767609843 Chr.11: 86022505-86022505 on Build GRCh38
rs878929487 Chr.11: 86022417-86022417 on Build GRCh38
rs768946453 Chr.11: 86022495-86022495 on Build GRCh38
rs531548903 Chr.11: 86022410-86022410 on Build GRCh38
rs995527987 Chr.11: 86022545-86022545 on Build GRCh38
rs774869921 Chr.11: 86022518-86022519 on Build GRCh38
rs759894965 Chr.11: 86022519-86022520 on Build GRCh38
rs747618387 Chr.11: 86022434-86022434 on Build GRCh38
rs779877592 Chr.11: 86022487-86022487 on Build GRCh38
rs1014468074 Chr.11: 86022401-86022401 on Build GRCh38
rs775830584 Chr.11: 86022468-86022468 on Build GRCh38
rs763333854 Chr.11: 86022500-86022501 on Build GRCh38
rs757874575 Chr.11: 86022453-86022453 on Build GRCh38
rs758162571 Chr.11: 86022481-86022481 on Build GRCh38
rs774741079 Chr.11: 86022502-86022502 on Build GRCh38
rs762163855 Chr.11: 86022504-86022504 on Build GRCh38
rs770274147 Chr.11: 86022480-86022480 on Build GRCh38
rs1020889379 Chr.11: 86022420-86022420 on Build GRCh38
rs763456109 Chr.11: 86022471-86022473 on Build GRCh38
rs200363412 Chr.11: 86022444-86022444 on Build GRCh38
rs773536823 Chr.11: 86022501-86022502 on Build GRCh38
rs778562552 Chr.11: 86022476-86022481 on Build GRCh38
rs753395320 Chr.11: 86022471-86022472 on Build GRCh38
rs146648955 Chr.11: 86022499-86022502 on Build GRCh38
rs766836971 Chr.11: 86022509-86022510 on Build GRCh38
rs746375569 Chr.11: 86022473-86022473 on Build GRCh38
rs374624574 Chr.11: 86022500-86022503 on Build GRCh38
rs777199260 Chr.11: 86022465-86022465 on Build GRCh38
rs145115354 Chr.11: 86022389-86022389 on Build GRCh38
rs768640519 Chr.11: 86022490-86022493 on Build GRCh38
rs754558342 Chr.11: 86022403-86022403 on Build GRCh38
rs776530570 Chr.11: 86022493-86022493 on Build GRCh38
rs756823045 Chr.11: 86022473-86022474 on Build GRCh38
rs753642858 Chr.11: 86022396-86022396 on Build GRCh38
rs773220359 Chr.11: 86022508-86022508 on Build GRCh38
rs967543022 Chr.11: 86022442-86022442 on Build GRCh38

View Full Product Details