Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 114443775-114444012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REXO2
Entrez Gene ID
25996
Gene Name
RNA exonuclease 2
Gene Aliases
CGI-114, REX2, RFN, SFN
UniGene
Hs.677190
Gene Chromosome Location
Chr.11: 114439386-114450279 on Build GRCh38
Transcripts
NM_015523.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
466350 c.285T>C p.D95D Chr.11: 114443909-114443909 on Build GRCh38
1264033 c.304G>A p.G102R Chr.11: 114443928-114443928 on Build GRCh38
5837835 c.290G>A p.C97Y Chr.11: 114443914-114443914 on Build GRCh38
5443131 c.243G>C p.L81L Chr.11: 114443867-114443867 on Build GRCh38
5480819 c.261T>C p.D87D Chr.11: 114443885-114443885 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781340317 Chr.11: 114443883-114443883 on Build GRCh38
rs770041778 Chr.11: 114443888-114443888 on Build GRCh38
rs764928376 Chr.11: 114443846-114443846 on Build GRCh38
rs750108555 Chr.11: 114443970-114443970 on Build GRCh38
rs763259090 Chr.11: 114443975-114443975 on Build GRCh38
rs866234894 Chr.11: 114443912-114443912 on Build GRCh38
rs772022921 Chr.11: 114443907-114443907 on Build GRCh38
rs143578716 Chr.11: 114443919-114443919 on Build GRCh38
rs760062610 Chr.11: 114443833-114443833 on Build GRCh38
rs761280301 Chr.11: 114443953-114443953 on Build GRCh38
rs372060779 Chr.11: 114443870-114443870 on Build GRCh38
rs375748634 Chr.11: 114443965-114443965 on Build GRCh38
rs372611718 Chr.11: 114443824-114443824 on Build GRCh38
rs766748226 Chr.11: 114443982-114443982 on Build GRCh38
rs780499509 Chr.11: 114443857-114443857 on Build GRCh38
rs760653189 Chr.11: 114443961-114443961 on Build GRCh38
rs776193591 Chr.11: 114443936-114443936 on Build GRCh38
rs186799877 Chr.11: 114443831-114443831 on Build GRCh38
rs750765972 Chr.11: 114443847-114443847 on Build GRCh38
rs115668978 Chr.11: 114443901-114443901 on Build GRCh38
rs1056243335 Chr.11: 114443977-114443977 on Build GRCh38
rs151260750 Chr.11: 114443927-114443927 on Build GRCh38
rs763682419 Chr.11: 114443839-114443839 on Build GRCh38
rs199983925 Chr.11: 114443909-114443909 on Build GRCh38
rs114077514 Chr.11: 114443885-114443885 on Build GRCh38
rs140096192 Chr.11: 114443862-114443862 on Build GRCh38
rs756325852 Chr.11: 114443871-114443871 on Build GRCh38
rs201888038 Chr.11: 114443841-114443841 on Build GRCh38
rs753372372 Chr.11: 114443840-114443840 on Build GRCh38
rs754958821 Chr.11: 114443882-114443882 on Build GRCh38
rs778185959 Chr.11: 114443892-114443892 on Build GRCh38

View Full Product Details