Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 31463189-31463414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMMP1L
Entrez Gene ID
196294
Gene Name
inner mitochondrial membrane peptidase subunit 1
Gene Aliases
IMMP1, IMP1, IMP1-LIKE
UniGene
Hs.502223
Gene Chromosome Location
Chr.11: 31432399-31509644 on Build GRCh38
Transcripts
NM_001304274.1 NM_144981.2 XM_005252812.3 XM_011519942.1 XM_011519943.2 XM_011519944.2 XM_011519945.2 XM_011519946.2 XM_011519947.2 XM_017017305.1 XM_017017306.1 XM_017017307.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
926329 c.32G>A p.R11Q Chr.11: 31463245-31463245 on Build GRCh38
311988 c.31C>A p.R11R Chr.11: 31463246-31463246 on Build GRCh38
321001 c.71A>G p.H24R Chr.11: 31463206-31463206 on Build GRCh38
4032347 c.42C>G p.G14G Chr.11: 31463235-31463235 on Build GRCh38
5781180 c.10G>T p.G4C Chr.11: 31463267-31463267 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185838481 Chr.11: 31463294-31463294 on Build GRCh38
rs1056478448 Chr.11: 31463338-31463338 on Build GRCh38
rs756626046 Chr.11: 31463221-31463221 on Build GRCh38
rs767086495 Chr.11: 31463217-31463217 on Build GRCh38
rs966310425 Chr.11: 31463379-31463379 on Build GRCh38
rs771566124 Chr.11: 31463300-31463300 on Build GRCh38
rs777823531 Chr.11: 31463279-31463279 on Build GRCh38
rs374791852 Chr.11: 31463310-31463310 on Build GRCh38
rs267602846 Chr.11: 31463267-31463267 on Build GRCh38
rs997747348 Chr.11: 31463383-31463383 on Build GRCh38
rs150556583 Chr.11: 31463235-31463235 on Build GRCh38
rs200943972 Chr.11: 31463218-31463218 on Build GRCh38
rs1005071466 Chr.11: 31463367-31463367 on Build GRCh38
rs759231886 Chr.11: 31463299-31463299 on Build GRCh38
rs892541206 Chr.11: 31463355-31463355 on Build GRCh38
rs148605225 Chr.11: 31463368-31463368 on Build GRCh38
rs767462747 Chr.11: 31463274-31463275 on Build GRCh38
rs540682544 Chr.11: 31463277-31463277 on Build GRCh38
rs180782155 Chr.11: 31463245-31463245 on Build GRCh38
rs375730259 Chr.11: 31463269-31463269 on Build GRCh38
rs200769250 Chr.11: 31463281-31463281 on Build GRCh38
rs758487097 Chr.11: 31463246-31463246 on Build GRCh38
rs775037875 Chr.11: 31463317-31463317 on Build GRCh38
rs78159446 Chr.11: 31463270-31463270 on Build GRCh38
rs1056162884 Chr.11: 31463258-31463258 on Build GRCh38
rs764570303 Chr.11: 31463224-31463224 on Build GRCh38
rs754265667 Chr.11: 31463233-31463233 on Build GRCh38
rs749145606 Chr.11: 31463280-31463280 on Build GRCh38
rs1042166248 Chr.11: 31463223-31463223 on Build GRCh38

View Full Product Details