Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 85114929-85115174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRCC1
Entrez Gene ID
85444
Gene Name
leucine rich repeat and coiled-coil centrosomal protein 1
Gene Aliases
CLERC, SAP2, VFL1
UniGene
Hs.193115
Gene Chromosome Location
Chr.8: 85107088-85146080 on Build GRCh38
Transcripts
NM_033402.4 XM_005251318.3 XM_005251319.4 XM_005251320.3 XM_005251322.3 XM_017013920.1 XM_017013921.1 XM_017013922.1 XM_017013923.1 XM_017013924.1 XM_017013925.1 XM_017013926.1 XM_017013927.1 XM_017013928.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6837276 c.596G>A p.C199Y Chr.8: 85115151-85115151 on Build GRCh38
346941 c.581T>A p.L194H Chr.8: 85115136-85115136 on Build GRCh38
6837277 c.536G>A p.C179Y Chr.8: 85115151-85115151 on Build GRCh38
346942 c.521T>A p.L174H Chr.8: 85115136-85115136 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748073058 Chr.8: 85115081-85115081 on Build GRCh38
rs141555401 Chr.8: 85115045-85115045 on Build GRCh38
rs770639195 Chr.8: 85115111-85115111 on Build GRCh38
rs530851164 Chr.8: 85115029-85115029 on Build GRCh38
rs16913590 Chr.8: 85114997-85114997 on Build GRCh38
rs781163742 Chr.8: 85115066-85115066 on Build GRCh38
rs573481162 Chr.8: 85114974-85114974 on Build GRCh38
rs901342084 Chr.8: 85115003-85115003 on Build GRCh38
rs952513993 Chr.8: 85115127-85115127 on Build GRCh38
rs982029155 Chr.8: 85115091-85115091 on Build GRCh38
rs1028084745 Chr.8: 85115016-85115016 on Build GRCh38
rs746818675 Chr.8: 85115055-85115055 on Build GRCh38
rs769879651 Chr.8: 85115087-85115087 on Build GRCh38
rs764408502 Chr.8: 85115134-85115134 on Build GRCh38
rs1026862143 Chr.8: 85115119-85115119 on Build GRCh38
rs369799406 Chr.8: 85115117-85115117 on Build GRCh38
rs905606190 Chr.8: 85114998-85114998 on Build GRCh38
rs759952689 Chr.8: 85115005-85115005 on Build GRCh38
rs761079593 Chr.8: 85115131-85115131 on Build GRCh38
rs768650424 Chr.8: 85115055-85115055 on Build GRCh38
rs565231763 Chr.8: 85115011-85115011 on Build GRCh38
rs903190981 Chr.8: 85114964-85114967 on Build GRCh38
rs748821311 Chr.8: 85115104-85115104 on Build GRCh38
rs768396058 Chr.8: 85115058-85115058 on Build GRCh38
rs997397376 Chr.8: 85115008-85115008 on Build GRCh38
rs776830325 Chr.8: 85115002-85115002 on Build GRCh38
rs777085889 Chr.8: 85115147-85115147 on Build GRCh38
rs376491425 Chr.8: 85114961-85114961 on Build GRCh38
rs960998441 Chr.8: 85115088-85115091 on Build GRCh38
rs948212142 Chr.8: 85114981-85114981 on Build GRCh38
rs772795989 Chr.8: 85115092-85115092 on Build GRCh38
rs773940416 Chr.8: 85115115-85115115 on Build GRCh38

View Full Product Details