Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 33825290-33825543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDCD6IP
Entrez Gene ID
10015
Gene Name
programmed cell death 6 interacting protein
Gene Aliases
AIP1, ALIX, DRIP4, HP95
UniGene
Hs.475896
Gene Chromosome Location
Chr.3: 33798571-33869707 on Build GRCh38
Transcripts
NM_001162429.2 NM_001256192.1 NM_013374.5 XM_011533252.1 XM_011533253.1 XM_017005488.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4116993 c.603A>T p.L201F Chr.3: 33825327-33825327 on Build GRCh38
2984624 c.581A>T p.Q194L Chr.3: 33825305-33825305 on Build GRCh38
1422459 c.595delT p.L201fs*1 Chr.3: 33825319-33825319 on Build GRCh38
3767406 c.616+9A>G p.? Chr.3: 33825349-33825349 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745761526 Chr.3: 33825385-33825385 on Build GRCh38
rs561065343 Chr.3: 33825410-33825410 on Build GRCh38
rs760399252 Chr.3: 33825327-33825327 on Build GRCh38
rs995382878 Chr.3: 33825456-33825456 on Build GRCh38
rs781406540 Chr.3: 33825373-33825373 on Build GRCh38
rs12495243 Chr.3: 33825437-33825437 on Build GRCh38
rs889773289 Chr.3: 33825481-33825481 on Build GRCh38
rs752847503 Chr.3: 33825366-33825378 on Build GRCh38
rs907002077 Chr.3: 33825365-33825365 on Build GRCh38
rs781474699 Chr.3: 33825319-33825319 on Build GRCh38
rs370551027 Chr.3: 33825379-33825379 on Build GRCh38
rs1028049470 Chr.3: 33825440-33825440 on Build GRCh38
rs1002669975 Chr.3: 33825372-33825372 on Build GRCh38
rs372388603 Chr.3: 33825350-33825350 on Build GRCh38
rs780253322 Chr.3: 33825366-33825366 on Build GRCh38
rs756828896 Chr.3: 33825347-33825347 on Build GRCh38
rs764273212 Chr.3: 33825354-33825354 on Build GRCh38
rs772114423 Chr.3: 33825392-33825392 on Build GRCh38
rs894395477 Chr.3: 33825427-33825427 on Build GRCh38
rs201621483 Chr.3: 33825496-33825496 on Build GRCh38
rs763892573 Chr.3: 33825334-33825334 on Build GRCh38
rs1028305435 Chr.3: 33825476-33825476 on Build GRCh38
rs755180669 Chr.3: 33825367-33825367 on Build GRCh38
rs12488699 Chr.3: 33825376-33825376 on Build GRCh38
rs753336839 Chr.3: 33825341-33825341 on Build GRCh38
rs939864630 Chr.3: 33825432-33825432 on Build GRCh38
rs528190811 Chr.3: 33825411-33825411 on Build GRCh38
rs773388906 Chr.3: 33825382-33825382 on Build GRCh38
rs945770519 Chr.3: 33825312-33825312 on Build GRCh38
rs1036794412 Chr.3: 33825434-33825434 on Build GRCh38
rs373719719 Chr.3: 33825348-33825348 on Build GRCh38

View Full Product Details