Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 39041942-39042211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NHLRC3
Entrez Gene ID
387921
Gene Name
NHL repeat containing 3
Gene Aliases
-
UniGene
Hs.507783
Gene Chromosome Location
Chr.13: 39038311-39050109 on Build GRCh38
Transcripts
NM_001012754.3 NM_001017370.2 NR_073109.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
947336 c.410A>G p.K137R Chr.13: 39042129-39042129 on Build GRCh38
4706841 c.419C>A p.S140Y Chr.13: 39042138-39042138 on Build GRCh38
1366846 c.406delA p.K137fs*22 Chr.13: 39042125-39042125 on Build GRCh38
5645836 c.432T>C p.L144L Chr.13: 39042151-39042151 on Build GRCh38
4706842 c.453A>G p.P151P Chr.13: 39042172-39042172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770694753 Chr.13: 39042094-39042099 on Build GRCh38
rs774005669 Chr.13: 39042061-39042061 on Build GRCh38
rs759568178 Chr.13: 39042084-39042084 on Build GRCh38
rs903012102 Chr.13: 39042115-39042115 on Build GRCh38
rs193193061 Chr.13: 39042068-39042068 on Build GRCh38
rs1055167296 Chr.13: 39042184-39042184 on Build GRCh38
rs373055809 Chr.13: 39042165-39042165 on Build GRCh38
rs749377019 Chr.13: 39042051-39042051 on Build GRCh38
rs371803735 Chr.13: 39042117-39042117 on Build GRCh38
rs779689901 Chr.13: 39042135-39042135 on Build GRCh38
rs763060878 Chr.13: 39042092-39042092 on Build GRCh38
rs566693779 Chr.13: 39042060-39042060 on Build GRCh38
rs775725384 Chr.13: 39042090-39042090 on Build GRCh38
rs551782375 Chr.13: 39042152-39042152 on Build GRCh38
rs375131866 Chr.13: 39042146-39042146 on Build GRCh38
rs965418886 Chr.13: 39041995-39041995 on Build GRCh38
rs931363641 Chr.13: 39042005-39042005 on Build GRCh38
rs751863983 Chr.13: 39042105-39042105 on Build GRCh38
rs1001881201 Chr.13: 39042162-39042162 on Build GRCh38
rs200562270 Chr.13: 39042140-39042140 on Build GRCh38
rs745775817 Chr.13: 39042175-39042176 on Build GRCh38
rs762385071 Chr.13: 39042111-39042111 on Build GRCh38
rs369490569 Chr.13: 39042155-39042155 on Build GRCh38
rs546830284 Chr.13: 39042003-39042003 on Build GRCh38
rs926690436 Chr.13: 39041985-39041985 on Build GRCh38
rs770030154 Chr.13: 39042089-39042089 on Build GRCh38
rs774194331 Chr.13: 39042139-39042139 on Build GRCh38
rs896510322 Chr.13: 39042054-39042054 on Build GRCh38
rs912581589 Chr.13: 39041978-39041978 on Build GRCh38
rs780179324 Chr.13: 39041983-39041983 on Build GRCh38
rs748186290 Chr.13: 39042067-39042067 on Build GRCh38
rs749974527 Chr.13: 39042122-39042122 on Build GRCh38
rs149579598 Chr.13: 39042171-39042171 on Build GRCh38
rs748861996 Chr.13: 39042057-39042057 on Build GRCh38
rs747843309 Chr.13: 39042174-39042174 on Build GRCh38
rs201377397 Chr.13: 39042102-39042102 on Build GRCh38
rs755725590 Chr.13: 39042128-39042128 on Build GRCh38
rs748395514 Chr.13: 39042056-39042059 on Build GRCh38
rs975794864 Chr.13: 39041998-39041998 on Build GRCh38
rs776709688 Chr.13: 39042077-39042077 on Build GRCh38
rs764575525 Chr.13: 39042098-39042098 on Build GRCh38

View Full Product Details