Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4899251-4899500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNE
Entrez Gene ID
1145
Gene Name
cholinergic receptor nicotinic epsilon subunit
Gene Aliases
ACHRE, CMS1D, CMS1E, CMS2A, CMS4A, CMS4B, CMS4C, FCCMS, SCCMS
UniGene
Hs.654535
Gene Chromosome Location
Chr.17: 4897769-4905019 on Build GRCh38
Transcripts
NM_000080.3 XM_017024115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7323571 c.1114G>C p.A372P Chr.17: 4899303-4899303 on Build GRCh38
4651312 c.1146G>T p.E382D Chr.17: 4899271-4899271 on Build GRCh38
6501036 c.1052C>G p.P351R Chr.17: 4899365-4899365 on Build GRCh38
3787359 c.1004C>T p.T335I Chr.17: 4899496-4899496 on Build GRCh38
6405486 c.1044G>T p.E348D Chr.17: 4899373-4899373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748353818 Chr.17: 4899391-4899391 on Build GRCh38
rs775550701 Chr.17: 4899422-4899422 on Build GRCh38
rs912512352 Chr.17: 4899302-4899302 on Build GRCh38
rs749376670 Chr.17: 4899409-4899409 on Build GRCh38
rs114454383 Chr.17: 4899483-4899483 on Build GRCh38
rs763837110 Chr.17: 4899315-4899315 on Build GRCh38
rs932632375 Chr.17: 4899290-4899290 on Build GRCh38
rs930096641 Chr.17: 4899278-4899278 on Build GRCh38
rs932032926 Chr.17: 4899326-4899345 on Build GRCh38
rs773187552 Chr.17: 4899342-4899342 on Build GRCh38
rs998988224 Chr.17: 4899339-4899339 on Build GRCh38
rs1046859320 Chr.17: 4899388-4899388 on Build GRCh38
rs781453095 Chr.17: 4899336-4899336 on Build GRCh38
rs372131102 Chr.17: 4899451-4899451 on Build GRCh38
rs879253723 Chr.17: 4899470-4899470 on Build GRCh38
rs954120463 Chr.17: 4899351-4899351 on Build GRCh38
rs2075763 Chr.17: 4899390-4899390 on Build GRCh38
rs747368522 Chr.17: 4899327-4899327 on Build GRCh38
rs974390828 Chr.17: 4899370-4899370 on Build GRCh38
rs907495464 Chr.17: 4899340-4899340 on Build GRCh38
rs886053123 Chr.17: 4899384-4899385 on Build GRCh38
rs768012683 Chr.17: 4899426-4899427 on Build GRCh38
rs928738409 Chr.17: 4899389-4899389 on Build GRCh38
rs1026988403 Chr.17: 4899347-4899347 on Build GRCh38
rs777843330 Chr.17: 4899402-4899402 on Build GRCh38
rs367944415 Chr.17: 4899435-4899435 on Build GRCh38
rs753193995 Chr.17: 4899430-4899431 on Build GRCh38
rs886053125 Chr.17: 4899470-4899470 on Build GRCh38
rs886042727 Chr.17: 4899337-4899337 on Build GRCh38
rs755351598 Chr.17: 4899425-4899426 on Build GRCh38
rs759577243 Chr.17: 4899312-4899312 on Build GRCh38
rs139922367 Chr.17: 4899481-4899481 on Build GRCh38
rs922032533 Chr.17: 4899313-4899336 on Build GRCh38
rs780327519 Chr.17: 4899324-4899324 on Build GRCh38
rs778313885 Chr.17: 4899319-4899319 on Build GRCh38
rs761361955 Chr.17: 4899386-4899387 on Build GRCh38
rs1047142793 Chr.17: 4899295-4899295 on Build GRCh38
rs769811534 Chr.17: 4899306-4899306 on Build GRCh38
rs1055824191 Chr.17: 4899393-4899393 on Build GRCh38
rs756306460 Chr.17: 4899398-4899398 on Build GRCh38
rs371919434 Chr.17: 4899314-4899314 on Build GRCh38
rs746986185 Chr.17: 4899433-4899433 on Build GRCh38
rs753495337 Chr.17: 4899317-4899317 on Build GRCh38
rs886043239 Chr.17: 4899324-4899324 on Build GRCh38
rs764744220 Chr.17: 4899362-4899362 on Build GRCh38
rs755463469 Chr.17: 4899460-4899460 on Build GRCh38
rs534346599 Chr.17: 4899423-4899423 on Build GRCh38
rs372862723 Chr.17: 4899298-4899298 on Build GRCh38
rs368940455 Chr.17: 4899280-4899280 on Build GRCh38
rs369641727 Chr.17: 4899478-4899478 on Build GRCh38
rs778130828 Chr.17: 4899476-4899476 on Build GRCh38
rs772143528 Chr.17: 4899346-4899346 on Build GRCh38
rs755391448 Chr.17: 4899334-4899334 on Build GRCh38
rs144047383 Chr.17: 4899468-4899468 on Build GRCh38
rs547989166 Chr.17: 4899276-4899276 on Build GRCh38
rs919759171 Chr.17: 4899277-4899277 on Build GRCh38
rs767374241 Chr.17: 4899313-4899313 on Build GRCh38
rs756786529 Chr.17: 4899275-4899275 on Build GRCh38
rs752226476 Chr.17: 4899337-4899338 on Build GRCh38
rs34440658 Chr.17: 4899409-4899409 on Build GRCh38
rs964034772 Chr.17: 4899366-4899366 on Build GRCh38
rs749936691 Chr.17: 4899369-4899369 on Build GRCh38
rs376093621 Chr.17: 4899279-4899279 on Build GRCh38
rs1053542785 Chr.17: 4899417-4899417 on Build GRCh38
rs752110136 Chr.17: 4899382-4899382 on Build GRCh38
rs763847983 Chr.17: 4899357-4899357 on Build GRCh38
rs942583372 Chr.17: 4899274-4899274 on Build GRCh38
rs192195094 Chr.17: 4899341-4899341 on Build GRCh38
rs786204773 Chr.17: 4899386-4899386 on Build GRCh38
rs575066645 Chr.17: 4899454-4899454 on Build GRCh38
rs775638720 Chr.17: 4899352-4899352 on Build GRCh38
rs756571784 Chr.17: 4899469-4899469 on Build GRCh38
rs768869197 Chr.17: 4899296-4899296 on Build GRCh38
rs986495754 Chr.17: 4899349-4899349 on Build GRCh38
rs974582156 Chr.17: 4899344-4899344 on Build GRCh38
rs764991750 Chr.17: 4899392-4899393 on Build GRCh38
rs553626728 Chr.17: 4899291-4899291 on Build GRCh38
rs886053124 Chr.17: 4899397-4899398 on Build GRCh38
rs765913515 Chr.17: 4899455-4899455 on Build GRCh38
rs766856649 Chr.17: 4899377-4899377 on Build GRCh38
rs777995077 Chr.17: 4899437-4899438 on Build GRCh38
rs774315307 Chr.17: 4899309-4899309 on Build GRCh38
rs1019584942 Chr.17: 4899482-4899482 on Build GRCh38
rs761459515 Chr.17: 4899441-4899441 on Build GRCh38
rs776299084 Chr.17: 4899440-4899440 on Build GRCh38
rs916370170 Chr.17: 4899419-4899419 on Build GRCh38
rs906186521 Chr.17: 4899443-4899443 on Build GRCh38
rs745744138 Chr.17: 4899343-4899343 on Build GRCh38
rs757968612 Chr.17: 4899375-4899375 on Build GRCh38
rs897318956 Chr.17: 4899465-4899465 on Build GRCh38
rs767865852 Chr.17: 4899462-4899462 on Build GRCh38
rs750889051 Chr.17: 4899322-4899322 on Build GRCh38
rs755303686 Chr.17: 4899385-4899385 on Build GRCh38
rs749442672 Chr.17: 4899443-4899444 on Build GRCh38
rs760538111 Chr.17: 4899356-4899356 on Build GRCh38
rs764790153 Chr.17: 4899446-4899446 on Build GRCh38
rs752020505 Chr.17: 4899459-4899459 on Build GRCh38
rs530015297 Chr.17: 4899414-4899414 on Build GRCh38
rs558633663 Chr.17: 4899361-4899361 on Build GRCh38
rs1053507936 Chr.17: 4899332-4899332 on Build GRCh38

View Full Product Details