Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143331039-143331310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN1
Entrez Gene ID
1180
Gene Name
chloride voltage-gated channel 1
Gene Aliases
CLC1
UniGene
Hs.121483
Gene Chromosome Location
Chr.7: 143316126-143352008 on Build GRCh38
Transcripts
NM_000083.2 NR_046453.1 XM_017011739.1 XM_017011740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
223218 c.1031A>T p.D344V Chr.7: 143331283-143331283 on Build GRCh38
1086741 c.1013G>A p.R338Q Chr.7: 143331265-143331265 on Build GRCh38
1448834 c.1012C>T p.R338* Chr.7: 143331264-143331264 on Build GRCh38
1086743 c.1049C>T p.A350V Chr.7: 143331301-143331301 on Build GRCh38
374616 c.980-2A>T p.? Chr.7: 143331230-143331230 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375215812 Chr.7: 143331236-143331236 on Build GRCh38
rs762636438 Chr.7: 143331194-143331194 on Build GRCh38
rs779035063 Chr.7: 143331242-143331242 on Build GRCh38
rs554038768 Chr.7: 143331193-143331193 on Build GRCh38
rs947551488 Chr.7: 143331091-143331091 on Build GRCh38
rs763744958 Chr.7: 143331203-143331203 on Build GRCh38
rs80356703 Chr.7: 143331265-143331265 on Build GRCh38
rs73456424 Chr.7: 143331097-143331097 on Build GRCh38
rs769517790 Chr.7: 143331268-143331268 on Build GRCh38
rs759761559 Chr.7: 143331264-143331264 on Build GRCh38
rs780421370 Chr.7: 143331235-143331235 on Build GRCh38
rs781765456 Chr.7: 143331212-143331212 on Build GRCh38
rs765034718 Chr.7: 143331192-143331192 on Build GRCh38
rs367699805 Chr.7: 143331205-143331205 on Build GRCh38
rs750881030 Chr.7: 143331215-143331215 on Build GRCh38
rs371875551 Chr.7: 143331206-143331206 on Build GRCh38
rs1036073626 Chr.7: 143331115-143331115 on Build GRCh38
rs756454039 Chr.7: 143331220-143331220 on Build GRCh38
rs553008115 Chr.7: 143331152-143331152 on Build GRCh38
rs748233796 Chr.7: 143331248-143331248 on Build GRCh38
rs762751787 Chr.7: 143331283-143331283 on Build GRCh38
rs772084781 Chr.7: 143331252-143331252 on Build GRCh38
rs775333425 Chr.7: 143331277-143331277 on Build GRCh38
rs776457257 Chr.7: 143331259-143331259 on Build GRCh38
rs140664749 Chr.7: 143331239-143331239 on Build GRCh38
rs747549814 Chr.7: 143331077-143331077 on Build GRCh38
rs903248055 Chr.7: 143331098-143331098 on Build GRCh38
rs777059595 Chr.7: 143331166-143331166 on Build GRCh38
rs1002979390 Chr.7: 143331101-143331101 on Build GRCh38
rs893429522 Chr.7: 143331065-143331065 on Build GRCh38

View Full Product Details