Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 154122877-154123134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF26
Entrez Gene ID
26084
Gene Name
Rho guanine nucleotide exchange factor 26
Gene Aliases
CSGEF, HMFN1864, SGEF
UniGene
Hs.240845
Gene Chromosome Location
Chr.3: 154121003-154257827 on Build GRCh38
Transcripts
NM_001251962.1 NM_001251963.1 NM_015595.3 XM_011512672.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4663160 c.998C>T p.S333L Chr.3: 154122990-154122990 on Build GRCh38
4957271 c.890A>T p.E297V Chr.3: 154122882-154122882 on Build GRCh38
4663157 c.938G>A p.R313Q Chr.3: 154122930-154122930 on Build GRCh38
6468578 c.1030C>T p.L344L Chr.3: 154123022-154123022 on Build GRCh38
3004264 c.1057A>T p.K353* Chr.3: 154123049-154123049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762256938 Chr.3: 154123048-154123048 on Build GRCh38
rs572938340 Chr.3: 154122972-154122972 on Build GRCh38
rs574123818 Chr.3: 154123070-154123070 on Build GRCh38
rs770535113 Chr.3: 154122936-154122936 on Build GRCh38
rs758588282 Chr.3: 154122984-154122984 on Build GRCh38
rs199951764 Chr.3: 154123046-154123046 on Build GRCh38
rs779880869 Chr.3: 154123073-154123073 on Build GRCh38
rs539257826 Chr.3: 154122943-154122943 on Build GRCh38
rs750490100 Chr.3: 154122983-154122983 on Build GRCh38
rs370810825 Chr.3: 154123087-154123087 on Build GRCh38
rs1000804071 Chr.3: 154123015-154123015 on Build GRCh38
rs777753661 Chr.3: 154122992-154122992 on Build GRCh38
rs555699610 Chr.3: 154123019-154123019 on Build GRCh38
rs775860770 Chr.3: 154122954-154122954 on Build GRCh38
rs371016104 Chr.3: 154123041-154123041 on Build GRCh38
rs374731357 Chr.3: 154123042-154123042 on Build GRCh38
rs368202288 Chr.3: 154123072-154123072 on Build GRCh38
rs557901822 Chr.3: 154122945-154122945 on Build GRCh38
rs746167643 Chr.3: 154123044-154123046 on Build GRCh38
rs756040991 Chr.3: 154122925-154122925 on Build GRCh38
rs771863470 Chr.3: 154123047-154123049 on Build GRCh38
rs1018525592 Chr.3: 154123068-154123068 on Build GRCh38
rs751867119 Chr.3: 154122987-154122987 on Build GRCh38
rs745378067 Chr.3: 154123017-154123017 on Build GRCh38
rs757286136 Chr.3: 154123006-154123006 on Build GRCh38
rs765675256 Chr.3: 154123050-154123050 on Build GRCh38
rs779359820 Chr.3: 154123043-154123045 on Build GRCh38
rs780991477 Chr.3: 154122911-154122911 on Build GRCh38
rs771641541 Chr.3: 154122952-154122952 on Build GRCh38
rs749200280 Chr.3: 154123000-154123000 on Build GRCh38
rs775268568 Chr.3: 154123091-154123092 on Build GRCh38
rs370035369 Chr.3: 154122912-154122912 on Build GRCh38
rs777096804 Chr.3: 154122980-154122980 on Build GRCh38
rs755952944 Chr.3: 154122988-154122988 on Build GRCh38
rs778432853 Chr.3: 154123007-154123007 on Build GRCh38
rs1052363661 Chr.3: 154123028-154123028 on Build GRCh38
rs750625968 Chr.3: 154122909-154122909 on Build GRCh38
rs371069494 Chr.3: 154123044-154123044 on Build GRCh38
rs752904270 Chr.3: 154123033-154123033 on Build GRCh38
rs764497151 Chr.3: 154122978-154122978 on Build GRCh38
rs899808579 Chr.3: 154122994-154122994 on Build GRCh38
rs540564396 Chr.3: 154122981-154122981 on Build GRCh38
rs375753893 Chr.3: 154123035-154123035 on Build GRCh38
rs749289934 Chr.3: 154122929-154122929 on Build GRCh38
rs372474018 Chr.3: 154123034-154123034 on Build GRCh38
rs891292575 Chr.3: 154122904-154122904 on Build GRCh38
rs777839865 Chr.3: 154122928-154122928 on Build GRCh38
rs766505826 Chr.3: 154122985-154122985 on Build GRCh38
rs7630182 Chr.3: 154123109-154123109 on Build GRCh38
rs746710814 Chr.3: 154123039-154123039 on Build GRCh38
rs535601495 Chr.3: 154122991-154122993 on Build GRCh38
rs373831084 Chr.3: 154123074-154123074 on Build GRCh38
rs747908710 Chr.3: 154122918-154122918 on Build GRCh38
rs544577271 Chr.3: 154123092-154123092 on Build GRCh38

View Full Product Details