Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201704237-201704502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALS2
Entrez Gene ID
57679
Gene Name
ALS2, alsin Rho guanine nucleotide exchange factor
Gene Aliases
ALS2CR6, ALSJ, IAHSP, PLSJ
UniGene
Hs.471096
Gene Chromosome Location
Chr.2: 201700263-201781172 on Build GRCh38
Transcripts
NM_020919.3 XM_006712654.2 XM_006712655.3 XM_011511530.2 XM_017004569.1 XM_017004571.1 XM_017004572.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4090253 c.4831C>T p.R1611W Chr.2: 201704461-201704461 on Build GRCh38
4496673 c.4807C>T p.P1603S Chr.2: 201704485-201704485 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772452966 Chr.2: 201704475-201704475 on Build GRCh38
rs1008860622 Chr.2: 201704438-201704438 on Build GRCh38
rs538300635 Chr.2: 201704320-201704320 on Build GRCh38
rs576039799 Chr.2: 201704348-201704348 on Build GRCh38
rs749049075 Chr.2: 201704434-201704434 on Build GRCh38
rs1014953504 Chr.2: 201704453-201704453 on Build GRCh38
rs968896144 Chr.2: 201704290-201704290 on Build GRCh38
rs369822894 Chr.2: 201704266-201704266 on Build GRCh38
rs370296035 Chr.2: 201704472-201704472 on Build GRCh38
rs543318613 Chr.2: 201704435-201704435 on Build GRCh38
rs1051467623 Chr.2: 201704370-201704370 on Build GRCh38
rs978891342 Chr.2: 201704369-201704369 on Build GRCh38
rs372820505 Chr.2: 201704420-201704420 on Build GRCh38
rs779186036 Chr.2: 201704461-201704461 on Build GRCh38
rs752516794 Chr.2: 201704435-201704436 on Build GRCh38
rs555220239 Chr.2: 201704474-201704474 on Build GRCh38
rs780722382 Chr.2: 201704403-201704403 on Build GRCh38
rs779872914 Chr.2: 201704431-201704431 on Build GRCh38
rs377218529 Chr.2: 201704441-201704441 on Build GRCh38
rs996804556 Chr.2: 201704276-201704276 on Build GRCh38
rs577567911 Chr.2: 201704275-201704275 on Build GRCh38
rs556905437 Chr.2: 201704346-201704346 on Build GRCh38
rs772339699 Chr.2: 201704421-201704421 on Build GRCh38
rs746735907 Chr.2: 201704304-201704304 on Build GRCh38

View Full Product Details