Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43884614-43884883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD5
Entrez Gene ID
84978
Gene Name
FERM domain containing 5
Gene Aliases
-
UniGene
Hs.578544
Gene Chromosome Location
Chr.15: 43870761-44195294 on Build GRCh38
Transcripts
NM_001286490.1 NM_001286491.1 NM_001322949.1 NM_001322950.1 NM_001322951.1 NM_032892.4 NR_104455.1 XM_005254730.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1629570 c.971C>T p.A324V Chr.15: 43884784-43884784 on Build GRCh38
5947375 c.995C>G p.A332G Chr.15: 43884760-43884760 on Build GRCh38
5849246 c.1018G>A p.E340K Chr.15: 43884737-43884737 on Build GRCh38
368061 c.966A>T p.R322R Chr.15: 43884789-43884789 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774105837 Chr.15: 43884739-43884739 on Build GRCh38
rs992324334 Chr.15: 43884721-43884721 on Build GRCh38
rs776687995 Chr.15: 43884844-43884844 on Build GRCh38
rs746323295 Chr.15: 43884783-43884783 on Build GRCh38
rs754699975 Chr.15: 43884691-43884691 on Build GRCh38
rs747170687 Chr.15: 43884713-43884713 on Build GRCh38
rs768934248 Chr.15: 43884843-43884843 on Build GRCh38
rs779254536 Chr.15: 43884704-43884704 on Build GRCh38
rs763523581 Chr.15: 43884798-43884799 on Build GRCh38
rs746295786 Chr.15: 43884705-43884705 on Build GRCh38
rs146504694 Chr.15: 43884738-43884738 on Build GRCh38
rs758771201 Chr.15: 43884805-43884805 on Build GRCh38
rs775725148 Chr.15: 43884711-43884711 on Build GRCh38
rs549809489 Chr.15: 43884723-43884723 on Build GRCh38
rs200347143 Chr.15: 43884744-43884744 on Build GRCh38
rs752411168 Chr.15: 43884756-43884756 on Build GRCh38
rs570129634 Chr.15: 43884637-43884637 on Build GRCh38
rs747362395 Chr.15: 43884832-43884832 on Build GRCh38
rs767353884 Chr.15: 43884749-43884749 on Build GRCh38
rs975188552 Chr.15: 43884671-43884671 on Build GRCh38
rs759404040 Chr.15: 43884748-43884748 on Build GRCh38
rs751439528 Chr.15: 43884689-43884689 on Build GRCh38
rs569734648 Chr.15: 43884762-43884762 on Build GRCh38
rs775926657 Chr.15: 43884703-43884703 on Build GRCh38
rs759052042 Chr.15: 43884795-43884795 on Build GRCh38
rs940663892 Chr.15: 43884790-43884790 on Build GRCh38
rs1054275860 Chr.15: 43884852-43884852 on Build GRCh38
rs762890594 Chr.15: 43884680-43884680 on Build GRCh38
rs538438510 Chr.15: 43884707-43884707 on Build GRCh38
rs573949349 Chr.15: 43884837-43884838 on Build GRCh38
rs372213408 Chr.15: 43884701-43884701 on Build GRCh38
rs966147102 Chr.15: 43884644-43884644 on Build GRCh38
rs779499948 Chr.15: 43884777-43884777 on Build GRCh38
rs763830882 Chr.15: 43884760-43884760 on Build GRCh38
rs1044040858 Chr.15: 43884804-43884804 on Build GRCh38
rs755885560 Chr.15: 43884758-43884758 on Build GRCh38
rs781012870 Chr.15: 43884700-43884700 on Build GRCh38
rs113310192 Chr.15: 43884745-43884745 on Build GRCh38
rs371992236 Chr.15: 43884768-43884768 on Build GRCh38
rs935932370 Chr.15: 43884836-43884836 on Build GRCh38
rs371219015 Chr.15: 43884702-43884702 on Build GRCh38
rs368267913 Chr.15: 43884688-43884688 on Build GRCh38
rs190093187 Chr.15: 43884716-43884716 on Build GRCh38
rs760218825 Chr.15: 43884779-43884779 on Build GRCh38
rs776750597 Chr.15: 43884717-43884717 on Build GRCh38
rs776279443 Chr.15: 43884801-43884803 on Build GRCh38
rs772533424 Chr.15: 43884695-43884695 on Build GRCh38
rs780342108 Chr.15: 43884817-43884817 on Build GRCh38
rs201528357 Chr.15: 43884703-43884703 on Build GRCh38
rs748310829 Chr.15: 43884846-43884846 on Build GRCh38

View Full Product Details