Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 106077712-106077983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf40
Entrez Gene ID
84417
Gene Name
chromosome 2 open reading frame 40
Gene Aliases
ECRG4
UniGene
Hs.43125
Gene Chromosome Location
Chr.2: 106063290-106078155 on Build GRCh38
Transcripts
NM_032411.2 XM_006712799.3 XM_011511993.2 XM_017005106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3564695 c.396G>A p.R132R Chr.2: 106077875-106077875 on Build GRCh38
1185975 c.436G>A p.D146N Chr.2: 106077915-106077915 on Build GRCh38
4667598 c.349G>A p.D117N Chr.2: 106077828-106077828 on Build GRCh38
3564693 c.325C>T p.R109* Chr.2: 106077804-106077804 on Build GRCh38
418643 c.347G>A p.G116D Chr.2: 106077826-106077826 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150617664 Chr.2: 106077827-106077827 on Build GRCh38
rs573510498 Chr.2: 106077780-106077780 on Build GRCh38
rs763765596 Chr.2: 106077735-106077735 on Build GRCh38
rs1024251675 Chr.2: 106077741-106077741 on Build GRCh38
rs373970608 Chr.2: 106077930-106077930 on Build GRCh38
rs117849769 Chr.2: 106077784-106077784 on Build GRCh38
rs1014041011 Chr.2: 106077900-106077900 on Build GRCh38
rs146511853 Chr.2: 106077828-106077828 on Build GRCh38
rs765005910 Chr.2: 106077918-106077920 on Build GRCh38
rs769838546 Chr.2: 106077805-106077805 on Build GRCh38
rs768759203 Chr.2: 106077944-106077944 on Build GRCh38
rs757602564 Chr.2: 106077841-106077841 on Build GRCh38
rs760358092 Chr.2: 106077897-106077897 on Build GRCh38
rs767390809 Chr.2: 106077894-106077894 on Build GRCh38
rs114065772 Chr.2: 106077943-106077943 on Build GRCh38
rs754503202 Chr.2: 106077908-106077908 on Build GRCh38
rs761339700 Chr.2: 106077837-106077839 on Build GRCh38
rs749937699 Chr.2: 106077944-106077945 on Build GRCh38
rs747271289 Chr.2: 106077794-106077794 on Build GRCh38
rs751253545 Chr.2: 106077753-106077753 on Build GRCh38
rs190568883 Chr.2: 106077939-106077939 on Build GRCh38
rs375563861 Chr.2: 106077804-106077804 on Build GRCh38
rs752036758 Chr.2: 106077915-106077915 on Build GRCh38
rs79393517 Chr.2: 106077748-106077749 on Build GRCh38
rs370404898 Chr.2: 106077913-106077913 on Build GRCh38
rs1046357080 Chr.2: 106077771-106077773 on Build GRCh38
rs971214381 Chr.2: 106077758-106077758 on Build GRCh38
rs750023164 Chr.2: 106077779-106077779 on Build GRCh38
rs762759460 Chr.2: 106077811-106077811 on Build GRCh38
rs757655683 Chr.2: 106077928-106077928 on Build GRCh38
rs532790057 Chr.2: 106077848-106077848 on Build GRCh38
rs761697747 Chr.2: 106077820-106077820 on Build GRCh38
rs148850657 Chr.2: 106077885-106077885 on Build GRCh38
rs746108267 Chr.2: 106077934-106077934 on Build GRCh38
rs769945889 Chr.2: 106077864-106077864 on Build GRCh38
rs200593375 Chr.2: 106077896-106077896 on Build GRCh38
rs376920426 Chr.2: 106077906-106077906 on Build GRCh38
rs762295020 Chr.2: 106077953-106077953 on Build GRCh38
rs115609696 Chr.2: 106077814-106077814 on Build GRCh38
rs779524318 Chr.2: 106077783-106077783 on Build GRCh38
rs903923412 Chr.2: 106077870-106077870 on Build GRCh38
rs1015609470 Chr.2: 106077819-106077819 on Build GRCh38
rs867836258 Chr.2: 106077886-106077886 on Build GRCh38
rs764563678 Chr.2: 106077914-106077914 on Build GRCh38
rs774472949 Chr.2: 106077945-106077945 on Build GRCh38
rs780855470 Chr.2: 106077776-106077776 on Build GRCh38
rs971309618 Chr.2: 106077917-106077917 on Build GRCh38
rs193020612 Chr.2: 106077787-106077787 on Build GRCh38
rs143187713 Chr.2: 106077813-106077813 on Build GRCh38
rs750078620 Chr.2: 106077824-106077824 on Build GRCh38
rs764905916 Chr.2: 106077840-106077840 on Build GRCh38
rs1042538022 Chr.2: 106077857-106077857 on Build GRCh38
rs765930940 Chr.2: 106077905-106077905 on Build GRCh38
rs774333431 Chr.2: 106077879-106077879 on Build GRCh38
rs780186072 Chr.2: 106077942-106077942 on Build GRCh38
rs778201695 Chr.2: 106077838-106077838 on Build GRCh38
rs895130323 Chr.2: 106077898-106077898 on Build GRCh38
rs111427569 Chr.2: 106077763-106077763 on Build GRCh38
rs185643114 Chr.2: 106077835-106077835 on Build GRCh38
rs749261815 Chr.2: 106077874-106077874 on Build GRCh38
rs962893833 Chr.2: 106077952-106077952 on Build GRCh38
rs76863264 Chr.2: 106077884-106077884 on Build GRCh38
rs772018594 Chr.2: 106077955-106077955 on Build GRCh38
rs377709469 Chr.2: 106077772-106077772 on Build GRCh38
rs767339420 Chr.2: 106077823-106077823 on Build GRCh38
rs775382887 Chr.2: 106077873-106077873 on Build GRCh38
rs771071087 Chr.2: 106077800-106077800 on Build GRCh38

View Full Product Details