Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92180902-92181147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB3
Entrez Gene ID
22849
Gene Name
cytoplasmic polyadenylation element binding protein 3
Gene Aliases
-
UniGene
Hs.131683
Gene Chromosome Location
Chr.10: 92046692-92291118 on Build GRCh38
Transcripts
NM_001178137.1 NM_014912.4 XM_005269630.4 XM_006717714.3 XM_006717715.3 XM_011539514.2 XM_011539515.2 XM_011539517.2 XM_011539518.2 XM_011539519.2 XM_011539520.2 XM_011539522.2 XM_011539523.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4448674 c.1166-1G>A p.? Chr.10: 92181020-92181020 on Build GRCh38
4869898 c.1111A>C p.N371H Chr.10: 92181005-92181005 on Build GRCh38
3981198 c.1097-9A>G p.? Chr.10: 92181028-92181028 on Build GRCh38
3441522 c.1184T>C p.F395S Chr.10: 92181001-92181001 on Build GRCh38
3981197 c.1166-9A>G p.? Chr.10: 92181028-92181028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764699604 Chr.10: 92180993-92180993 on Build GRCh38
rs780742695 Chr.10: 92181024-92181028 on Build GRCh38
rs200036143 Chr.10: 92180927-92180927 on Build GRCh38
rs373130171 Chr.10: 92180928-92180928 on Build GRCh38
rs143457838 Chr.10: 92181014-92181014 on Build GRCh38
rs746837928 Chr.10: 92181048-92181048 on Build GRCh38
rs748177894 Chr.10: 92181059-92181059 on Build GRCh38
rs916115091 Chr.10: 92181124-92181124 on Build GRCh38
rs888165525 Chr.10: 92181047-92181047 on Build GRCh38
rs769616822 Chr.10: 92181063-92181063 on Build GRCh38
rs1027708855 Chr.10: 92181102-92181102 on Build GRCh38
rs779113763 Chr.10: 92181016-92181016 on Build GRCh38
rs971765139 Chr.10: 92181101-92181101 on Build GRCh38
rs769292574 Chr.10: 92180940-92180940 on Build GRCh38
rs780201270 Chr.10: 92181045-92181045 on Build GRCh38
rs137908945 Chr.10: 92181017-92181017 on Build GRCh38
rs140308172 Chr.10: 92180974-92180974 on Build GRCh38
rs762736647 Chr.10: 92180948-92180948 on Build GRCh38
rs375964350 Chr.10: 92181057-92181057 on Build GRCh38
rs772586937 Chr.10: 92180944-92180944 on Build GRCh38
rs375966304 Chr.10: 92181015-92181015 on Build GRCh38
rs766825880 Chr.10: 92181068-92181068 on Build GRCh38
rs372490000 Chr.10: 92181052-92181052 on Build GRCh38
rs1055388296 Chr.10: 92181042-92181042 on Build GRCh38
rs766102129 Chr.10: 92180954-92180954 on Build GRCh38
rs1005722918 Chr.10: 92181067-92181067 on Build GRCh38
rs1036776637 Chr.10: 92181119-92181119 on Build GRCh38
rs991633593 Chr.10: 92181109-92181109 on Build GRCh38
rs552163049 Chr.10: 92180933-92180933 on Build GRCh38
rs951851403 Chr.10: 92181107-92181107 on Build GRCh38
rs758694732 Chr.10: 92181028-92181028 on Build GRCh38
rs961898419 Chr.10: 92181088-92181088 on Build GRCh38
rs761235285 Chr.10: 92180938-92180938 on Build GRCh38
rs879654962 Chr.10: 92181087-92181087 on Build GRCh38
rs753165597 Chr.10: 92180973-92180973 on Build GRCh38
rs11186846 Chr.10: 92181104-92181104 on Build GRCh38

View Full Product Details