Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101353135-101353408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTK
Entrez Gene ID
695
Gene Name
Bruton tyrosine kinase
Gene Aliases
AGMX1, AT, ATK, BPK, IMD1, PSCTK1, XLA
UniGene
Hs.159494
Gene Chromosome Location
Chr.X: 101349447-101390796 on Build GRCh38
Transcripts
NM_000061.2 NM_001287344.1 NM_001287345.1

SNP Detail

SNP ID Chromosome Location
rs782784765 Chr.X: 101353364-101353364 on Build GRCh38
rs781930404 Chr.X: 101353279-101353279 on Build GRCh38
rs200762027 Chr.X: 101353167-101353167 on Build GRCh38
rs372424571 Chr.X: 101353170-101353170 on Build GRCh38
rs782035073 Chr.X: 101353366-101353366 on Build GRCh38
rs128621209 Chr.X: 101353264-101353264 on Build GRCh38
rs782732287 Chr.X: 101353369-101353369 on Build GRCh38
rs1010302002 Chr.X: 101353176-101353176 on Build GRCh38
rs781882779 Chr.X: 101353383-101353383 on Build GRCh38
rs782244933 Chr.X: 101353249-101353249 on Build GRCh38
rs782134435 Chr.X: 101353362-101353362 on Build GRCh38
rs128621208 Chr.X: 101353282-101353282 on Build GRCh38
rs128621206 Chr.X: 101353336-101353336 on Build GRCh38
rs782609610 Chr.X: 101353189-101353189 on Build GRCh38
rs886039321 Chr.X: 101353342-101353342 on Build GRCh38
rs128621207 Chr.X: 101353329-101353329 on Build GRCh38
rs782503550 Chr.X: 101353218-101353218 on Build GRCh38
rs1135363 Chr.X: 101353203-101353203 on Build GRCh38
rs3027646 Chr.X: 101353220-101353220 on Build GRCh38
rs782493736 Chr.X: 101353379-101353379 on Build GRCh38
rs782211137 Chr.X: 101353290-101353290 on Build GRCh38
rs782046439 Chr.X: 101353368-101353368 on Build GRCh38
rs1057287168 Chr.X: 101353162-101353162 on Build GRCh38
rs128622211 Chr.X: 101353196-101353196 on Build GRCh38
rs781985424 Chr.X: 101353349-101353349 on Build GRCh38
rs782389631 Chr.X: 101353301-101353301 on Build GRCh38
rs128621210 Chr.X: 101353213-101353213 on Build GRCh38
rs782283931 Chr.X: 101353242-101353242 on Build GRCh38
rs781829443 Chr.X: 101353377-101353377 on Build GRCh38

View Full Product Details