Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50640331-50640604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPKAPK3
Entrez Gene ID
7867
Gene Name
mitogen-activated protein kinase-activated protein kinase 3
Gene Aliases
3PK, MAPKAP-K3, MAPKAP3, MAPKAPK-3, MK-3
UniGene
Hs.234521
Gene Chromosome Location
Chr.3: 50611862-50649297 on Build GRCh38
Transcripts
NM_001243925.1 NM_001243926.1 NM_004635.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5631310 c.339T>C p.C113C Chr.3: 50640485-50640485 on Build GRCh38
7175914 c.348C>T p.I116I Chr.3: 50640494-50640494 on Build GRCh38
1046543 c.316A>G p.N106D Chr.3: 50640462-50640462 on Build GRCh38
2851527 c.296G>A p.C99Y Chr.3: 50640442-50640442 on Build GRCh38
3824233 c.323A>C p.H108P Chr.3: 50640469-50640469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777383707 Chr.3: 50640391-50640391 on Build GRCh38
rs575368406 Chr.3: 50640417-50640417 on Build GRCh38
rs748745169 Chr.3: 50640406-50640406 on Build GRCh38
rs769899324 Chr.3: 50640465-50640465 on Build GRCh38
rs767380329 Chr.3: 50640371-50640371 on Build GRCh38
rs143980632 Chr.3: 50640426-50640426 on Build GRCh38
rs773395153 Chr.3: 50640471-50640471 on Build GRCh38
rs751453467 Chr.3: 50640355-50640355 on Build GRCh38
rs1009293754 Chr.3: 50640511-50640511 on Build GRCh38
rs755897736 Chr.3: 50640538-50640538 on Build GRCh38
rs764577115 Chr.3: 50640360-50640360 on Build GRCh38
rs752419500 Chr.3: 50640388-50640388 on Build GRCh38
rs759497061 Chr.3: 50640481-50640481 on Build GRCh38
rs372266899 Chr.3: 50640539-50640539 on Build GRCh38
rs756725967 Chr.3: 50640544-50640544 on Build GRCh38
rs141282634 Chr.3: 50640373-50640373 on Build GRCh38
rs868088281 Chr.3: 50640533-50640533 on Build GRCh38
rs893542590 Chr.3: 50640507-50640507 on Build GRCh38
rs1020729854 Chr.3: 50640532-50640532 on Build GRCh38
rs752509312 Chr.3: 50640522-50640522 on Build GRCh38
rs77366433 Chr.3: 50640433-50640433 on Build GRCh38
rs540276598 Chr.3: 50640550-50640550 on Build GRCh38
rs762952384 Chr.3: 50640475-50640475 on Build GRCh38
rs539709580 Chr.3: 50640425-50640425 on Build GRCh38
rs776935311 Chr.3: 50640446-50640446 on Build GRCh38
rs867974734 Chr.3: 50640467-50640467 on Build GRCh38
rs1029588764 Chr.3: 50640504-50640504 on Build GRCh38
rs953995018 Chr.3: 50640567-50640567 on Build GRCh38
rs748422112 Chr.3: 50640455-50640455 on Build GRCh38
rs1032039030 Chr.3: 50640496-50640496 on Build GRCh38
rs558117363 Chr.3: 50640479-50640479 on Build GRCh38
rs376848184 Chr.3: 50640545-50640545 on Build GRCh38
rs755805527 Chr.3: 50640390-50640390 on Build GRCh38
rs1028156635 Chr.3: 50640563-50640563 on Build GRCh38
rs767468360 Chr.3: 50640503-50640503 on Build GRCh38
rs766209026 Chr.3: 50640353-50640353 on Build GRCh38
rs200955406 Chr.3: 50640480-50640480 on Build GRCh38
rs608961 Chr.3: 50640570-50640570 on Build GRCh38
rs754340615 Chr.3: 50640518-50640518 on Build GRCh38

View Full Product Details