Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28780766-28781035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS9
Entrez Gene ID
55756
Gene Name
integrator complex subunit 9
Gene Aliases
CPSF2L, INT9, RC74
UniGene
Hs.162397
Gene Chromosome Location
Chr.8: 28767658-28890226 on Build GRCh38
Transcripts
NM_001145159.2 NM_001172562.1 NM_018250.3 NR_026826.2 XM_006716356.2 XM_006716357.3 XM_011544573.1 XM_011544574.1 XM_011544575.2 XM_011544576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1098934 c.1165C>T p.Q389* Chr.8: 28780928-28780928 on Build GRCh38
1098935 c.1144G>T p.D382Y Chr.8: 28780949-28780949 on Build GRCh38
5874529 c.1211A>G p.D404G Chr.8: 28780882-28780882 on Build GRCh38
1098936 c.1124A>T p.H375L Chr.8: 28780969-28780969 on Build GRCh38
6358655 c.1244C>G p.S415C Chr.8: 28780849-28780849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750163507 Chr.8: 28780952-28780952 on Build GRCh38
rs970600098 Chr.8: 28780909-28780909 on Build GRCh38
rs377391616 Chr.8: 28780848-28780848 on Build GRCh38
rs752918898 Chr.8: 28780887-28780887 on Build GRCh38
rs370384415 Chr.8: 28780906-28780906 on Build GRCh38
rs770846836 Chr.8: 28780855-28780855 on Build GRCh38
rs771987070 Chr.8: 28780937-28780937 on Build GRCh38
rs762934236 Chr.8: 28781002-28781003 on Build GRCh38
rs749661745 Chr.8: 28781003-28781003 on Build GRCh38
rs756371472 Chr.8: 28780891-28780891 on Build GRCh38
rs371334216 Chr.8: 28780949-28780949 on Build GRCh38
rs548025826 Chr.8: 28780881-28780881 on Build GRCh38
rs760766584 Chr.8: 28780940-28780940 on Build GRCh38
rs771652177 Chr.8: 28780921-28780921 on Build GRCh38
rs746547645 Chr.8: 28780825-28780825 on Build GRCh38
rs149685676 Chr.8: 28780938-28780938 on Build GRCh38
rs575319275 Chr.8: 28780956-28780956 on Build GRCh38
rs759725411 Chr.8: 28780883-28780883 on Build GRCh38
rs767014548 Chr.8: 28780874-28780874 on Build GRCh38
rs931625681 Chr.8: 28780944-28780944 on Build GRCh38
rs201577112 Chr.8: 28781007-28781007 on Build GRCh38
rs776186633 Chr.8: 28780831-28780831 on Build GRCh38
rs751559479 Chr.8: 28780960-28780960 on Build GRCh38
rs1000238374 Chr.8: 28780869-28780869 on Build GRCh38
rs374478546 Chr.8: 28780953-28780953 on Build GRCh38
rs771481068 Chr.8: 28781004-28781004 on Build GRCh38
rs764872038 Chr.8: 28780892-28780892 on Build GRCh38
rs199637042 Chr.8: 28781005-28781005 on Build GRCh38
rs111650232 Chr.8: 28780824-28780824 on Build GRCh38
rs759675081 Chr.8: 28780932-28780932 on Build GRCh38
rs758334285 Chr.8: 28780800-28780800 on Build GRCh38
rs770115032 Chr.8: 28780841-28780841 on Build GRCh38
rs754554220 Chr.8: 28780907-28780907 on Build GRCh38
rs780016544 Chr.8: 28780902-28780902 on Build GRCh38
rs747700535 Chr.8: 28780916-28780916 on Build GRCh38
rs57302168 Chr.8: 28780796-28780796 on Build GRCh38
rs772855738 Chr.8: 28780946-28780946 on Build GRCh38
rs202065556 Chr.8: 28780901-28780901 on Build GRCh38
rs150219151 Chr.8: 28780852-28780852 on Build GRCh38
rs141502256 Chr.8: 28780983-28780983 on Build GRCh38
rs34731600 Chr.8: 28780986-28780986 on Build GRCh38
rs1009701451 Chr.8: 28780951-28780951 on Build GRCh38
rs775598012 Chr.8: 28780930-28780931 on Build GRCh38
rs754925342 Chr.8: 28780962-28780962 on Build GRCh38
rs777518238 Chr.8: 28780995-28780995 on Build GRCh38
rs532880369 Chr.8: 28780801-28780804 on Build GRCh38
rs777947693 Chr.8: 28780920-28780920 on Build GRCh38
rs767847217 Chr.8: 28780886-28780886 on Build GRCh38
rs775059997 Chr.8: 28780931-28780931 on Build GRCh38
rs771530190 Chr.8: 28780926-28780926 on Build GRCh38
rs34879077 Chr.8: 28780908-28780908 on Build GRCh38
rs200884133 Chr.8: 28780981-28780981 on Build GRCh38
rs141006653 Chr.8: 28780835-28780835 on Build GRCh38
rs747123765 Chr.8: 28780808-28780809 on Build GRCh38

View Full Product Details