Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 152651607-152651858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRQ
Entrez Gene ID
55879
Gene Name
gamma-aminobutyric acid type A receptor theta subunit
Gene Aliases
THETA
UniGene
Hs.283081
Gene Chromosome Location
Chr.X: 152637895-152659827 on Build GRCh38
Transcripts
NM_018558.3 XM_011531184.2

SNP Detail

SNP ID Chromosome Location
rs887979681 Chr.X: 152651776-152651776 on Build GRCh38
rs782737720 Chr.X: 152651831-152651831 on Build GRCh38
rs781994330 Chr.X: 152651777-152651777 on Build GRCh38
rs782708286 Chr.X: 152651783-152651783 on Build GRCh38
rs146427374 Chr.X: 152651750-152651750 on Build GRCh38
rs782598556 Chr.X: 152651719-152651719 on Build GRCh38
rs782349381 Chr.X: 152651806-152651806 on Build GRCh38
rs782695736 Chr.X: 152651690-152651690 on Build GRCh38
rs147876428 Chr.X: 152651751-152651751 on Build GRCh38
rs145128065 Chr.X: 152651819-152651819 on Build GRCh38
rs782486010 Chr.X: 152651715-152651715 on Build GRCh38
rs370519346 Chr.X: 152651821-152651821 on Build GRCh38
rs781897245 Chr.X: 152651697-152651697 on Build GRCh38
rs782675594 Chr.X: 152651730-152651730 on Build GRCh38
rs898089439 Chr.X: 152651835-152651835 on Build GRCh38
rs782332148 Chr.X: 152651728-152651728 on Build GRCh38
rs142261040 Chr.X: 152651729-152651729 on Build GRCh38
rs782050509 Chr.X: 152651712-152651712 on Build GRCh38
rs367579214 Chr.X: 152651714-152651714 on Build GRCh38
rs74350629 Chr.X: 152651657-152651657 on Build GRCh38
rs782057587 Chr.X: 152651700-152651700 on Build GRCh38
rs781863240 Chr.X: 152651820-152651820 on Build GRCh38
rs781812649 Chr.X: 152651812-152651812 on Build GRCh38
rs782062972 Chr.X: 152651638-152651638 on Build GRCh38
rs782112897 Chr.X: 152651822-152651822 on Build GRCh38
rs782106419 Chr.X: 152651800-152651800 on Build GRCh38
rs949309487 Chr.X: 152651712-152651712 on Build GRCh38
rs1036492731 Chr.X: 152651830-152651830 on Build GRCh38
rs782162359 Chr.X: 152651818-152651818 on Build GRCh38
rs913826191 Chr.X: 152651669-152651669 on Build GRCh38
rs373794425 Chr.X: 152651703-152651703 on Build GRCh38

View Full Product Details