Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38952263-38952514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHA2
Entrez Gene ID
59339
Gene Name
pleckstrin homology domain containing A2
Gene Aliases
TAPP2
UniGene
Hs.369123
Gene Chromosome Location
Chr.8: 38901235-38973912 on Build GRCh38
Transcripts
NM_021623.1 XM_011544605.2 XM_011544606.2 XM_011544607.2 XM_017013718.1 XM_017013719.1 XM_017013720.1

SNP Detail

SNP ID Chromosome Location
rs1049336772 Chr.8: 38952407-38952407 on Build GRCh38
rs763632968 Chr.8: 38952303-38952303 on Build GRCh38
rs548951623 Chr.8: 38952337-38952337 on Build GRCh38
rs577287566 Chr.8: 38952467-38952467 on Build GRCh38
rs981624223 Chr.8: 38952439-38952439 on Build GRCh38
rs756934318 Chr.8: 38952313-38952313 on Build GRCh38
rs777817218 Chr.8: 38952490-38952490 on Build GRCh38
rs937551106 Chr.8: 38952482-38952482 on Build GRCh38
rs769012632 Chr.8: 38952361-38952361 on Build GRCh38
rs375718252 Chr.8: 38952344-38952344 on Build GRCh38
rs752432402 Chr.8: 38952283-38952283 on Build GRCh38
rs1040106372 Chr.8: 38952372-38952372 on Build GRCh38
rs756018373 Chr.8: 38952385-38952385 on Build GRCh38
rs914357410 Chr.8: 38952492-38952492 on Build GRCh38
rs1012816150 Chr.8: 38952427-38952427 on Build GRCh38
rs897693023 Chr.8: 38952375-38952376 on Build GRCh38
rs555852608 Chr.8: 38952415-38952415 on Build GRCh38
rs377061403 Chr.8: 38952381-38952381 on Build GRCh38
rs753483339 Chr.8: 38952306-38952306 on Build GRCh38
rs778329476 Chr.8: 38952331-38952331 on Build GRCh38
rs760562087 Chr.8: 38952294-38952294 on Build GRCh38
rs747312406 Chr.8: 38952348-38952348 on Build GRCh38
rs1055881168 Chr.8: 38952490-38952490 on Build GRCh38
rs111655245 Chr.8: 38952342-38952342 on Build GRCh38
rs147372019 Chr.8: 38952419-38952420 on Build GRCh38

View Full Product Details