Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 27866967-27867214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H1B
Entrez Gene ID
3009
Gene Name
histone cluster 1, H1b
Gene Aliases
H1, H1.5, H1B, H1F5, H1s-3
UniGene
Hs.131956
Gene Chromosome Location
Chr.6: 27866792-27867581 on Build GRCh38
Transcripts
NM_005322.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7142123 c.548C>A p.P183Q Chr.6: 27866982-27866982 on Build GRCh38
4690395 c.480G>A p.K160K Chr.6: 27867050-27867050 on Build GRCh38
6190638 c.538G>A p.A180T Chr.6: 27866992-27866992 on Build GRCh38
1442768 c.352_353insG p.E118fs*13 Chr.6: 27867177-27867178 on Build GRCh38
3745104 c.434C>T p.A145V Chr.6: 27867096-27867096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747685654 Chr.6: 27867059-27867059 on Build GRCh38
rs960161336 Chr.6: 27867133-27867133 on Build GRCh38
rs772021812 Chr.6: 27867155-27867155 on Build GRCh38
rs750807127 Chr.6: 27867090-27867090 on Build GRCh38
rs201327319 Chr.6: 27867170-27867170 on Build GRCh38
rs745567232 Chr.6: 27867067-27867069 on Build GRCh38
rs143393068 Chr.6: 27867056-27867056 on Build GRCh38
rs776470457 Chr.6: 27867071-27867071 on Build GRCh38
rs768117877 Chr.6: 27867021-27867021 on Build GRCh38
rs764722398 Chr.6: 27867050-27867050 on Build GRCh38
rs764242128 Chr.6: 27867080-27867080 on Build GRCh38
rs796439621 Chr.6: 27866989-27866989 on Build GRCh38
rs775898359 Chr.6: 27867108-27867108 on Build GRCh38
rs764891265 Chr.6: 27867093-27867093 on Build GRCh38
rs373151254 Chr.6: 27867022-27867022 on Build GRCh38
rs199758872 Chr.6: 27867138-27867138 on Build GRCh38
rs533040512 Chr.6: 27867107-27867109 on Build GRCh38
rs114406891 Chr.6: 27867053-27867053 on Build GRCh38
rs765210491 Chr.6: 27866992-27866992 on Build GRCh38
rs890228059 Chr.6: 27867013-27867013 on Build GRCh38
rs376086559 Chr.6: 27866997-27866997 on Build GRCh38
rs749095413 Chr.6: 27867107-27867130 on Build GRCh38
rs773541479 Chr.6: 27867121-27867121 on Build GRCh38
rs770056338 Chr.6: 27867074-27867074 on Build GRCh38
rs749797142 Chr.6: 27867173-27867173 on Build GRCh38
rs778964097 Chr.6: 27867146-27867146 on Build GRCh38
rs767856102 Chr.6: 27867087-27867087 on Build GRCh38
rs760484298 Chr.6: 27867036-27867036 on Build GRCh38
rs563961120 Chr.6: 27867015-27867015 on Build GRCh38
rs769005534 Chr.6: 27867145-27867145 on Build GRCh38
rs542637132 Chr.6: 27867028-27867028 on Build GRCh38
rs775808412 Chr.6: 27867078-27867078 on Build GRCh38
rs557604077 Chr.6: 27867135-27867135 on Build GRCh38
rs545007006 Chr.6: 27867056-27867067 on Build GRCh38
rs141210758 Chr.6: 27867044-27867044 on Build GRCh38
rs947308495 Chr.6: 27867160-27867160 on Build GRCh38
rs145569332 Chr.6: 27867182-27867182 on Build GRCh38
rs773808007 Chr.6: 27867161-27867161 on Build GRCh38
rs1036344606 Chr.6: 27867047-27867055 on Build GRCh38
rs928346209 Chr.6: 27867112-27867112 on Build GRCh38
rs759795708 Chr.6: 27867045-27867045 on Build GRCh38
rs768915463 Chr.6: 27867115-27867115 on Build GRCh38
rs775065810 Chr.6: 27867083-27867083 on Build GRCh38
rs778363954 Chr.6: 27867009-27867009 on Build GRCh38
rs763764720 Chr.6: 27867175-27867175 on Build GRCh38
rs770736739 Chr.6: 27867039-27867039 on Build GRCh38
rs372646605 Chr.6: 27867116-27867116 on Build GRCh38
rs926661430 Chr.6: 27867076-27867076 on Build GRCh38
rs148649312 Chr.6: 27867027-27867027 on Build GRCh38
rs766329707 Chr.6: 27867031-27867031 on Build GRCh38
rs911218988 Chr.6: 27867171-27867171 on Build GRCh38
rs969813689 Chr.6: 27867105-27867105 on Build GRCh38
rs780923320 Chr.6: 27867101-27867101 on Build GRCh38
rs745310887 Chr.6: 27867096-27867104 on Build GRCh38
rs751151753 Chr.6: 27867178-27867178 on Build GRCh38
rs749521475 Chr.6: 27867114-27867114 on Build GRCh38
rs370099818 Chr.6: 27866998-27866998 on Build GRCh38
rs1038798603 Chr.6: 27867129-27867129 on Build GRCh38
rs768398619 Chr.6: 27867153-27867188 on Build GRCh38
rs747600314 Chr.6: 27867023-27867023 on Build GRCh38
rs745700733 Chr.6: 27867102-27867102 on Build GRCh38
rs368971921 Chr.6: 27867086-27867086 on Build GRCh38
rs149411113 Chr.6: 27867012-27867012 on Build GRCh38
rs145278803 Chr.6: 27867079-27867079 on Build GRCh38
rs749034153 Chr.6: 27867140-27867140 on Build GRCh38
rs35906036 Chr.6: 27867017-27867017 on Build GRCh38
rs753493023 Chr.6: 27867054-27867054 on Build GRCh38
rs769692895 Chr.6: 27867106-27867106 on Build GRCh38
rs780734078 Chr.6: 27867055-27867056 on Build GRCh38
rs1025862705 Chr.6: 27866991-27866991 on Build GRCh38
rs541014157 Chr.6: 27867186-27867186 on Build GRCh38
rs745588089 Chr.6: 27867072-27867072 on Build GRCh38
rs367931912 Chr.6: 27867046-27867046 on Build GRCh38
rs748280261 Chr.6: 27867147-27867147 on Build GRCh38
rs138950128 Chr.6: 27867001-27867001 on Build GRCh38
rs140485202 Chr.6: 27867126-27867126 on Build GRCh38
rs200524277 Chr.6: 27867127-27867127 on Build GRCh38
rs753318525 Chr.6: 27867003-27867003 on Build GRCh38
rs149729430 Chr.6: 27867168-27867168 on Build GRCh38
rs757943776 Chr.6: 27867096-27867096 on Build GRCh38
rs763089363 Chr.6: 27867047-27867047 on Build GRCh38
rs756204649 Chr.6: 27867164-27867164 on Build GRCh38
rs200387249 Chr.6: 27867042-27867042 on Build GRCh38
rs11970638 Chr.6: 27867099-27867099 on Build GRCh38
rs748940475 Chr.6: 27867018-27867018 on Build GRCh38
rs768102848 Chr.6: 27867120-27867120 on Build GRCh38
rs971253532 Chr.6: 27867066-27867066 on Build GRCh38
rs758958536 Chr.6: 27867006-27867006 on Build GRCh38
rs781241531 Chr.6: 27867128-27867128 on Build GRCh38
rs146157091 Chr.6: 27866993-27866993 on Build GRCh38
rs746973357 Chr.6: 27867098-27867098 on Build GRCh38
rs1041269822 Chr.6: 27867067-27867067 on Build GRCh38
rs750300228 Chr.6: 27867131-27867131 on Build GRCh38
rs758029486 Chr.6: 27867124-27867124 on Build GRCh38
rs777735956 Chr.6: 27867060-27867060 on Build GRCh38
rs750436881 Chr.6: 27867187-27867187 on Build GRCh38
rs375871661 Chr.6: 27867123-27867123 on Build GRCh38
rs779982283 Chr.6: 27867139-27867139 on Build GRCh38
rs1047535184 Chr.6: 27867163-27867163 on Build GRCh38
rs774195585 Chr.6: 27867189-27867190 on Build GRCh38
rs774587530 Chr.6: 27867117-27867117 on Build GRCh38
rs139479440 Chr.6: 27867062-27867062 on Build GRCh38

View Full Product Details