Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 88046596-88046855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP16
Entrez Gene ID
4325
Gene Name
matrix metallopeptidase 16
Gene Aliases
C8orf57, MMP-X2, MT-MMP2, MT-MMP3, MT3-MMP
UniGene
Hs.492187
Gene Chromosome Location
Chr.8: 88037232-88327488 on Build GRCh38
Transcripts
NM_005941.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1700325 c.1408G>A p.D470N Chr.8: 88046750-88046750 on Build GRCh38
4466754 c.1450C>T p.P484S Chr.8: 88046708-88046708 on Build GRCh38
486736 c.1382G>A p.R461K Chr.8: 88046776-88046776 on Build GRCh38
3901928 c.1478A>C p.H493P Chr.8: 88046680-88046680 on Build GRCh38
7328457 c.1386T>C p.Y462Y Chr.8: 88046772-88046772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748048886 Chr.8: 88046630-88046630 on Build GRCh38
rs375297114 Chr.8: 88046646-88046646 on Build GRCh38
rs766663475 Chr.8: 88046736-88046736 on Build GRCh38
rs759446380 Chr.8: 88046665-88046665 on Build GRCh38
rs748881258 Chr.8: 88046816-88046816 on Build GRCh38
rs148465952 Chr.8: 88046699-88046699 on Build GRCh38
rs765305474 Chr.8: 88046682-88046682 on Build GRCh38
rs767362210 Chr.8: 88046789-88046789 on Build GRCh38
rs761144612 Chr.8: 88046648-88046648 on Build GRCh38
rs773350264 Chr.8: 88046709-88046709 on Build GRCh38
rs773976049 Chr.8: 88046794-88046795 on Build GRCh38
rs762250691 Chr.8: 88046671-88046671 on Build GRCh38
rs752681249 Chr.8: 88046795-88046795 on Build GRCh38
rs777837424 Chr.8: 88046811-88046811 on Build GRCh38
rs373547136 Chr.8: 88046635-88046635 on Build GRCh38
rs761516388 Chr.8: 88046799-88046801 on Build GRCh38
rs754823857 Chr.8: 88046786-88046786 on Build GRCh38
rs751689921 Chr.8: 88046772-88046772 on Build GRCh38
rs756692651 Chr.8: 88046819-88046819 on Build GRCh38
rs769438790 Chr.8: 88046631-88046631 on Build GRCh38
rs770671507 Chr.8: 88046642-88046642 on Build GRCh38
rs1042079518 Chr.8: 88046641-88046641 on Build GRCh38
rs1050892041 Chr.8: 88046798-88046798 on Build GRCh38
rs532743913 Chr.8: 88046822-88046822 on Build GRCh38
rs997950104 Chr.8: 88046753-88046753 on Build GRCh38
rs756184927 Chr.8: 88046810-88046810 on Build GRCh38
rs762929864 Chr.8: 88046727-88046727 on Build GRCh38
rs202016106 Chr.8: 88046657-88046657 on Build GRCh38
rs748800899 Chr.8: 88046637-88046637 on Build GRCh38

View Full Product Details