Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 155084970-155085219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MME
Entrez Gene ID
4311
Gene Name
membrane metallo-endopeptidase
Gene Aliases
CALLA, CD10, CMT2T, NEP, SFE
UniGene
Hs.307734
Gene Chromosome Location
Chr.3: 155024124-155183729 on Build GRCh38
Transcripts
NM_000902.3 NM_007287.2 NM_007288.2 NM_007289.2 XM_006713646.3 XM_006713647.3 XM_011512855.2 XM_011512856.2 XM_011512857.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3702349 c.182A>T p.D61V Chr.3: 155085080-155085080 on Build GRCh38
6544964 c.185G>T p.C62F Chr.3: 155085083-155085083 on Build GRCh38
4137007 c.196+2T>A p.? Chr.3: 155085096-155085096 on Build GRCh38
1257821 c.167T>A p.I56N Chr.3: 155085065-155085065 on Build GRCh38
6493850 c.196delG p.A66fs*4 Chr.3: 155085094-155085094 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749573166 Chr.3: 155085063-155085063 on Build GRCh38
rs573470222 Chr.3: 155085125-155085125 on Build GRCh38
rs201076610 Chr.3: 155085059-155085059 on Build GRCh38
rs753448967 Chr.3: 155085020-155085020 on Build GRCh38
rs143147554 Chr.3: 155085112-155085112 on Build GRCh38
rs556193537 Chr.3: 155085151-155085151 on Build GRCh38
rs768014053 Chr.3: 155085146-155085146 on Build GRCh38
rs947966494 Chr.3: 155085182-155085182 on Build GRCh38
rs202220058 Chr.3: 155085143-155085143 on Build GRCh38
rs199801768 Chr.3: 155085042-155085042 on Build GRCh38
rs777987661 Chr.3: 155085056-155085056 on Build GRCh38
rs771899867 Chr.3: 155085104-155085104 on Build GRCh38
rs374391710 Chr.3: 155085175-155085175 on Build GRCh38
rs775192412 Chr.3: 155085105-155085105 on Build GRCh38
rs199926724 Chr.3: 155085050-155085050 on Build GRCh38
rs769934048 Chr.3: 155085054-155085054 on Build GRCh38
rs761509638 Chr.3: 155085021-155085021 on Build GRCh38
rs751888468 Chr.3: 155085133-155085133 on Build GRCh38
rs763866586 Chr.3: 155085095-155085097 on Build GRCh38
rs201622614 Chr.3: 155085048-155085048 on Build GRCh38
rs764896118 Chr.3: 155085124-155085124 on Build GRCh38
rs149028419 Chr.3: 155085029-155085029 on Build GRCh38
rs201910473 Chr.3: 155085091-155085091 on Build GRCh38
rs763629171 Chr.3: 155085010-155085010 on Build GRCh38
rs188259964 Chr.3: 155085044-155085044 on Build GRCh38
rs755468821 Chr.3: 155085134-155085134 on Build GRCh38
rs780349049 Chr.3: 155085039-155085039 on Build GRCh38
rs758774449 Chr.3: 155085038-155085038 on Build GRCh38
rs995785969 Chr.3: 155085022-155085022 on Build GRCh38
rs35926730 Chr.3: 155085060-155085060 on Build GRCh38
rs4607069 Chr.3: 155085131-155085131 on Build GRCh38
rs73875803 Chr.3: 155085113-155085113 on Build GRCh38
rs202035845 Chr.3: 155085026-155085026 on Build GRCh38
rs750757127 Chr.3: 155085035-155085035 on Build GRCh38
rs762553367 Chr.3: 155085127-155085127 on Build GRCh38
rs572236471 Chr.3: 155085003-155085003 on Build GRCh38
rs577995933 Chr.3: 155085157-155085157 on Build GRCh38

View Full Product Details