Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 123187010-123187283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDM6
Entrez Gene ID
93166
Gene Name
PR domain 6
Gene Aliases
PRISM
UniGene
Hs.135118
Gene Chromosome Location
Chr.5: 123089146-123194266 on Build GRCh38
Transcripts
NM_001136239.1 XM_011543726.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3135189 c.1711C>A p.Q571K Chr.5: 123187124-123187124 on Build GRCh38
7325634 c.1759_1760delGA p.S588fs*>8 Chr.5: 123187172-123187173 on Build GRCh38
3135190 c.1713G>T p.Q571H Chr.5: 123187126-123187126 on Build GRCh38
5402037 c.1777G>A p.E593K Chr.5: 123187190-123187190 on Build GRCh38
7323005 c.1676G>A p.C559Y Chr.5: 123187089-123187089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369490038 Chr.5: 123187090-123187090 on Build GRCh38
rs746204208 Chr.5: 123187073-123187073 on Build GRCh38
rs1055598167 Chr.5: 123187056-123187056 on Build GRCh38
rs758010129 Chr.5: 123187064-123187064 on Build GRCh38
rs774379926 Chr.5: 123187041-123187041 on Build GRCh38
rs335198 Chr.5: 123187050-123187050 on Build GRCh38
rs941901656 Chr.5: 123187230-123187230 on Build GRCh38
rs749812428 Chr.5: 123187104-123187104 on Build GRCh38
rs200894287 Chr.5: 123187037-123187037 on Build GRCh38
rs538574724 Chr.5: 123187094-123187094 on Build GRCh38
rs778888246 Chr.5: 123187119-123187119 on Build GRCh38
rs887665609 Chr.5: 123187227-123187227 on Build GRCh38
rs963337273 Chr.5: 123187189-123187189 on Build GRCh38
rs912045236 Chr.5: 123187253-123187253 on Build GRCh38
rs1041508063 Chr.5: 123187036-123187036 on Build GRCh38
rs764659923 Chr.5: 123187047-123187047 on Build GRCh38
rs986807050 Chr.5: 123187049-123187049 on Build GRCh38
rs373102899 Chr.5: 123187203-123187203 on Build GRCh38
rs756542248 Chr.5: 123187091-123187091 on Build GRCh38
rs1045388452 Chr.5: 123187209-123187209 on Build GRCh38
rs369418081 Chr.5: 123187210-123187210 on Build GRCh38
rs373228627 Chr.5: 123187107-123187107 on Build GRCh38
rs992860171 Chr.5: 123187163-123187163 on Build GRCh38
rs894415131 Chr.5: 123187077-123187077 on Build GRCh38
rs466025 Chr.5: 123187055-123187055 on Build GRCh38
rs74354350 Chr.5: 123187176-123187176 on Build GRCh38
rs556457540 Chr.5: 123187133-123187133 on Build GRCh38
rs948724070 Chr.5: 123187187-123187187 on Build GRCh38
rs760542283 Chr.5: 123187202-123187202 on Build GRCh38
rs1036150773 Chr.5: 123187247-123187247 on Build GRCh38
rs773608431 Chr.5: 123187190-123187190 on Build GRCh38
rs777532348 Chr.5: 123187071-123187071 on Build GRCh38
rs148505766 Chr.5: 123187030-123187030 on Build GRCh38
rs375354972 Chr.5: 123187150-123187150 on Build GRCh38
rs938733052 Chr.5: 123187152-123187152 on Build GRCh38

View Full Product Details