Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21550947-21551208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf51
Entrez Gene ID
339263
Gene Name
chromosome 17 open reading frame 51
Gene Aliases
-
UniGene
Hs.514016
Gene Chromosome Location
Chr.17: 21524606-21551678 on Build GRCh38
Transcripts
NM_001113434.4 XM_005256621.3 XM_006721514.2 XM_011523822.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3766047 c.355T>C p.S119P Chr.17: 21551068-21551068 on Build GRCh38
6403478 c.348C>A p.S116R Chr.17: 21551075-21551075 on Build GRCh38
5346737 c.343A>T p.S115C Chr.17: 21551080-21551080 on Build GRCh38
6373721 c.347G>T p.S116I Chr.17: 21551076-21551076 on Build GRCh38
5972089 c.306G>C p.Q102H Chr.17: 21551117-21551117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539038513 Chr.17: 21551076-21551076 on Build GRCh38
rs571511305 Chr.17: 21551045-21551045 on Build GRCh38
rs781976335 Chr.17: 21551156-21551156 on Build GRCh38
rs782357273 Chr.17: 21551154-21551154 on Build GRCh38
rs782813680 Chr.17: 21551145-21551145 on Build GRCh38
rs372929778 Chr.17: 21551005-21551005 on Build GRCh38
rs369645906 Chr.17: 21551051-21551051 on Build GRCh38
rs781979288 Chr.17: 21551174-21551174 on Build GRCh38
rs574137272 Chr.17: 21551097-21551097 on Build GRCh38
rs557343982 Chr.17: 21551098-21551098 on Build GRCh38
rs546690821 Chr.17: 21550991-21550991 on Build GRCh38
rs201484956 Chr.17: 21551105-21551105 on Build GRCh38
rs567410766 Chr.17: 21550975-21550975 on Build GRCh38
rs200894326 Chr.17: 21551043-21551043 on Build GRCh38
rs781933490 Chr.17: 21551103-21551103 on Build GRCh38
rs534836985 Chr.17: 21550990-21550990 on Build GRCh38

View Full Product Details