Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75107438-75107697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD31
Entrez Gene ID
256006
Gene Name
ankyrin repeat domain 31
Gene Aliases
-
UniGene
Hs.482533
Gene Chromosome Location
Chr.5: 75068297-75236897 on Build GRCh38
Transcripts
NM_001164443.1 XM_011543298.2 XM_011543299.1 XM_011543300.2 XM_011543301.2 XM_011543302.2 XM_017009317.1 XM_017009318.1 XM_017009319.1 XM_017009320.1 XM_017009321.1 XM_017009322.1

SNP Detail

SNP ID Chromosome Location
rs760356990 Chr.5: 75107639-75107639 on Build GRCh38
rs759509117 Chr.5: 75107539-75107539 on Build GRCh38
rs900599342 Chr.5: 75107531-75107531 on Build GRCh38
rs1018097687 Chr.5: 75107621-75107621 on Build GRCh38
rs765154054 Chr.5: 75107558-75107558 on Build GRCh38
rs187640209 Chr.5: 75107613-75107613 on Build GRCh38
rs753762043 Chr.5: 75107653-75107653 on Build GRCh38
rs34233525 Chr.5: 75107477-75107478 on Build GRCh38
rs766389829 Chr.5: 75107647-75107647 on Build GRCh38
rs750371805 Chr.5: 75107584-75107584 on Build GRCh38
rs777252991 Chr.5: 75107494-75107494 on Build GRCh38
rs763282070 Chr.5: 75107585-75107585 on Build GRCh38
rs903683279 Chr.5: 75107637-75107637 on Build GRCh38
rs929966480 Chr.5: 75107507-75107507 on Build GRCh38
rs553352858 Chr.5: 75107617-75107617 on Build GRCh38
rs755248977 Chr.5: 75107553-75107553 on Build GRCh38
rs1037816095 Chr.5: 75107497-75107497 on Build GRCh38
rs764171416 Chr.5: 75107586-75107586 on Build GRCh38
rs750261213 Chr.5: 75107592-75107592 on Build GRCh38
rs775643093 Chr.5: 75107579-75107579 on Build GRCh38
rs750155018 Chr.5: 75107497-75107498 on Build GRCh38
rs112521958 Chr.5: 75107658-75107658 on Build GRCh38
rs759743544 Chr.5: 75107535-75107535 on Build GRCh38
rs769986159 Chr.5: 75107578-75107578 on Build GRCh38

View Full Product Details