Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 45442871-45443132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX5
Entrez Gene ID
240
Gene Name
arachidonate 5-lipoxygenase
Gene Aliases
5-LO, 5-LOX, 5LPG, LOG5
UniGene
Hs.89499
Gene Chromosome Location
Chr.10: 45374166-45446121 on Build GRCh38
Transcripts
NM_000698.4 NM_001256153.2 NM_001256154.2 NM_001320861.1 NM_001320862.1 XM_017016012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7243014 c.1340C>A p.A447D Chr.10: 45443105-45443105 on Build GRCh38
4641358 c.1316G>A p.R439K Chr.10: 45443081-45443081 on Build GRCh38
7296296 c.1299C>T p.H433H Chr.10: 45443064-45443064 on Build GRCh38
1244862 c.1300G>A p.V434M Chr.10: 45443065-45443065 on Build GRCh38
1347872 c.1334C>T p.T445I Chr.10: 45443099-45443099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756199477 Chr.10: 45443010-45443010 on Build GRCh38
rs903655140 Chr.10: 45443020-45443020 on Build GRCh38
rs779380506 Chr.10: 45443033-45443033 on Build GRCh38
rs554154845 Chr.10: 45443065-45443065 on Build GRCh38
rs537072496 Chr.10: 45443043-45443043 on Build GRCh38
rs9282591 Chr.10: 45442907-45442907 on Build GRCh38
rs761325956 Chr.10: 45443052-45443052 on Build GRCh38
rs890918317 Chr.10: 45442951-45442951 on Build GRCh38
rs147327397 Chr.10: 45443061-45443061 on Build GRCh38
rs1041030806 Chr.10: 45442924-45442924 on Build GRCh38
rs1003841751 Chr.10: 45442952-45442952 on Build GRCh38
rs1013946436 Chr.10: 45443078-45443078 on Build GRCh38
rs769243207 Chr.10: 45443058-45443058 on Build GRCh38
rs749766003 Chr.10: 45443016-45443016 on Build GRCh38
rs202085482 Chr.10: 45443106-45443106 on Build GRCh38
rs1010268448 Chr.10: 45442957-45442957 on Build GRCh38
rs759246654 Chr.10: 45443062-45443062 on Build GRCh38
rs996710436 Chr.10: 45442904-45442904 on Build GRCh38
rs375848132 Chr.10: 45443107-45443107 on Build GRCh38
rs780844592 Chr.10: 45443091-45443091 on Build GRCh38
rs28395869 Chr.10: 45443104-45443104 on Build GRCh38
rs773162136 Chr.10: 45443060-45443060 on Build GRCh38
rs746202423 Chr.10: 45443041-45443041 on Build GRCh38
rs778441503 Chr.10: 45443013-45443013 on Build GRCh38
rs757837007 Chr.10: 45443076-45443076 on Build GRCh38
rs179363899 Chr.10: 45443004-45443004 on Build GRCh38
rs962290583 Chr.10: 45442987-45442987 on Build GRCh38
rs182044631 Chr.10: 45443009-45443009 on Build GRCh38
rs778036906 Chr.10: 45443068-45443068 on Build GRCh38
rs537506989 Chr.10: 45442939-45442939 on Build GRCh38
rs755278305 Chr.10: 45443007-45443007 on Build GRCh38
rs139206290 Chr.10: 45443112-45443112 on Build GRCh38
rs746248416 Chr.10: 45443084-45443084 on Build GRCh38
rs777548521 Chr.10: 45443050-45443052 on Build GRCh38
rs995514178 Chr.10: 45443001-45443001 on Build GRCh38
rs200646971 Chr.10: 45442956-45442956 on Build GRCh38
rs572629091 Chr.10: 45443087-45443087 on Build GRCh38
rs781262591 Chr.10: 45443055-45443055 on Build GRCh38
rs911497736 Chr.10: 45442901-45442901 on Build GRCh38
rs779279218 Chr.10: 45443083-45443083 on Build GRCh38
rs753942816 Chr.10: 45443069-45443069 on Build GRCh38
rs944316485 Chr.10: 45442903-45442903 on Build GRCh38
rs1027892958 Chr.10: 45443003-45443003 on Build GRCh38
rs772760111 Chr.10: 45443110-45443110 on Build GRCh38
rs776198562 Chr.10: 45443045-45443045 on Build GRCh38
rs767787897 Chr.10: 45443064-45443064 on Build GRCh38
rs376983759 Chr.10: 45443027-45443027 on Build GRCh38
rs1015680802 Chr.10: 45442973-45442973 on Build GRCh38
rs756287174 Chr.10: 45443067-45443067 on Build GRCh38

View Full Product Details