Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2132816-2133055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2
Entrez Gene ID
3481
Gene Name
insulin like growth factor 2
Gene Aliases
C11orf43, GRDF, IGF-II, PP9974
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2149603 on Build GRCh38
Transcripts
NM_000612.5 NM_001007139.5 NM_001127598.2 NM_001291861.2 NM_001291862.2 NR_003512.3

Gene Symbol
INS-IGF2
Entrez Gene ID
723961
Gene Name
INS-IGF2 readthrough
Gene Aliases
INSIGF
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2161209 on Build GRCh38
Transcripts
NM_000612.5 NM_001007139.5 NM_001127598.2 NM_001291861.2 NM_001291862.2 NR_003512.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5505310 c.657C>T p.T219T Chr.11: 2133041-2133041 on Build GRCh38
5596860 c.683C>T p.P228L Chr.11: 2133015-2133015 on Build GRCh38
4852080 c.498C>A p.P166P Chr.11: 2133032-2133032 on Build GRCh38
4766692 c.536G>A p.R179Q Chr.11: 2132994-2132994 on Build GRCh38
6936716 c.676G>A p.G226S Chr.11: 2133022-2133022 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756776702 Chr.11: 2132981-2132981 on Build GRCh38
rs953987752 Chr.11: 2132842-2132842 on Build GRCh38
rs771725807 Chr.11: 2132858-2132859 on Build GRCh38
rs377316111 Chr.11: 2133031-2133031 on Build GRCh38
rs1052860378 Chr.11: 2132883-2132883 on Build GRCh38
rs760902159 Chr.11: 2132946-2132946 on Build GRCh38
rs554804044 Chr.11: 2133025-2133025 on Build GRCh38
rs200036157 Chr.11: 2133026-2133026 on Build GRCh38
rs763805360 Chr.11: 2132974-2132974 on Build GRCh38
rs1050844499 Chr.11: 2132924-2132924 on Build GRCh38
rs747026832 Chr.11: 2133021-2133021 on Build GRCh38
rs948917125 Chr.11: 2132854-2132854 on Build GRCh38
rs972572597 Chr.11: 2132892-2132892 on Build GRCh38
rs912453779 Chr.11: 2132852-2132852 on Build GRCh38
rs11545014 Chr.11: 2133013-2133013 on Build GRCh38
rs9282726 Chr.11: 2132973-2132973 on Build GRCh38
rs1014211286 Chr.11: 2132995-2132995 on Build GRCh38
rs902646764 Chr.11: 2133014-2133014 on Build GRCh38
rs759186937 Chr.11: 2132960-2132960 on Build GRCh38
rs751058590 Chr.11: 2132976-2132976 on Build GRCh38
rs773392012 Chr.11: 2132936-2132936 on Build GRCh38
rs767815580 Chr.11: 2133032-2133032 on Build GRCh38
rs935689640 Chr.11: 2132869-2132869 on Build GRCh38
rs1006752092 Chr.11: 2132928-2132928 on Build GRCh38
rs200131466 Chr.11: 2132957-2132957 on Build GRCh38
rs34337549 Chr.11: 2132877-2132877 on Build GRCh38
rs763681027 Chr.11: 2133035-2133035 on Build GRCh38
rs2230949 Chr.11: 2132958-2132958 on Build GRCh38
rs773624887 Chr.11: 2133018-2133018 on Build GRCh38
rs925639358 Chr.11: 2132864-2132866 on Build GRCh38
rs3213234 Chr.11: 2132927-2132927 on Build GRCh38
rs889623176 Chr.11: 2132925-2132925 on Build GRCh38
rs992974582 Chr.11: 2132837-2132837 on Build GRCh38
rs538267300 Chr.11: 2132902-2132902 on Build GRCh38
rs376978352 Chr.11: 2132878-2132878 on Build GRCh38
rs12993 Chr.11: 2132990-2132990 on Build GRCh38
rs891509994 Chr.11: 2132887-2132887 on Build GRCh38
rs1001308758 Chr.11: 2132836-2132836 on Build GRCh38
rs755455183 Chr.11: 2133011-2133012 on Build GRCh38
rs549471313 Chr.11: 2132851-2132851 on Build GRCh38
rs3741214 Chr.11: 2132961-2132961 on Build GRCh38
rs771138084 Chr.11: 2133022-2133022 on Build GRCh38
rs749836523 Chr.11: 2133008-2133008 on Build GRCh38
rs1048095045 Chr.11: 2132926-2132926 on Build GRCh38
rs570973772 Chr.11: 2132884-2132884 on Build GRCh38
rs565575035 Chr.11: 2132994-2132994 on Build GRCh38
rs138399775 Chr.11: 2133020-2133020 on Build GRCh38
rs1050342 Chr.11: 2133012-2133012 on Build GRCh38
rs931067409 Chr.11: 2132923-2132923 on Build GRCh38
rs763531701 Chr.11: 2132891-2132891 on Build GRCh38
rs1032351201 Chr.11: 2132838-2132838 on Build GRCh38
rs1044703347 Chr.11: 2132863-2132863 on Build GRCh38

View Full Product Details