Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63800407-63800672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX42
Entrez Gene ID
11325
Gene Name
DEAD-box helicase 42
Gene Aliases
DDX42P, RHELP, RNAHP, SF3B8, SF3b125
UniGene
Hs.702010
Gene Chromosome Location
Chr.17: 63773856-63819317 on Build GRCh38
Transcripts
NM_007372.3 NM_203499.2 XM_006721657.1 XM_011524259.2 XM_017024111.1 XM_017024112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2795334 c.580delA p.I196fs*29 Chr.17: 63800576-63800576 on Build GRCh38
212765 c.559G>A p.G187R Chr.17: 63800555-63800555 on Build GRCh38
5773817 c.619G>T p.E207* Chr.17: 63800615-63800615 on Build GRCh38
3520790 c.518T>G p.V173G Chr.17: 63800514-63800514 on Build GRCh38
213150 c.511G>C p.G171R Chr.17: 63800507-63800507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929082052 Chr.17: 63800646-63800646 on Build GRCh38
rs753255162 Chr.17: 63800564-63800564 on Build GRCh38
rs754868344 Chr.17: 63800632-63800632 on Build GRCh38
rs138225391 Chr.17: 63800437-63800437 on Build GRCh38
rs751432016 Chr.17: 63800603-63800603 on Build GRCh38
rs149577156 Chr.17: 63800554-63800554 on Build GRCh38
rs774716914 Chr.17: 63800512-63800512 on Build GRCh38
rs373347096 Chr.17: 63800464-63800464 on Build GRCh38
rs1009392960 Chr.17: 63800436-63800436 on Build GRCh38
rs992259661 Chr.17: 63800582-63800582 on Build GRCh38
rs769703657 Chr.17: 63800637-63800637 on Build GRCh38
rs1020887698 Chr.17: 63800566-63800566 on Build GRCh38
rs748075616 Chr.17: 63800442-63800442 on Build GRCh38
rs757435317 Chr.17: 63800596-63800596 on Build GRCh38
rs779228678 Chr.17: 63800600-63800600 on Build GRCh38
rs753890978 Chr.17: 63800589-63800589 on Build GRCh38
rs781119487 Chr.17: 63800633-63800633 on Build GRCh38
rs967454746 Chr.17: 63800577-63800577 on Build GRCh38
rs772772214 Chr.17: 63800454-63800454 on Build GRCh38
rs776670518 Chr.17: 63800440-63800440 on Build GRCh38
rs147272438 Chr.17: 63800628-63800628 on Build GRCh38
rs143708043 Chr.17: 63800586-63800586 on Build GRCh38
rs765996092 Chr.17: 63800510-63800510 on Build GRCh38
rs979351083 Chr.17: 63800487-63800487 on Build GRCh38
rs764149045 Chr.17: 63800588-63800588 on Build GRCh38
rs1050666219 Chr.17: 63800505-63800505 on Build GRCh38
rs751624402 Chr.17: 63800646-63800647 on Build GRCh38
rs369374729 Chr.17: 63800448-63800448 on Build GRCh38
rs201646387 Chr.17: 63800536-63800536 on Build GRCh38

View Full Product Details