Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20655393-20655642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDOST
Entrez Gene ID
1650
Gene Name
dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit
Gene Aliases
AGER1, CDG1R, OKSWcl45, OST, OST48, WBP1
UniGene
Hs.523145
Gene Chromosome Location
Chr.1: 20651767-20661544 on Build GRCh38
Transcripts
NM_005216.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1601604 c.547G>T p.A183S Chr.1: 20655495-20655495 on Build GRCh38
1929149 c.513G>A p.T171T Chr.1: 20655529-20655529 on Build GRCh38
1929150 c.508-1G>T p.? Chr.1: 20655535-20655535 on Build GRCh38
118659 c.582C>T p.P194P Chr.1: 20655460-20655460 on Build GRCh38
3482885 c.549C>T p.A183A Chr.1: 20655493-20655493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376070047 Chr.1: 20655523-20655523 on Build GRCh38
rs373001976 Chr.1: 20655548-20655548 on Build GRCh38
rs748859395 Chr.1: 20655545-20655545 on Build GRCh38
rs747589765 Chr.1: 20655450-20655450 on Build GRCh38
rs372948054 Chr.1: 20655489-20655489 on Build GRCh38
rs914065741 Chr.1: 20655599-20655599 on Build GRCh38
rs745612594 Chr.1: 20655570-20655570 on Build GRCh38
rs774007750 Chr.1: 20655480-20655480 on Build GRCh38
rs924948918 Chr.1: 20655533-20655533 on Build GRCh38
rs369049692 Chr.1: 20655544-20655544 on Build GRCh38
rs770937124 Chr.1: 20655470-20655470 on Build GRCh38
rs904279388 Chr.1: 20655576-20655577 on Build GRCh38
rs769322830 Chr.1: 20655457-20655457 on Build GRCh38
rs111334879 Chr.1: 20655483-20655483 on Build GRCh38
rs757889180 Chr.1: 20655418-20655418 on Build GRCh38
rs149487745 Chr.1: 20655484-20655484 on Build GRCh38
rs754011834 Chr.1: 20655486-20655486 on Build GRCh38
rs146767054 Chr.1: 20655482-20655482 on Build GRCh38
rs1001022882 Chr.1: 20655596-20655596 on Build GRCh38
rs745406431 Chr.1: 20655547-20655548 on Build GRCh38
rs750695197 Chr.1: 20655496-20655496 on Build GRCh38
rs886045871 Chr.1: 20655532-20655532 on Build GRCh38
rs755809942 Chr.1: 20655538-20655538 on Build GRCh38
rs777635772 Chr.1: 20655459-20655459 on Build GRCh38
rs765830229 Chr.1: 20655494-20655494 on Build GRCh38
rs778917497 Chr.1: 20655556-20655556 on Build GRCh38
rs933186948 Chr.1: 20655460-20655460 on Build GRCh38
rs771926276 Chr.1: 20655573-20655573 on Build GRCh38
rs936394586 Chr.1: 20655543-20655543 on Build GRCh38
rs779117025 Chr.1: 20655423-20655423 on Build GRCh38
rs369573026 Chr.1: 20655429-20655429 on Build GRCh38
rs771814545 Chr.1: 20655580-20655581 on Build GRCh38
rs749197629 Chr.1: 20655468-20655468 on Build GRCh38
rs975020316 Chr.1: 20655529-20655529 on Build GRCh38
rs759393357 Chr.1: 20655481-20655481 on Build GRCh38
rs189359933 Chr.1: 20655595-20655595 on Build GRCh38
rs1043222730 Chr.1: 20655566-20655566 on Build GRCh38
rs780909764 Chr.1: 20655449-20655449 on Build GRCh38
rs756270668 Chr.1: 20655413-20655413 on Build GRCh38
rs754420505 Chr.1: 20655416-20655416 on Build GRCh38
rs780666099 Chr.1: 20655526-20655526 on Build GRCh38
rs993998618 Chr.1: 20655454-20655454 on Build GRCh38
rs754573232 Chr.1: 20655439-20655439 on Build GRCh38
rs76266717 Chr.1: 20655415-20655415 on Build GRCh38
rs759658987 Chr.1: 20655549-20655549 on Build GRCh38
rs11547742 Chr.1: 20655530-20655530 on Build GRCh38

View Full Product Details