Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156841461-156841712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSRR
Entrez Gene ID
3645
Gene Name
insulin receptor related receptor
Gene Aliases
IRR
UniGene
Hs.248138
Gene Chromosome Location
Chr.1: 156840873-156858920 on Build GRCh38
Transcripts
NM_014215.2 NM_001007792.1

Gene Symbol
NTRK1
Entrez Gene ID
4914
Gene Name
neurotrophic receptor tyrosine kinase 1
Gene Aliases
MTC, TRK, TRK1, TRKA, Trk-A, p140-TrkA
UniGene
Hs.406293
Gene Chromosome Location
Chr.1: 156815750-156881850 on Build GRCh38
Transcripts
NM_014215.2 NM_001007792.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6980130 c.3481C>A p.P1161T Chr.1: 156841711-156841711 on Build GRCh38
5968880 c.3484G>A p.E1162K Chr.1: 156841708-156841708 on Build GRCh38
1688775 c.3488C>T p.S1163F Chr.1: 156841704-156841704 on Build GRCh38
3475405 c.3504C>T p.I1168I Chr.1: 156841688-156841688 on Build GRCh38
6216091 c.3527+8C>T p.? Chr.1: 156841657-156841657 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760117540 Chr.1: 156841541-156841541 on Build GRCh38
rs781207074 Chr.1: 156841501-156841501 on Build GRCh38
rs35335439 Chr.1: 156841662-156841663 on Build GRCh38
rs756041845 Chr.1: 156841480-156841480 on Build GRCh38
rs377055667 Chr.1: 156841566-156841566 on Build GRCh38
rs775010265 Chr.1: 156841658-156841658 on Build GRCh38
rs761696263 Chr.1: 156841673-156841673 on Build GRCh38
rs755143958 Chr.1: 156841575-156841575 on Build GRCh38
rs768850524 Chr.1: 156841686-156841686 on Build GRCh38
rs763035973 Chr.1: 156841633-156841633 on Build GRCh38
rs778144377 Chr.1: 156841518-156841518 on Build GRCh38
rs757050728 Chr.1: 156841487-156841487 on Build GRCh38
rs751827758 Chr.1: 156841492-156841492 on Build GRCh38
rs770274884 Chr.1: 156841507-156841507 on Build GRCh38
rs928421594 Chr.1: 156841623-156841623 on Build GRCh38
rs767771384 Chr.1: 156841678-156841678 on Build GRCh38
rs763962327 Chr.1: 156841485-156841485 on Build GRCh38
rs760229277 Chr.1: 156841661-156841661 on Build GRCh38
rs546133638 Chr.1: 156841677-156841677 on Build GRCh38
rs138965981 Chr.1: 156841682-156841682 on Build GRCh38
rs771886063 Chr.1: 156841533-156841533 on Build GRCh38
rs576165880 Chr.1: 156841665-156841665 on Build GRCh38
rs192415965 Chr.1: 156841627-156841627 on Build GRCh38
rs775019889 Chr.1: 156841534-156841534 on Build GRCh38
rs749862084 Chr.1: 156841647-156841647 on Build GRCh38
rs762117969 Chr.1: 156841656-156841656 on Build GRCh38
rs374122291 Chr.1: 156841652-156841652 on Build GRCh38
rs772767878 Chr.1: 156841576-156841577 on Build GRCh38
rs866669566 Chr.1: 156841688-156841688 on Build GRCh38
rs748197635 Chr.1: 156841502-156841502 on Build GRCh38
rs574446043 Chr.1: 156841526-156841526 on Build GRCh38
rs200304796 Chr.1: 156841674-156841674 on Build GRCh38
rs752751634 Chr.1: 156841620-156841620 on Build GRCh38
rs750249647 Chr.1: 156841567-156841567 on Build GRCh38
rs55694053 Chr.1: 156841679-156841679 on Build GRCh38
rs146967458 Chr.1: 156841519-156841519 on Build GRCh38
rs753787252 Chr.1: 156841561-156841561 on Build GRCh38
rs184789044 Chr.1: 156841651-156841651 on Build GRCh38
rs774129177 Chr.1: 156841520-156841520 on Build GRCh38
rs781281568 Chr.1: 156841578-156841578 on Build GRCh38
rs201428101 Chr.1: 156841657-156841657 on Build GRCh38
rs774816847 Chr.1: 156841605-156841605 on Build GRCh38
rs755019469 Chr.1: 156841496-156841496 on Build GRCh38
rs753613162 Chr.1: 156841486-156841486 on Build GRCh38
rs951148500 Chr.1: 156841595-156841595 on Build GRCh38
rs372532208 Chr.1: 156841553-156841553 on Build GRCh38
rs780251872 Chr.1: 156841488-156841488 on Build GRCh38
rs899260228 Chr.1: 156841515-156841515 on Build GRCh38
rs761392074 Chr.1: 156841518-156841519 on Build GRCh38
rs546911132 Chr.1: 156841566-156841567 on Build GRCh38
rs964356412 Chr.1: 156841650-156841650 on Build GRCh38
rs370465263 Chr.1: 156841622-156841622 on Build GRCh38
rs987375243 Chr.1: 156841606-156841606 on Build GRCh38

View Full Product Details