Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 61877170-61877443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT4
Entrez Gene ID
10575
Gene Name
chaperonin containing TCP1 subunit 4
Gene Aliases
CCT-DELTA, Cctd, SRB
UniGene
Hs.421509
Gene Chromosome Location
Chr.2: 61868127-61888671 on Build GRCh38
Transcripts
NM_001256721.1 NM_006430.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1752604 c.636G>A p.K212K Chr.2: 61877401-61877401 on Build GRCh38
5726915 c.613C>T p.L205F Chr.2: 61877424-61877424 on Build GRCh38
341322 c.610G>C p.D204H Chr.2: 61877427-61877427 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs184610212 Chr.2: 61877270-61877270 on Build GRCh38
rs1050474591 Chr.2: 61877332-61877332 on Build GRCh38
rs746302991 Chr.2: 61877404-61877404 on Build GRCh38
rs781596389 Chr.2: 61877395-61877395 on Build GRCh38
rs923143707 Chr.2: 61877260-61877260 on Build GRCh38
rs1002478326 Chr.2: 61877337-61877337 on Build GRCh38
rs535822789 Chr.2: 61877271-61877271 on Build GRCh38
rs961681605 Chr.2: 61877222-61877222 on Build GRCh38
rs1046401534 Chr.2: 61877324-61877324 on Build GRCh38
rs1033865246 Chr.2: 61877198-61877198 on Build GRCh38
rs756672679 Chr.2: 61877398-61877400 on Build GRCh38
rs546418200 Chr.2: 61877326-61877326 on Build GRCh38
rs542190105 Chr.2: 61877349-61877349 on Build GRCh38
rs577373424 Chr.2: 61877211-61877211 on Build GRCh38
rs1053861876 Chr.2: 61877303-61877303 on Build GRCh38
rs776930350 Chr.2: 61877394-61877394 on Build GRCh38
rs976595206 Chr.2: 61877315-61877315 on Build GRCh38
rs189880501 Chr.2: 61877418-61877418 on Build GRCh38
rs978422253 Chr.2: 61877283-61877283 on Build GRCh38
rs766324495 Chr.2: 61877402-61877402 on Build GRCh38
rs372411100 Chr.2: 61877382-61877382 on Build GRCh38
rs7571143 Chr.2: 61877208-61877208 on Build GRCh38
rs936515331 Chr.2: 61877272-61877272 on Build GRCh38
rs770338304 Chr.2: 61877414-61877414 on Build GRCh38
rs758005184 Chr.2: 61877375-61877375 on Build GRCh38
rs755953210 Chr.2: 61877343-61877343 on Build GRCh38
rs367831859 Chr.2: 61877258-61877258 on Build GRCh38
rs1038379603 Chr.2: 61877204-61877204 on Build GRCh38
rs892592990 Chr.2: 61877308-61877308 on Build GRCh38
rs147439665 Chr.2: 61877369-61877369 on Build GRCh38
rs754998244 Chr.2: 61877366-61877366 on Build GRCh38
rs777424157 Chr.2: 61877377-61877377 on Build GRCh38
rs955965513 Chr.2: 61877304-61877304 on Build GRCh38
rs933263055 Chr.2: 61877266-61877266 on Build GRCh38
rs958677163 Chr.2: 61877207-61877207 on Build GRCh38
rs4795 Chr.2: 61877401-61877401 on Build GRCh38
rs932454985 Chr.2: 61877331-61877331 on Build GRCh38
rs1009530694 Chr.2: 61877315-61877315 on Build GRCh38
rs924553008 Chr.2: 61877287-61877288 on Build GRCh38
rs560527802 Chr.2: 61877359-61877359 on Build GRCh38
rs752256824 Chr.2: 61877370-61877370 on Build GRCh38
rs768950542 Chr.2: 61877276-61877276 on Build GRCh38
rs139589025 Chr.2: 61877398-61877398 on Build GRCh38

View Full Product Details