Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4955430-4955681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO3
Entrez Gene ID
2027
Gene Name
enolase 3
Gene Aliases
GSD13, MSE
UniGene
Hs.224171
Gene Chromosome Location
Chr.17: 4949182-4957131 on Build GRCh38
Transcripts
NM_001193503.1 NM_001976.4 NM_053013.3 XM_005256521.2 XM_011523729.1 XM_017024346.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4130387 c.724A>T p.I242F Chr.17: 4955463-4955463 on Build GRCh38
6081513 c.775C>G p.L259V Chr.17: 4955514-4955514 on Build GRCh38
7287683 c.698C>T p.A233V Chr.17: 4955437-4955437 on Build GRCh38
4382985 c.772G>T p.D258Y Chr.17: 4955511-4955511 on Build GRCh38
6654088 c.829G>A p.G277R Chr.17: 4955568-4955568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886053166 Chr.17: 4955524-4955524 on Build GRCh38
rs199785068 Chr.17: 4955568-4955568 on Build GRCh38
rs377257809 Chr.17: 4955547-4955547 on Build GRCh38
rs886053167 Chr.17: 4955527-4955528 on Build GRCh38
rs548538086 Chr.17: 4955525-4955526 on Build GRCh38
rs766583341 Chr.17: 4955465-4955465 on Build GRCh38
rs188548745 Chr.17: 4955510-4955510 on Build GRCh38
rs763526971 Chr.17: 4955464-4955464 on Build GRCh38
rs756256626 Chr.17: 4955475-4955475 on Build GRCh38
rs760982486 Chr.17: 4955536-4955536 on Build GRCh38
rs149830213 Chr.17: 4955638-4955638 on Build GRCh38
rs374804367 Chr.17: 4955625-4955625 on Build GRCh38
rs780920675 Chr.17: 4955618-4955618 on Build GRCh38
rs769513367 Chr.17: 4955566-4955566 on Build GRCh38
rs368063034 Chr.17: 4955615-4955615 on Build GRCh38
rs144800857 Chr.17: 4955653-4955653 on Build GRCh38
rs144524946 Chr.17: 4955544-4955544 on Build GRCh38
rs1038733889 Chr.17: 4955599-4955599 on Build GRCh38
rs779920188 Chr.17: 4955573-4955573 on Build GRCh38
rs149301373 Chr.17: 4955541-4955541 on Build GRCh38
rs1029287688 Chr.17: 4955468-4955468 on Build GRCh38
rs776265768 Chr.17: 4955504-4955504 on Build GRCh38
rs561878819 Chr.17: 4955499-4955499 on Build GRCh38
rs772969351 Chr.17: 4955497-4955497 on Build GRCh38
rs754910286 Chr.17: 4955617-4955617 on Build GRCh38
rs753008114 Chr.17: 4955474-4955474 on Build GRCh38
rs773582449 Chr.17: 4955459-4955459 on Build GRCh38
rs772499633 Chr.17: 4955578-4955578 on Build GRCh38
rs772780015 Chr.17: 4955511-4955511 on Build GRCh38
rs201670239 Chr.17: 4955466-4955466 on Build GRCh38
rs754758809 Chr.17: 4955545-4955545 on Build GRCh38
rs753975550 Chr.17: 4955540-4955540 on Build GRCh38
rs35119507 Chr.17: 4955567-4955567 on Build GRCh38
rs778664924 Chr.17: 4955447-4955447 on Build GRCh38
rs764872268 Chr.17: 4955455-4955455 on Build GRCh38
rs763327091 Chr.17: 4955513-4955513 on Build GRCh38
rs191645770 Chr.17: 4955616-4955616 on Build GRCh38
rs768482299 Chr.17: 4955500-4955500 on Build GRCh38
rs111931609 Chr.17: 4955631-4955631 on Build GRCh38
rs370822982 Chr.17: 4955571-4955571 on Build GRCh38
rs745727037 Chr.17: 4955489-4955489 on Build GRCh38
rs369366731 Chr.17: 4955453-4955453 on Build GRCh38
rs370699915 Chr.17: 4955612-4955612 on Build GRCh38
rs147327991 Chr.17: 4955607-4955607 on Build GRCh38
rs375776604 Chr.17: 4955470-4955470 on Build GRCh38
rs771888314 Chr.17: 4955566-4955572 on Build GRCh38
rs200479387 Chr.17: 4955476-4955476 on Build GRCh38
rs935631054 Chr.17: 4955543-4955543 on Build GRCh38
rs370752383 Chr.17: 4955620-4955620 on Build GRCh38
rs747728478 Chr.17: 4955548-4955548 on Build GRCh38
rs75748087 Chr.17: 4955528-4955528 on Build GRCh38
rs776189298 Chr.17: 4955645-4955645 on Build GRCh38
rs376782089 Chr.17: 4955592-4955592 on Build GRCh38
rs748865733 Chr.17: 4955527-4955527 on Build GRCh38
rs375868102 Chr.17: 4955643-4955643 on Build GRCh38
rs372643524 Chr.17: 4955637-4955637 on Build GRCh38
rs766929167 Chr.17: 4955519-4955519 on Build GRCh38
rs761950302 Chr.17: 4955611-4955611 on Build GRCh38
rs776029162 Chr.17: 4955583-4955583 on Build GRCh38
rs758391667 Chr.17: 4955495-4955495 on Build GRCh38
rs768320684 Chr.17: 4955449-4955449 on Build GRCh38
rs182385874 Chr.17: 4955644-4955644 on Build GRCh38
rs760762359 Chr.17: 4955585-4955585 on Build GRCh38
rs199801526 Chr.17: 4955593-4955593 on Build GRCh38
rs777450548 Chr.17: 4955626-4955626 on Build GRCh38
rs111385580 Chr.17: 4955605-4955605 on Build GRCh38

View Full Product Details