Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132141647-132141906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX51
Entrez Gene ID
317781
Gene Name
DEAD-box helicase 51
Gene Aliases
-
UniGene
Hs.445168
Gene Chromosome Location
Chr.12: 132136594-132144335 on Build GRCh38
Transcripts
NM_175066.3 XM_011538256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3753064 c.965C>T p.A322V Chr.12: 132141880-132141880 on Build GRCh38
4040315 c.947C>T p.T316M Chr.12: 132141898-132141898 on Build GRCh38
430761 c.995+1G>C p.? Chr.12: 132141849-132141849 on Build GRCh38
6847208 c.968A>C p.K323T Chr.12: 132141877-132141877 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769573650 Chr.12: 132141838-132141838 on Build GRCh38
rs542443732 Chr.12: 132141678-132141678 on Build GRCh38
rs1031960552 Chr.12: 132141782-132141782 on Build GRCh38
rs1014966058 Chr.12: 132141722-132141722 on Build GRCh38
rs749618841 Chr.12: 132141821-132141821 on Build GRCh38
rs1021551368 Chr.12: 132141825-132141825 on Build GRCh38
rs552056437 Chr.12: 132141845-132141845 on Build GRCh38
rs573043189 Chr.12: 132141690-132141690 on Build GRCh38
rs377623915 Chr.12: 132141807-132141807 on Build GRCh38
rs530393568 Chr.12: 132141714-132141714 on Build GRCh38
rs545654209 Chr.12: 132141764-132141764 on Build GRCh38
rs923372915 Chr.12: 132141842-132141842 on Build GRCh38
rs758154765 Chr.12: 132141822-132141822 on Build GRCh38
rs753516664 Chr.12: 132141685-132141685 on Build GRCh38
rs942263821 Chr.12: 132141719-132141719 on Build GRCh38
rs1027463342 Chr.12: 132141700-132141700 on Build GRCh38
rs767968422 Chr.12: 132141804-132141804 on Build GRCh38
rs756510998 Chr.12: 132141817-132141817 on Build GRCh38
rs763896404 Chr.12: 132141673-132141673 on Build GRCh38
rs553278189 Chr.12: 132141689-132141689 on Build GRCh38
rs1038009911 Chr.12: 132141730-132141730 on Build GRCh38
rs914750584 Chr.12: 132141805-132141805 on Build GRCh38
rs566406256 Chr.12: 132141766-132141766 on Build GRCh38
rs767640556 Chr.12: 132141856-132141856 on Build GRCh38
rs560940157 Chr.12: 132141836-132141837 on Build GRCh38
rs573796850 Chr.12: 132141668-132141668 on Build GRCh38
rs750410878 Chr.12: 132141860-132141860 on Build GRCh38
rs774545137 Chr.12: 132141849-132141849 on Build GRCh38
rs116281803 Chr.12: 132141882-132141882 on Build GRCh38
rs769235366 Chr.12: 132141738-132141738 on Build GRCh38
rs746564827 Chr.12: 132141830-132141830 on Build GRCh38
rs753397339 Chr.12: 132141849-132141849 on Build GRCh38
rs117087653 Chr.12: 132141695-132141695 on Build GRCh38
rs60927391 Chr.12: 132141880-132141880 on Build GRCh38
rs370652076 Chr.12: 132141831-132141831 on Build GRCh38
rs756805080 Chr.12: 132141884-132141884 on Build GRCh38
rs528794538 Chr.12: 132141774-132141774 on Build GRCh38
rs1012057980 Chr.12: 132141696-132141696 on Build GRCh38
rs933492194 Chr.12: 132141713-132141713 on Build GRCh38
rs866867846 Chr.12: 132141873-132141873 on Build GRCh38
rs112903364 Chr.12: 132141744-132141744 on Build GRCh38
rs990286020 Chr.12: 132141779-132141779 on Build GRCh38
rs745835481 Chr.12: 132141834-132141834 on Build GRCh38
rs201397300 Chr.12: 132141850-132141850 on Build GRCh38
rs777533210 Chr.12: 132141824-132141824 on Build GRCh38
rs761964646 Chr.12: 132141875-132141875 on Build GRCh38
rs559059187 Chr.12: 132141868-132141868 on Build GRCh38
rs1013055473 Chr.12: 132141733-132141733 on Build GRCh38
rs756666377 Chr.12: 132141861-132141861 on Build GRCh38
rs757339976 Chr.12: 132141799-132141799 on Build GRCh38
rs922210706 Chr.12: 132141741-132141741 on Build GRCh38
rs564541249 Chr.12: 132141704-132141704 on Build GRCh38
rs766951147 Chr.12: 132141864-132141864 on Build GRCh38
rs560803524 Chr.12: 132141739-132141739 on Build GRCh38
rs147746512 Chr.12: 132141717-132141717 on Build GRCh38
rs754293715 Chr.12: 132141878-132141878 on Build GRCh38
rs898179210 Chr.12: 132141732-132141732 on Build GRCh38
rs895043386 Chr.12: 132141855-132141855 on Build GRCh38
rs780780937 Chr.12: 132141886-132141886 on Build GRCh38
rs776495247 Chr.12: 132141833-132141833 on Build GRCh38
rs983600829 Chr.12: 132141872-132141872 on Build GRCh38
rs764349900 Chr.12: 132141846-132141846 on Build GRCh38
rs1002700673 Chr.12: 132141781-132141781 on Build GRCh38

View Full Product Details