Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136369807-136370032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD9
Entrez Gene ID
64170
Gene Name
caspase recruitment domain family member 9
Gene Aliases
CANDF2, hCARD9
UniGene
Hs.694071
Gene Chromosome Location
Chr.9: 136363956-136373681 on Build GRCh38
Transcripts
NM_052813.4 NM_052814.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6620170 c.952-6delC p.? Chr.9: 136369881-136369881 on Build GRCh38
6027725 c.998G>A p.R333H Chr.9: 136369829-136369829 on Build GRCh38
4785390 c.1002G>T p.K334N Chr.9: 136369825-136369825 on Build GRCh38
6620169 c.952-6delC p.? Chr.9: 136369881-136369881 on Build GRCh38
4626386 c.991G>T p.A331S Chr.9: 136369836-136369836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758654216 Chr.9: 136369883-136369883 on Build GRCh38
rs374601657 Chr.9: 136369858-136369858 on Build GRCh38
rs761950582 Chr.9: 136369861-136369863 on Build GRCh38
rs180714639 Chr.9: 136369967-136369967 on Build GRCh38
rs112626270 Chr.9: 136369927-136369927 on Build GRCh38
rs148158774 Chr.9: 136369867-136369867 on Build GRCh38
rs373264509 Chr.9: 136369864-136369864 on Build GRCh38
rs774264495 Chr.9: 136369853-136369853 on Build GRCh38
rs755663524 Chr.9: 136369861-136369861 on Build GRCh38
rs555805929 Chr.9: 136369848-136369848 on Build GRCh38
rs1039812317 Chr.9: 136369902-136369902 on Build GRCh38
rs951178457 Chr.9: 136369998-136369998 on Build GRCh38
rs541356831 Chr.9: 136369887-136369887 on Build GRCh38
rs774389606 Chr.9: 136369914-136369914 on Build GRCh38
rs144114150 Chr.9: 136369941-136369941 on Build GRCh38
rs769988935 Chr.9: 136369852-136369852 on Build GRCh38
rs147295759 Chr.9: 136369951-136369951 on Build GRCh38
rs918697942 Chr.9: 136369997-136369997 on Build GRCh38
rs755467048 Chr.9: 136369871-136369871 on Build GRCh38
rs917706972 Chr.9: 136370007-136370007 on Build GRCh38
rs368069693 Chr.9: 136369891-136369891 on Build GRCh38
rs986663077 Chr.9: 136370009-136370009 on Build GRCh38
rs774103518 Chr.9: 136369990-136369990 on Build GRCh38
rs962750940 Chr.9: 136369970-136369970 on Build GRCh38
rs1020044015 Chr.9: 136369935-136369935 on Build GRCh38
rs772922499 Chr.9: 136369921-136369921 on Build GRCh38
rs960977830 Chr.9: 136369948-136369948 on Build GRCh38
rs533239815 Chr.9: 136369943-136369943 on Build GRCh38
rs950548740 Chr.9: 136369962-136369962 on Build GRCh38
rs1026550571 Chr.9: 136369952-136369952 on Build GRCh38
rs749994446 Chr.9: 136369860-136369860 on Build GRCh38
rs773193573 Chr.9: 136369913-136369914 on Build GRCh38
rs370912459 Chr.9: 136369849-136369849 on Build GRCh38
rs747130473 Chr.9: 136369886-136369886 on Build GRCh38
rs761411125 Chr.9: 136369856-136369856 on Build GRCh38
rs772118143 Chr.9: 136369864-136369866 on Build GRCh38
rs753078506 Chr.9: 136369877-136369877 on Build GRCh38
rs143669690 Chr.9: 136369851-136369851 on Build GRCh38
rs773208112 Chr.9: 136369834-136369834 on Build GRCh38
rs775284320 Chr.9: 136369858-136369860 on Build GRCh38
rs778065099 Chr.9: 136369884-136369884 on Build GRCh38
rs781405446 Chr.9: 136369897-136369897 on Build GRCh38
rs974534700 Chr.9: 136369983-136369983 on Build GRCh38
rs1035221535 Chr.9: 136369944-136369944 on Build GRCh38
rs761690518 Chr.9: 136369916-136369916 on Build GRCh38
rs779301338 Chr.9: 136369874-136369874 on Build GRCh38
rs950283552 Chr.9: 136369882-136369882 on Build GRCh38
rs371476104 Chr.9: 136369905-136369905 on Build GRCh38
rs768682407 Chr.9: 136369913-136369913 on Build GRCh38
rs771707041 Chr.9: 136369917-136369917 on Build GRCh38
rs376536772 Chr.9: 136369919-136369919 on Build GRCh38

View Full Product Details