Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10555092-10555339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRI1
Entrez Gene ID
65095
Gene Name
KRI1 homolog
Gene Aliases
-
UniGene
Hs.709967
Gene Chromosome Location
Chr.19: 10553085-10566031 on Build GRCh38
Transcripts
NM_023008.3 XM_011528190.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4131173 c.1692C>A p.C564* Chr.19: 10555293-10555293 on Build GRCh38
7105380 c.1793G>A p.R598Q Chr.19: 10555093-10555093 on Build GRCh38
5626451 c.1767G>A p.K589K Chr.19: 10555119-10555119 on Build GRCh38
7155100 c.1757C>T p.S586L Chr.19: 10555129-10555129 on Build GRCh38
6083851 c.1785A>C p.S595S Chr.19: 10555101-10555101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780818104 Chr.19: 10555225-10555225 on Build GRCh38
rs745379463 Chr.19: 10555245-10555246 on Build GRCh38
rs1055287196 Chr.19: 10555272-10555272 on Build GRCh38
rs765168060 Chr.19: 10555165-10555165 on Build GRCh38
rs765537702 Chr.19: 10555267-10555268 on Build GRCh38
rs1020254071 Chr.19: 10555128-10555128 on Build GRCh38
rs763167226 Chr.19: 10555228-10555228 on Build GRCh38
rs780685407 Chr.19: 10555239-10555240 on Build GRCh38
rs746469131 Chr.19: 10555209-10555209 on Build GRCh38
rs984572646 Chr.19: 10555236-10555236 on Build GRCh38
rs766667598 Chr.19: 10555259-10555260 on Build GRCh38
rs776788582 Chr.19: 10555288-10555288 on Build GRCh38
rs965945209 Chr.19: 10555146-10555146 on Build GRCh38
rs138703013 Chr.19: 10555308-10555308 on Build GRCh38
rs774651293 Chr.19: 10555229-10555229 on Build GRCh38
rs752993431 Chr.19: 10555273-10555274 on Build GRCh38
rs753228518 Chr.19: 10555238-10555239 on Build GRCh38
rs369389358 Chr.19: 10555235-10555236 on Build GRCh38
rs375415935 Chr.19: 10555310-10555310 on Build GRCh38
rs762018490 Chr.19: 10555289-10555289 on Build GRCh38
rs773489973 Chr.19: 10555256-10555260 on Build GRCh38
rs750262453 Chr.19: 10555187-10555187 on Build GRCh38
rs34470828 Chr.19: 10555237-10555238 on Build GRCh38
rs144763482 Chr.19: 10555295-10555295 on Build GRCh38
rs374926114 Chr.19: 10555202-10555203 on Build GRCh38
rs764356582 Chr.19: 10555155-10555155 on Build GRCh38
rs866649472 Chr.19: 10555166-10555166 on Build GRCh38
rs114840087 Chr.19: 10555262-10555262 on Build GRCh38
rs754041629 Chr.19: 10555264-10555268 on Build GRCh38
rs113400152 Chr.19: 10555248-10555248 on Build GRCh38
rs149026065 Chr.19: 10555127-10555127 on Build GRCh38
rs772063879 Chr.19: 10555252-10555253 on Build GRCh38
rs7251746 Chr.19: 10555275-10555275 on Build GRCh38
rs779858576 Chr.19: 10555208-10555208 on Build GRCh38
rs760798550 Chr.19: 10555253-10555253 on Build GRCh38
rs796266209 Chr.19: 10555235-10555236 on Build GRCh38
rs375101497 Chr.19: 10555265-10555265 on Build GRCh38
rs774729864 Chr.19: 10555137-10555137 on Build GRCh38
rs761851198 Chr.19: 10555251-10555252 on Build GRCh38
rs572476315 Chr.19: 10555252-10555252 on Build GRCh38
rs1013747030 Chr.19: 10555284-10555284 on Build GRCh38
rs953908052 Chr.19: 10555188-10555188 on Build GRCh38
rs144204938 Chr.19: 10555268-10555268 on Build GRCh38
rs143028139 Chr.19: 10555148-10555148 on Build GRCh38
rs759759044 Chr.19: 10555263-10555264 on Build GRCh38
rs199581846 Chr.19: 10555210-10555210 on Build GRCh38
rs142680117 Chr.19: 10555152-10555152 on Build GRCh38
rs543106938 Chr.19: 10555267-10555267 on Build GRCh38
rs539354663 Chr.19: 10555239-10555239 on Build GRCh38
rs562739044 Chr.19: 10555300-10555300 on Build GRCh38
rs760822112 Chr.19: 10555256-10555257 on Build GRCh38
rs759410881 Chr.19: 10555315-10555315 on Build GRCh38
rs771264894 Chr.19: 10555227-10555227 on Build GRCh38
rs774809000 Chr.19: 10555131-10555131 on Build GRCh38
rs754503756 Chr.19: 10555222-10555222 on Build GRCh38
rs757118490 Chr.19: 10555157-10555157 on Build GRCh38
rs141953114 Chr.19: 10555222-10555223 on Build GRCh38
rs377006523 Chr.19: 10555251-10555251 on Build GRCh38
rs778241796 Chr.19: 10555296-10555296 on Build GRCh38
rs967147967 Chr.19: 10555147-10555147 on Build GRCh38
rs774146115 Chr.19: 10555248-10555249 on Build GRCh38
rs751517530 Chr.19: 10555266-10555266 on Build GRCh38
rs769496306 Chr.19: 10555246-10555247 on Build GRCh38
rs767445525 Chr.19: 10555317-10555317 on Build GRCh38
rs148315651 Chr.19: 10555256-10555256 on Build GRCh38
rs373535536 Chr.19: 10555291-10555291 on Build GRCh38
rs765585165 Chr.19: 10555264-10555264 on Build GRCh38
rs561218567 Chr.19: 10555270-10555270 on Build GRCh38
rs769303956 Chr.19: 10555280-10555280 on Build GRCh38
rs747838728 Chr.19: 10555226-10555226 on Build GRCh38
rs758313205 Chr.19: 10555191-10555191 on Build GRCh38
rs749985732 Chr.19: 10555300-10555301 on Build GRCh38
rs114277769 Chr.19: 10555250-10555250 on Build GRCh38
rs752161333 Chr.19: 10555318-10555318 on Build GRCh38
rs772367586 Chr.19: 10555274-10555274 on Build GRCh38
rs143447983 Chr.19: 10555118-10555118 on Build GRCh38
rs767933842 Chr.19: 10555145-10555145 on Build GRCh38
rs71162094 Chr.19: 10555247-10555248 on Build GRCh38
rs199551803 Chr.19: 10555117-10555117 on Build GRCh38
rs554182935 Chr.19: 10555245-10555245 on Build GRCh38
rs747533021 Chr.19: 10555278-10555278 on Build GRCh38
rs758971458 Chr.19: 10555279-10555283 on Build GRCh38
rs6146467 Chr.19: 10555236-10555237 on Build GRCh38
rs754175130 Chr.19: 10555156-10555156 on Build GRCh38
rs755725872 Chr.19: 10555269-10555269 on Build GRCh38
rs117140161 Chr.19: 10555257-10555257 on Build GRCh38

View Full Product Details