Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138071602-138071875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DZIP1L
Entrez Gene ID
199221
Gene Name
DAZ interacting zinc finger protein 1 like
Gene Aliases
DZIP2
UniGene
Hs.351403
Gene Chromosome Location
Chr.3: 138061985-138115862 on Build GRCh38
Transcripts
NM_001170538.1 NM_173543.2 XM_005247198.3 XM_005247200.4 XM_006713526.1 XM_006713527.3 XM_011512518.2 XM_011512520.2 XM_011512521.2 XM_011512522.1 XM_011512524.2 XM_017005841.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3587924 c.1479C>T p.V493V Chr.3: 138071779-138071779 on Build GRCh38
4864791 c.1433G>A p.G478E Chr.3: 138071825-138071825 on Build GRCh38
3587922 c.1500G>A p.R500R Chr.3: 138071758-138071758 on Build GRCh38
6650967 c.1523T>C p.L508P Chr.3: 138071735-138071735 on Build GRCh38
3587927 c.1440G>T p.S480S Chr.3: 138071818-138071818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529011516 Chr.3: 138071724-138071724 on Build GRCh38
rs769667583 Chr.3: 138071846-138071846 on Build GRCh38
rs774066560 Chr.3: 138071632-138071632 on Build GRCh38
rs151287945 Chr.3: 138071746-138071746 on Build GRCh38
rs749943377 Chr.3: 138071788-138071788 on Build GRCh38
rs548651176 Chr.3: 138071747-138071747 on Build GRCh38
rs761584698 Chr.3: 138071755-138071755 on Build GRCh38
rs758893246 Chr.3: 138071764-138071764 on Build GRCh38
rs757890112 Chr.3: 138071797-138071797 on Build GRCh38
rs375025881 Chr.3: 138071665-138071665 on Build GRCh38
rs538574418 Chr.3: 138071779-138071779 on Build GRCh38
rs17856977 Chr.3: 138071672-138071672 on Build GRCh38
rs768525363 Chr.3: 138071744-138071744 on Build GRCh38
rs746836568 Chr.3: 138071726-138071726 on Build GRCh38
rs756795805 Chr.3: 138071708-138071708 on Build GRCh38
rs751968525 Chr.3: 138071694-138071694 on Build GRCh38
rs540859814 Chr.3: 138071655-138071655 on Build GRCh38
rs773167708 Chr.3: 138071848-138071848 on Build GRCh38
rs765168818 Chr.3: 138071668-138071668 on Build GRCh38
rs900747145 Chr.3: 138071836-138071836 on Build GRCh38
rs966215307 Chr.3: 138071637-138071637 on Build GRCh38
rs918332522 Chr.3: 138071666-138071666 on Build GRCh38
rs768572145 Chr.3: 138071813-138071813 on Build GRCh38
rs747959405 Chr.3: 138071819-138071819 on Build GRCh38
rs889086878 Chr.3: 138071799-138071799 on Build GRCh38
rs1010905067 Chr.3: 138071770-138071770 on Build GRCh38
rs184030146 Chr.3: 138071760-138071760 on Build GRCh38
rs915796405 Chr.3: 138071733-138071733 on Build GRCh38
rs766608532 Chr.3: 138071691-138071691 on Build GRCh38
rs148790083 Chr.3: 138071717-138071717 on Build GRCh38
rs556276730 Chr.3: 138071780-138071780 on Build GRCh38
rs202011537 Chr.3: 138071817-138071817 on Build GRCh38
rs147779217 Chr.3: 138071703-138071703 on Build GRCh38
rs375872409 Chr.3: 138071769-138071769 on Build GRCh38
rs781686459 Chr.3: 138071700-138071700 on Build GRCh38
rs745447444 Chr.3: 138071633-138071633 on Build GRCh38
rs779878262 Chr.3: 138071802-138071802 on Build GRCh38
rs753058491 Chr.3: 138071775-138071775 on Build GRCh38
rs775487339 Chr.3: 138071649-138071649 on Build GRCh38
rs745372080 Chr.3: 138071722-138071722 on Build GRCh38
rs776674487 Chr.3: 138071661-138071661 on Build GRCh38
rs150466957 Chr.3: 138071774-138071774 on Build GRCh38
rs140443333 Chr.3: 138071677-138071677 on Build GRCh38
rs929750748 Chr.3: 138071688-138071688 on Build GRCh38
rs144186460 Chr.3: 138071698-138071698 on Build GRCh38
rs747614039 Chr.3: 138071810-138071810 on Build GRCh38
rs566510085 Chr.3: 138071759-138071759 on Build GRCh38
rs147166319 Chr.3: 138071656-138071656 on Build GRCh38
rs746599482 Chr.3: 138071803-138071803 on Build GRCh38
rs140548803 Chr.3: 138071766-138071766 on Build GRCh38
rs750660072 Chr.3: 138071676-138071676 on Build GRCh38
rs771752719 Chr.3: 138071638-138071638 on Build GRCh38

View Full Product Details