Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74751126-74751399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP1A2
Entrez Gene ID
1544
Gene Name
cytochrome P450 family 1 subfamily A member 2
Gene Aliases
CP12, P3-450, P450(PA)
UniGene
Hs.1361
Gene Chromosome Location
Chr.15: 74748843-74756600 on Build GRCh38
Transcripts
NM_000761.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
964893 c.857C>A p.A286D Chr.15: 74751214-74751214 on Build GRCh38
378204 c.927C>A p.N309K Chr.15: 74751284-74751284 on Build GRCh38
3794401 c.887G>C p.R296T Chr.15: 74751244-74751244 on Build GRCh38
3503979 c.844G>A p.D282N Chr.15: 74751201-74751201 on Build GRCh38
4561158 c.872G>A p.S291N Chr.15: 74751229-74751229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780704581 Chr.15: 74751277-74751277 on Build GRCh38
rs751754228 Chr.15: 74751163-74751163 on Build GRCh38
rs777121760 Chr.15: 74751358-74751358 on Build GRCh38
rs867784817 Chr.15: 74751306-74751306 on Build GRCh38
rs866210877 Chr.15: 74751240-74751240 on Build GRCh38
rs750707671 Chr.15: 74751168-74751168 on Build GRCh38
rs181281039 Chr.15: 74751349-74751349 on Build GRCh38
rs767248026 Chr.15: 74751357-74751357 on Build GRCh38
rs763014583 Chr.15: 74751153-74751153 on Build GRCh38
rs774506756 Chr.15: 74751302-74751302 on Build GRCh38
rs908670064 Chr.15: 74751146-74751146 on Build GRCh38
rs760143725 Chr.15: 74751329-74751329 on Build GRCh38
rs990290810 Chr.15: 74751343-74751343 on Build GRCh38
rs995853768 Chr.15: 74751321-74751321 on Build GRCh38
rs752325506 Chr.15: 74751182-74751182 on Build GRCh38
rs762114920 Chr.15: 74751224-74751224 on Build GRCh38
rs55860387 Chr.15: 74751308-74751308 on Build GRCh38
rs756936781 Chr.15: 74751262-74751262 on Build GRCh38
rs751145178 Chr.15: 74751345-74751345 on Build GRCh38
rs28399419 Chr.15: 74751328-74751328 on Build GRCh38
rs764355637 Chr.15: 74751154-74751154 on Build GRCh38
rs745581564 Chr.15: 74751213-74751213 on Build GRCh38
rs1055486806 Chr.15: 74751187-74751187 on Build GRCh38
rs771876603 Chr.15: 74751226-74751226 on Build GRCh38
rs200115475 Chr.15: 74751166-74751166 on Build GRCh38
rs755877881 Chr.15: 74751284-74751284 on Build GRCh38
rs775392030 Chr.15: 74751217-74751217 on Build GRCh38
rs1015783320 Chr.15: 74751364-74751364 on Build GRCh38
rs773632297 Chr.15: 74751228-74751228 on Build GRCh38
rs17861157 Chr.15: 74751251-74751251 on Build GRCh38
rs765595047 Chr.15: 74751336-74751336 on Build GRCh38
rs878910455 Chr.15: 74751251-74751252 on Build GRCh38
rs368776150 Chr.15: 74751208-74751208 on Build GRCh38
rs745733376 Chr.15: 74751282-74751282 on Build GRCh38
rs35796837 Chr.15: 74751252-74751252 on Build GRCh38
rs780645075 Chr.15: 74751177-74751177 on Build GRCh38
rs767051591 Chr.15: 74751238-74751238 on Build GRCh38
rs780002436 Chr.15: 74751286-74751286 on Build GRCh38
rs746031187 Chr.15: 74751211-74751211 on Build GRCh38
rs940101068 Chr.15: 74751164-74751164 on Build GRCh38
rs267604322 Chr.15: 74751200-74751200 on Build GRCh38
rs761170528 Chr.15: 74751323-74751323 on Build GRCh38
rs774252909 Chr.15: 74751223-74751223 on Build GRCh38
rs34058039 Chr.15: 74751287-74751287 on Build GRCh38
rs1022231223 Chr.15: 74751304-74751304 on Build GRCh38
rs531577033 Chr.15: 74751218-74751218 on Build GRCh38
rs890187686 Chr.15: 74751239-74751239 on Build GRCh38
rs749252751 Chr.15: 74751293-74751293 on Build GRCh38
rs189723059 Chr.15: 74751327-74751327 on Build GRCh38
rs760946724 Chr.15: 74751231-74751231 on Build GRCh38
rs28399418 Chr.15: 74751297-74751297 on Build GRCh38
rs76407270 Chr.15: 74751170-74751170 on Build GRCh38
rs755719888 Chr.15: 74751361-74751361 on Build GRCh38
rs56205522 Chr.15: 74751374-74751374 on Build GRCh38
rs527553740 Chr.15: 74751173-74751173 on Build GRCh38
rs772408961 Chr.15: 74751318-74751318 on Build GRCh38
rs267604321 Chr.15: 74751199-74751199 on Build GRCh38
rs746644895 Chr.15: 74751201-74751201 on Build GRCh38
rs750975192 Chr.15: 74751253-74751253 on Build GRCh38
rs768687059 Chr.15: 74751221-74751221 on Build GRCh38
rs56037745 Chr.15: 74751209-74751209 on Build GRCh38
rs45468096 Chr.15: 74751198-74751198 on Build GRCh38
rs773296531 Chr.15: 74751322-74751322 on Build GRCh38
rs559714610 Chr.15: 74751165-74751165 on Build GRCh38

View Full Product Details