Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201285182-201285407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP8
Entrez Gene ID
841
Gene Name
caspase 8
Gene Aliases
ALPS2B, CAP4, Casp-8, FLICE, MACH, MCH5
UniGene
Hs.599762
Gene Chromosome Location
Chr.2: 201233443-201287711 on Build GRCh38
Transcripts
NM_001080124.1 NM_001080125.1 NM_001228.4 NM_033355.3 NM_033356.3 NR_111983.1 XM_005246885.1 XM_005246886.1 XM_005246887.1 XM_005246888.1 XM_005246889.1 XM_005246890.3 XM_005246891.4 XM_005246892.1 XM_005246894.3 XM_006712789.1 XM_006712790.3 XM_011511969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5973539 c.1231C>T p.P411S Chr.2: 201285193-201285193 on Build GRCh38
5009943 c.1297G>A p.A433T Chr.2: 201285259-201285259 on Build GRCh38
1326524 c.1352C>T p.P451L Chr.2: 201285314-201285314 on Build GRCh38
1014880 c.1347G>A p.T449T Chr.2: 201285183-201285183 on Build GRCh38
6940631 c.1424_1447del24 p.E476_S483delEGTWYIQS Chr.2: 201285260-201285283 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746401108 Chr.2: 201285317-201285317 on Build GRCh38
rs758753163 Chr.2: 201285227-201285227 on Build GRCh38
rs966626638 Chr.2: 201285357-201285357 on Build GRCh38
rs752967014 Chr.2: 201285214-201285214 on Build GRCh38
rs771714764 Chr.2: 201285352-201285352 on Build GRCh38
rs759356977 Chr.2: 201285204-201285204 on Build GRCh38
rs747738525 Chr.2: 201285336-201285336 on Build GRCh38
rs535474839 Chr.2: 201285210-201285210 on Build GRCh38
rs34750049 Chr.2: 201285366-201285366 on Build GRCh38
rs746547222 Chr.2: 201285359-201285359 on Build GRCh38
rs587776665 Chr.2: 201285241-201285242 on Build GRCh38
rs760174966 Chr.2: 201285368-201285368 on Build GRCh38
rs764213462 Chr.2: 201285228-201285228 on Build GRCh38
rs752277547 Chr.2: 201285361-201285361 on Build GRCh38
rs781707597 Chr.2: 201285279-201285279 on Build GRCh38
rs769700812 Chr.2: 201285365-201285365 on Build GRCh38
rs751664523 Chr.2: 201285255-201285255 on Build GRCh38
rs766919472 Chr.2: 201285360-201285361 on Build GRCh38
rs369459056 Chr.2: 201285332-201285332 on Build GRCh38
rs931648756 Chr.2: 201285307-201285307 on Build GRCh38
rs768000891 Chr.2: 201285319-201285319 on Build GRCh38
rs777103308 Chr.2: 201285354-201285354 on Build GRCh38
rs375465590 Chr.2: 201285329-201285329 on Build GRCh38
rs1036564097 Chr.2: 201285369-201285369 on Build GRCh38
rs757665080 Chr.2: 201285269-201285269 on Build GRCh38
rs372712049 Chr.2: 201285363-201285363 on Build GRCh38

View Full Product Details