Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122841115-122841376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUZD1
Entrez Gene ID
50624
Gene Name
CUB and zona pellucida like domains 1
Gene Aliases
ERG-1, ERG1, ITMAP1, UO-44
UniGene
Hs.647182
Gene Chromosome Location
Chr.10: 122832155-122846175 on Build GRCh38
Transcripts
NM_022034.5 NR_037912.1 NR_037915.1

Gene Symbol
FAM24B-CUZD1
Entrez Gene ID
100533195
Gene Name
FAM24B-CUZD1 readthrough
Gene Aliases
-
UniGene
Hs.647182
Gene Chromosome Location
Chr.10: 122832155-122879641 on Build GRCh38
Transcripts
NM_022034.5 NR_037912.1 NR_037915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
915720 c.215T>G p.I72S Chr.10: 122841196-122841196 on Build GRCh38
3435349 c.104G>A p.S35N Chr.10: 122841307-122841307 on Build GRCh38
7090172 c.111G>T p.G37G Chr.10: 122841300-122841300 on Build GRCh38
6865796 c.196G>A p.E66K Chr.10: 122841215-122841215 on Build GRCh38
2020826 c.113delG p.G38fs*11 Chr.10: 122841298-122841298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779742489 Chr.10: 122841173-122841173 on Build GRCh38
rs754492546 Chr.10: 122841192-122841192 on Build GRCh38
rs372082723 Chr.10: 122841229-122841229 on Build GRCh38
rs367768151 Chr.10: 122841182-122841182 on Build GRCh38
rs765224448 Chr.10: 122841312-122841313 on Build GRCh38
rs112767873 Chr.10: 122841149-122841149 on Build GRCh38
rs749383852 Chr.10: 122841336-122841336 on Build GRCh38
rs752191102 Chr.10: 122841281-122841281 on Build GRCh38
rs764921462 Chr.10: 122841279-122841279 on Build GRCh38
rs754538690 Chr.10: 122841276-122841276 on Build GRCh38
rs767418253 Chr.10: 122841256-122841256 on Build GRCh38
rs1006746944 Chr.10: 122841224-122841224 on Build GRCh38
rs998313360 Chr.10: 122841272-122841272 on Build GRCh38
rs888101522 Chr.10: 122841230-122841230 on Build GRCh38
rs909541909 Chr.10: 122841292-122841292 on Build GRCh38
rs568710672 Chr.10: 122841139-122841139 on Build GRCh38
rs775774628 Chr.10: 122841335-122841335 on Build GRCh38
rs750604872 Chr.10: 122841306-122841306 on Build GRCh38
rs746046760 Chr.10: 122841243-122841243 on Build GRCh38
rs1050911949 Chr.10: 122841350-122841350 on Build GRCh38
rs374674283 Chr.10: 122841167-122841167 on Build GRCh38
rs201593810 Chr.10: 122841311-122841311 on Build GRCh38
rs763055684 Chr.10: 122841253-122841253 on Build GRCh38
rs754687984 Chr.10: 122841256-122841256 on Build GRCh38
rs766099629 Chr.10: 122841268-122841268 on Build GRCh38
rs778437302 Chr.10: 122841284-122841284 on Build GRCh38
rs753523432 Chr.10: 122841269-122841269 on Build GRCh38
rs760479790 Chr.10: 122841264-122841264 on Build GRCh38
rs375448676 Chr.10: 122841329-122841329 on Build GRCh38
rs551074034 Chr.10: 122841226-122841226 on Build GRCh38
rs758880808 Chr.10: 122841200-122841200 on Build GRCh38
rs769946487 Chr.10: 122841331-122841331 on Build GRCh38
rs769935598 Chr.10: 122841244-122841244 on Build GRCh38
rs755896246 Chr.10: 122841170-122841170 on Build GRCh38
rs368148874 Chr.10: 122841138-122841138 on Build GRCh38
rs778246579 Chr.10: 122841199-122841199 on Build GRCh38
rs369745931 Chr.10: 122841344-122841344 on Build GRCh38
rs896940943 Chr.10: 122841325-122841325 on Build GRCh38
rs34246902 Chr.10: 122841318-122841318 on Build GRCh38
rs759422721 Chr.10: 122841142-122841142 on Build GRCh38
rs768726266 Chr.10: 122841339-122841339 on Build GRCh38
rs747452055 Chr.10: 122841298-122841298 on Build GRCh38
rs776997506 Chr.10: 122841233-122841233 on Build GRCh38
rs753689743 Chr.10: 122841219-122841222 on Build GRCh38
rs35299879 Chr.10: 122841282-122841282 on Build GRCh38
rs773123161 Chr.10: 122841260-122841260 on Build GRCh38
rs141881176 Chr.10: 122841248-122841248 on Build GRCh38
rs781648542 Chr.10: 122841316-122841316 on Build GRCh38

View Full Product Details