Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 125038741-125039002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0196
Entrez Gene ID
9897
Gene Name
KIAA0196
Gene Aliases
RTSC, RTSC1, SPG8
UniGene
Hs.270043
Gene Chromosome Location
Chr.8: 125024260-125091819 on Build GRCh38
Transcripts
NM_014846.3 XM_005251120.3 XM_011517409.1 XM_017014113.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3924610 c.2996C>T p.S999L Chr.8: 125038918-125038918 on Build GRCh38
461486 c.3034G>C p.E1012Q Chr.8: 125038880-125038880 on Build GRCh38
3644972 c.3001C>T p.P1001S Chr.8: 125038913-125038913 on Build GRCh38
6991195 c.3065T>C p.I1022T Chr.8: 125038849-125038849 on Build GRCh38
6565579 c.2966C>A p.A989E Chr.8: 125038948-125038948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769011883 Chr.8: 125038798-125038798 on Build GRCh38
rs770423051 Chr.8: 125038808-125038808 on Build GRCh38
rs763980360 Chr.8: 125038933-125038933 on Build GRCh38
rs775235313 Chr.8: 125038925-125038925 on Build GRCh38
rs749572396 Chr.8: 125038797-125038797 on Build GRCh38
rs762772290 Chr.8: 125038833-125038833 on Build GRCh38
rs139652594 Chr.8: 125038917-125038917 on Build GRCh38
rs760270136 Chr.8: 125038824-125038825 on Build GRCh38
rs371764965 Chr.8: 125038784-125038784 on Build GRCh38
rs767591968 Chr.8: 125038966-125038966 on Build GRCh38
rs983999629 Chr.8: 125038927-125038927 on Build GRCh38
rs528306856 Chr.8: 125038804-125038804 on Build GRCh38
rs749808080 Chr.8: 125038885-125038885 on Build GRCh38
rs746633398 Chr.8: 125038910-125038910 on Build GRCh38
rs35700689 Chr.8: 125038793-125038794 on Build GRCh38
rs34849329 Chr.8: 125038958-125038958 on Build GRCh38
rs991575383 Chr.8: 125038800-125038800 on Build GRCh38
rs780629887 Chr.8: 125038796-125038796 on Build GRCh38
rs746350762 Chr.8: 125038793-125038793 on Build GRCh38
rs971723373 Chr.8: 125038792-125038792 on Build GRCh38
rs142883794 Chr.8: 125038942-125038942 on Build GRCh38
rs755321757 Chr.8: 125038886-125038886 on Build GRCh38
rs769353866 Chr.8: 125038923-125038923 on Build GRCh38
rs200191392 Chr.8: 125038916-125038916 on Build GRCh38
rs780357095 Chr.8: 125038884-125038884 on Build GRCh38
rs767608029 Chr.8: 125038867-125038867 on Build GRCh38
rs779317231 Chr.8: 125038902-125038902 on Build GRCh38
rs770619335 Chr.8: 125038913-125038913 on Build GRCh38
rs756246696 Chr.8: 125038878-125038878 on Build GRCh38
rs141278432 Chr.8: 125038805-125038805 on Build GRCh38
rs770249644 Chr.8: 125038794-125038794 on Build GRCh38
rs759495858 Chr.8: 125038813-125038813 on Build GRCh38
rs763996850 Chr.8: 125038841-125038841 on Build GRCh38
rs763045985 Chr.8: 125038929-125038929 on Build GRCh38
rs150819213 Chr.8: 125038849-125038849 on Build GRCh38
rs774332269 Chr.8: 125038937-125038937 on Build GRCh38
rs750418643 Chr.8: 125038876-125038876 on Build GRCh38
rs752712933 Chr.8: 125038830-125038830 on Build GRCh38
rs776045038 Chr.8: 125038812-125038812 on Build GRCh38
rs568411535 Chr.8: 125038814-125038814 on Build GRCh38
rs765926045 Chr.8: 125038889-125038890 on Build GRCh38

View Full Product Details