Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95974155-95974428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAL
Entrez Gene ID
3034
Gene Name
histidine ammonia-lyase
Gene Aliases
HIS, HSTD
UniGene
Hs.190783
Gene Chromosome Location
Chr.12: 95972662-95996365 on Build GRCh38
Transcripts
NM_001258333.1 NM_001258334.1 NM_002108.3 XM_011538249.2 XM_017019246.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3782774 c.1954C>T p.P652S Chr.12: 95974252-95974252 on Build GRCh38
1579560 c.1955C>T p.P652L Chr.12: 95974251-95974251 on Build GRCh38
212166 c.1963G>T p.E655* Chr.12: 95974243-95974243 on Build GRCh38
1299982 c.1938G>A p.K646K Chr.12: 95974268-95974268 on Build GRCh38
6677510 c.1918delT p.S640fs*>18 Chr.12: 95974288-95974288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773263806 Chr.12: 95974351-95974351 on Build GRCh38
rs955249045 Chr.12: 95974287-95974288 on Build GRCh38
rs748512974 Chr.12: 95974346-95974346 on Build GRCh38
rs769152394 Chr.12: 95974330-95974330 on Build GRCh38
rs865904307 Chr.12: 95974252-95974252 on Build GRCh38
rs777205930 Chr.12: 95974339-95974339 on Build GRCh38
rs780634001 Chr.12: 95974326-95974326 on Build GRCh38
rs1012465644 Chr.12: 95974247-95974247 on Build GRCh38
rs904292772 Chr.12: 95974347-95974347 on Build GRCh38
rs773761824 Chr.12: 95974308-95974308 on Build GRCh38
rs865839161 Chr.12: 95974194-95974194 on Build GRCh38
rs200780494 Chr.12: 95974255-95974255 on Build GRCh38
rs148503094 Chr.12: 95974385-95974385 on Build GRCh38
rs762907413 Chr.12: 95974246-95974246 on Build GRCh38
rs775830260 Chr.12: 95974193-95974193 on Build GRCh38
rs144698271 Chr.12: 95974273-95974273 on Build GRCh38
rs778515949 Chr.12: 95974317-95974317 on Build GRCh38
rs539448301 Chr.12: 95974238-95974238 on Build GRCh38
rs574414004 Chr.12: 95974268-95974268 on Build GRCh38
rs750562351 Chr.12: 95974239-95974239 on Build GRCh38
rs769097289 Chr.12: 95974230-95974230 on Build GRCh38
rs1034089392 Chr.12: 95974380-95974380 on Build GRCh38
rs112610390 Chr.12: 95974250-95974250 on Build GRCh38
rs973991513 Chr.12: 95974364-95974364 on Build GRCh38
rs757807781 Chr.12: 95974324-95974324 on Build GRCh38
rs942857961 Chr.12: 95974227-95974227 on Build GRCh38
rs142634377 Chr.12: 95974366-95974366 on Build GRCh38
rs761132742 Chr.12: 95974214-95974214 on Build GRCh38
rs749836087 Chr.12: 95974348-95974348 on Build GRCh38
rs753623981 Chr.12: 95974271-95974271 on Build GRCh38
rs372136349 Chr.12: 95974231-95974231 on Build GRCh38
rs372760582 Chr.12: 95974393-95974393 on Build GRCh38
rs564880759 Chr.12: 95974322-95974322 on Build GRCh38
rs755949907 Chr.12: 95974258-95974258 on Build GRCh38
rs1031038163 Chr.12: 95974263-95974263 on Build GRCh38
rs1008021276 Chr.12: 95974337-95974337 on Build GRCh38
rs775500986 Chr.12: 95974391-95974391 on Build GRCh38
rs762063319 Chr.12: 95974233-95974233 on Build GRCh38
rs901383469 Chr.12: 95974251-95974251 on Build GRCh38
rs764003960 Chr.12: 95974396-95974396 on Build GRCh38
rs369075607 Chr.12: 95974348-95974348 on Build GRCh38
rs138504011 Chr.12: 95974236-95974236 on Build GRCh38

View Full Product Details