Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85801079-85801350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX4I1
Entrez Gene ID
1327
Gene Name
cytochrome c oxidase subunit 4I1
Gene Aliases
COX IV-1, COX4, COX4-1, COXIV, COXIV-1
UniGene
Hs.433419
Gene Chromosome Location
Chr.16: 85799567-85807003 on Build GRCh38
Transcripts
NM_001318786.1 NM_001318788.1 NM_001318794.1 NM_001318797.1 NM_001318802.1 NM_001861.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4846965 c.53C>G p.S18C Chr.16: 85801258-85801258 on Build GRCh38
4063355 c.13A>C p.R5R Chr.16: 85801218-85801218 on Build GRCh38
5706255 c.37C>T p.R13* Chr.16: 85801242-85801242 on Build GRCh38
4418235 c.7G>A p.A3T Chr.16: 85801212-85801212 on Build GRCh38
974527 c.64C>T p.R22* Chr.16: 85801269-85801269 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754772407 Chr.16: 85801256-85801256 on Build GRCh38
rs986078720 Chr.16: 85801120-85801120 on Build GRCh38
rs202146194 Chr.16: 85801270-85801270 on Build GRCh38
rs113661756 Chr.16: 85801279-85801279 on Build GRCh38
rs200718347 Chr.16: 85801168-85801168 on Build GRCh38
rs750534183 Chr.16: 85801228-85801228 on Build GRCh38
rs772666331 Chr.16: 85801283-85801283 on Build GRCh38
rs1026322677 Chr.16: 85801312-85801312 on Build GRCh38
rs11557187 Chr.16: 85801212-85801212 on Build GRCh38
rs187001915 Chr.16: 85801106-85801106 on Build GRCh38
rs749233847 Chr.16: 85801252-85801252 on Build GRCh38
rs574946540 Chr.16: 85801328-85801328 on Build GRCh38
rs568511286 Chr.16: 85801134-85801134 on Build GRCh38
rs776030186 Chr.16: 85801303-85801303 on Build GRCh38
rs971942769 Chr.16: 85801308-85801308 on Build GRCh38
rs756080790 Chr.16: 85801232-85801232 on Build GRCh38
rs183118839 Chr.16: 85801103-85801103 on Build GRCh38
rs199880666 Chr.16: 85801306-85801306 on Build GRCh38
rs1041068718 Chr.16: 85801222-85801222 on Build GRCh38
rs888984413 Chr.16: 85801290-85801291 on Build GRCh38
rs770474032 Chr.16: 85801301-85801301 on Build GRCh38
rs558251587 Chr.16: 85801179-85801179 on Build GRCh38
rs966420452 Chr.16: 85801131-85801131 on Build GRCh38
rs746541450 Chr.16: 85801297-85801297 on Build GRCh38
rs753295579 Chr.16: 85801315-85801315 on Build GRCh38
rs537535282 Chr.16: 85801220-85801220 on Build GRCh38
rs757478293 Chr.16: 85801223-85801223 on Build GRCh38
rs142776923 Chr.16: 85801260-85801260 on Build GRCh38
rs373552102 Chr.16: 85801214-85801214 on Build GRCh38
rs375272236 Chr.16: 85801198-85801198 on Build GRCh38
rs991112397 Chr.16: 85801148-85801148 on Build GRCh38
rs1010675152 Chr.16: 85801269-85801269 on Build GRCh38
rs768853744 Chr.16: 85801213-85801214 on Build GRCh38
rs771611429 Chr.16: 85801184-85801184 on Build GRCh38
rs777065486 Chr.16: 85801258-85801259 on Build GRCh38
rs751801715 Chr.16: 85801219-85801219 on Build GRCh38
rs765761702 Chr.16: 85801205-85801205 on Build GRCh38
rs773081275 Chr.16: 85801136-85801136 on Build GRCh38
rs367882670 Chr.16: 85801174-85801174 on Build GRCh38
rs372008173 Chr.16: 85801175-85801175 on Build GRCh38
rs550964101 Chr.16: 85801145-85801145 on Build GRCh38
rs759981105 Chr.16: 85801201-85801201 on Build GRCh38
rs191821030 Chr.16: 85801259-85801259 on Build GRCh38
rs901138897 Chr.16: 85801262-85801262 on Build GRCh38
rs148220134 Chr.16: 85801151-85801151 on Build GRCh38
rs747800437 Chr.16: 85801181-85801181 on Build GRCh38
rs781487417 Chr.16: 85801158-85801158 on Build GRCh38
rs774033649 Chr.16: 85801178-85801178 on Build GRCh38
rs1050471783 Chr.16: 85801302-85801302 on Build GRCh38
rs767646220 Chr.16: 85801226-85801226 on Build GRCh38
rs539727402 Chr.16: 85801164-85801164 on Build GRCh38
rs780164636 Chr.16: 85801248-85801248 on Build GRCh38
rs764402410 Chr.16: 85801217-85801217 on Build GRCh38
rs1045983779 Chr.16: 85801186-85801186 on Build GRCh38
rs757079313 Chr.16: 85801180-85801180 on Build GRCh38
rs762159019 Chr.16: 85801294-85801297 on Build GRCh38
rs981755528 Chr.16: 85801133-85801133 on Build GRCh38
rs985255305 Chr.16: 85801147-85801147 on Build GRCh38
rs539207920 Chr.16: 85801137-85801137 on Build GRCh38
rs756131951 Chr.16: 85801165-85801165 on Build GRCh38
rs1018860428 Chr.16: 85801130-85801130 on Build GRCh38

View Full Product Details