Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141361426-141361655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018922.2 NM_018923.2 NM_032096.1

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018922.2 NM_018923.2 NM_032096.1

Gene Symbol
PCDHGA3
Entrez Gene ID
56112
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
PCDH-GAMMA-A3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141344034-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018922.2 NM_018923.2 NM_032096.1

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141355025-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018922.2 NM_018923.2 NM_032096.1

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141350261-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018922.2 NM_018923.2 NM_032096.1

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141360136-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018922.2 NM_018923.2 NM_032096.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4166441 c.1404C>A p.G468G Chr.5: 141361539-141361539 on Build GRCh38
7196114 c.1294C>G p.L432V Chr.5: 141361429-141361429 on Build GRCh38
7192871 c.1444G>T p.G482C Chr.5: 141361579-141361579 on Build GRCh38
6169366 c.1490A>T p.K497M Chr.5: 141361625-141361625 on Build GRCh38
6741077 c.1512C>T p.Y504Y Chr.5: 141361647-141361647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754978831 Chr.5: 141361539-141361539 on Build GRCh38
rs368091522 Chr.5: 141361503-141361503 on Build GRCh38
rs548966955 Chr.5: 141361465-141361465 on Build GRCh38
rs199916536 Chr.5: 141361554-141361554 on Build GRCh38
rs761232947 Chr.5: 141361597-141361597 on Build GRCh38
rs781508901 Chr.5: 141361468-141361468 on Build GRCh38
rs919763957 Chr.5: 141361614-141361614 on Build GRCh38
rs759187963 Chr.5: 141361482-141361482 on Build GRCh38
rs534427524 Chr.5: 141361631-141361631 on Build GRCh38
rs765204837 Chr.5: 141361599-141361599 on Build GRCh38
rs765118611 Chr.5: 141361514-141361514 on Build GRCh38
rs773626542 Chr.5: 141361474-141361474 on Build GRCh38
rs751712279 Chr.5: 141361628-141361628 on Build GRCh38
rs747981041 Chr.5: 141361625-141361626 on Build GRCh38
rs758759325 Chr.5: 141361453-141361453 on Build GRCh38
rs372928108 Chr.5: 141361618-141361618 on Build GRCh38
rs748594092 Chr.5: 141361540-141361540 on Build GRCh38
rs753696787 Chr.5: 141361506-141361506 on Build GRCh38
rs780371360 Chr.5: 141361527-141361527 on Build GRCh38
rs749597862 Chr.5: 141361550-141361550 on Build GRCh38
rs990440167 Chr.5: 141361626-141361626 on Build GRCh38
rs771647276 Chr.5: 141361484-141361484 on Build GRCh38
rs370361048 Chr.5: 141361546-141361546 on Build GRCh38
rs768038039 Chr.5: 141361589-141361589 on Build GRCh38
rs746722919 Chr.5: 141361582-141361582 on Build GRCh38
rs751857437 Chr.5: 141361528-141361528 on Build GRCh38
rs374369531 Chr.5: 141361548-141361548 on Build GRCh38
rs748732304 Chr.5: 141361471-141361471 on Build GRCh38
rs142115917 Chr.5: 141361509-141361509 on Build GRCh38
rs775139870 Chr.5: 141361490-141361490 on Build GRCh38
rs571629532 Chr.5: 141361602-141361602 on Build GRCh38
rs537842997 Chr.5: 141361518-141361518 on Build GRCh38
rs776269447 Chr.5: 141361591-141361591 on Build GRCh38
rs778689212 Chr.5: 141361541-141361542 on Build GRCh38
rs1031903185 Chr.5: 141361616-141361616 on Build GRCh38
rs775084773 Chr.5: 141361558-141361558 on Build GRCh38
rs768400418 Chr.5: 141361530-141361532 on Build GRCh38
rs551885796 Chr.5: 141361524-141361524 on Build GRCh38
rs942275534 Chr.5: 141361447-141361447 on Build GRCh38
rs374176708 Chr.5: 141361473-141361473 on Build GRCh38
rs760056101 Chr.5: 141361494-141361494 on Build GRCh38
rs780424534 Chr.5: 141361454-141361454 on Build GRCh38
rs367687761 Chr.5: 141361606-141361606 on Build GRCh38
rs750174731 Chr.5: 141361516-141361516 on Build GRCh38
rs529112132 Chr.5: 141361456-141361456 on Build GRCh38

View Full Product Details