Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 155116260-155116533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MME
Entrez Gene ID
4311
Gene Name
membrane metallo-endopeptidase
Gene Aliases
CALLA, CD10, CMT2T, NEP, SFE
UniGene
Hs.307734
Gene Chromosome Location
Chr.3: 155024124-155183729 on Build GRCh38
Transcripts
NM_000902.3 NM_007287.2 NM_007288.2 NM_007289.2 XM_006713646.3 XM_006713647.3 XM_011512855.2 XM_011512856.2 XM_011512857.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1420147 c.412G>A p.A138T Chr.3: 155116532-155116532 on Build GRCh38
1752977 c.402G>C p.Q134H Chr.3: 155116522-155116522 on Build GRCh38
1327781 c.361G>A p.V121I Chr.3: 155116481-155116481 on Build GRCh38
6163744 c.385G>C p.D129H Chr.3: 155116505-155116505 on Build GRCh38
728627 c.376A>G p.K126E Chr.3: 155116496-155116496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024494453 Chr.3: 155116363-155116364 on Build GRCh38
rs757982719 Chr.3: 155116478-155116478 on Build GRCh38
rs746340868 Chr.3: 155116335-155116335 on Build GRCh38
rs746723545 Chr.3: 155116498-155116498 on Build GRCh38
rs753688812 Chr.3: 155116454-155116454 on Build GRCh38
rs926079283 Chr.3: 155116507-155116507 on Build GRCh38
rs955332880 Chr.3: 155116421-155116421 on Build GRCh38
rs772367510 Chr.3: 155116407-155116407 on Build GRCh38
rs1036544823 Chr.3: 155116426-155116426 on Build GRCh38
rs979227777 Chr.3: 155116491-155116491 on Build GRCh38
rs187010759 Chr.3: 155116439-155116439 on Build GRCh38
rs191913757 Chr.3: 155116352-155116352 on Build GRCh38
rs779697692 Chr.3: 155116479-155116479 on Build GRCh38
rs199523224 Chr.3: 155116508-155116508 on Build GRCh38
rs906048897 Chr.3: 155116364-155116364 on Build GRCh38
rs183031256 Chr.3: 155116364-155116364 on Build GRCh38
rs908864625 Chr.3: 155116386-155116386 on Build GRCh38
rs58186318 Chr.3: 155116490-155116490 on Build GRCh38
rs1020140269 Chr.3: 155116312-155116312 on Build GRCh38
rs528249649 Chr.3: 155116381-155116381 on Build GRCh38
rs191808045 Chr.3: 155116452-155116452 on Build GRCh38
rs372337211 Chr.3: 155116511-155116511 on Build GRCh38
rs200313798 Chr.3: 155116509-155116509 on Build GRCh38
rs778238089 Chr.3: 155116467-155116467 on Build GRCh38
rs892053752 Chr.3: 155116328-155116328 on Build GRCh38
rs757268431 Chr.3: 155116466-155116466 on Build GRCh38
rs201718664 Chr.3: 155116483-155116483 on Build GRCh38
rs745418124 Chr.3: 155116476-155116476 on Build GRCh38
rs976240999 Chr.3: 155116332-155116332 on Build GRCh38
rs866204763 Chr.3: 155116311-155116311 on Build GRCh38
rs963875874 Chr.3: 155116489-155116489 on Build GRCh38
rs965837036 Chr.3: 155116329-155116329 on Build GRCh38
rs200444530 Chr.3: 155116455-155116455 on Build GRCh38
rs748652992 Chr.3: 155116441-155116441 on Build GRCh38
rs576256723 Chr.3: 155116408-155116408 on Build GRCh38
rs200239992 Chr.3: 155116437-155116437 on Build GRCh38
rs988701608 Chr.3: 155116310-155116310 on Build GRCh38
rs202218776 Chr.3: 155116449-155116449 on Build GRCh38

View Full Product Details