Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19657658-19657931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH3A2
Entrez Gene ID
224
Gene Name
aldehyde dehydrogenase 3 family member A2
Gene Aliases
ALDH10, FALDH, SLS
UniGene
Hs.499886
Gene Chromosome Location
Chr.17: 19648136-19677596 on Build GRCh38
Transcripts
NM_000382.2 NM_001031806.1 XM_011523732.1 XM_011523733.1 XM_017024355.1 XM_017024356.1 XM_017024357.1 XM_017024358.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6322142 c.713G>T p.G238V Chr.17: 19657777-19657777 on Build GRCh38
6322141 c.713G>T p.G238V Chr.17: 19657777-19657777 on Build GRCh38
4883197 c.793G>T p.V265L Chr.17: 19657857-19657857 on Build GRCh38
4445330 c.681-3C>G p.? Chr.17: 19657742-19657742 on Build GRCh38
4445329 c.681-3C>G p.? Chr.17: 19657742-19657742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760371280 Chr.17: 19657722-19657722 on Build GRCh38
rs3034928 Chr.17: 19657706-19657713 on Build GRCh38
rs776513696 Chr.17: 19657854-19657854 on Build GRCh38
rs767994960 Chr.17: 19657697-19657704 on Build GRCh38
rs757359379 Chr.17: 19657862-19657862 on Build GRCh38
rs752758799 Chr.17: 19657763-19657763 on Build GRCh38
rs763105379 Chr.17: 19657879-19657879 on Build GRCh38
rs786204677 Chr.17: 19657867-19657867 on Build GRCh38
rs756554580 Chr.17: 19657796-19657796 on Build GRCh38
rs1057517080 Chr.17: 19657863-19657863 on Build GRCh38
rs1057517190 Chr.17: 19657833-19657834 on Build GRCh38
rs771641423 Chr.17: 19657717-19657717 on Build GRCh38
rs72547569 Chr.17: 19657862-19657862 on Build GRCh38
rs72547568 Chr.17: 19657797-19657797 on Build GRCh38
rs3841513 Chr.17: 19657699-19657706 on Build GRCh38
rs1033560863 Chr.17: 19657876-19657876 on Build GRCh38
rs775626554 Chr.17: 19657878-19657878 on Build GRCh38
rs372698552 Chr.17: 19657708-19657715 on Build GRCh38
rs765282472 Chr.17: 19657756-19657756 on Build GRCh38
rs372689616 Chr.17: 19657884-19657884 on Build GRCh38
rs148103086 Chr.17: 19657810-19657810 on Build GRCh38
rs764210473 Chr.17: 19657767-19657767 on Build GRCh38
rs766240333 Chr.17: 19657730-19657730 on Build GRCh38
rs901879859 Chr.17: 19657790-19657790 on Build GRCh38
rs761280501 Chr.17: 19657700-19657701 on Build GRCh38
rs747669738 Chr.17: 19657715-19657715 on Build GRCh38
rs781577862 Chr.17: 19657788-19657788 on Build GRCh38
rs747649145 Chr.17: 19657801-19657801 on Build GRCh38
rs369385931 Chr.17: 19657840-19657840 on Build GRCh38
rs72547567 Chr.17: 19657774-19657774 on Build GRCh38
rs746253303 Chr.17: 19657792-19657792 on Build GRCh38
rs759501619 Chr.17: 19657748-19657748 on Build GRCh38
rs773802049 Chr.17: 19657689-19657689 on Build GRCh38
rs866392702 Chr.17: 19657747-19657747 on Build GRCh38
rs764626215 Chr.17: 19657703-19657708 on Build GRCh38
rs776325315 Chr.17: 19657735-19657735 on Build GRCh38
rs772979684 Chr.17: 19657719-19657719 on Build GRCh38
rs769851025 Chr.17: 19657877-19657877 on Build GRCh38
rs886052688 Chr.17: 19657850-19657850 on Build GRCh38
rs751858811 Chr.17: 19657781-19657781 on Build GRCh38
rs77114981 Chr.17: 19657708-19657708 on Build GRCh38
rs758618354 Chr.17: 19657764-19657764 on Build GRCh38
rs369605228 Chr.17: 19657806-19657806 on Build GRCh38
rs72547566 Chr.17: 19657746-19657746 on Build GRCh38
rs770314692 Chr.17: 19657687-19657687 on Build GRCh38
rs141842478 Chr.17: 19657838-19657838 on Build GRCh38
rs753999443 Chr.17: 19657842-19657842 on Build GRCh38
rs375324873 Chr.17: 19657833-19657833 on Build GRCh38
rs757529940 Chr.17: 19657784-19657784 on Build GRCh38
rs897308650 Chr.17: 19657758-19657758 on Build GRCh38
rs1057517278 Chr.17: 19657863-19657863 on Build GRCh38
rs1057517352 Chr.17: 19657863-19657868 on Build GRCh38
rs534772670 Chr.17: 19657702-19657702 on Build GRCh38

View Full Product Details