Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132141852-132142111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX51
Entrez Gene ID
317781
Gene Name
DEAD-box helicase 51
Gene Aliases
-
UniGene
Hs.445168
Gene Chromosome Location
Chr.12: 132136594-132144335 on Build GRCh38
Transcripts
NM_175066.3 XM_011538256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3753064 c.965C>T p.A322V Chr.12: 132141880-132141880 on Build GRCh38
5970607 c.930G>A p.L310L Chr.12: 132141915-132141915 on Build GRCh38
6364186 c.929T>C p.L310P Chr.12: 132141916-132141916 on Build GRCh38
5970608 c.905A>G p.N302S Chr.12: 132141940-132141940 on Build GRCh38
5631749 c.930G>C p.L310L Chr.12: 132141915-132141915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200074001 Chr.12: 132141996-132141996 on Build GRCh38
rs543781716 Chr.12: 132141987-132141987 on Build GRCh38
rs780680296 Chr.12: 132141943-132141943 on Build GRCh38
rs756804939 Chr.12: 132141943-132141943 on Build GRCh38
rs774067276 Chr.12: 132141906-132141906 on Build GRCh38
rs776782181 Chr.12: 132141965-132141965 on Build GRCh38
rs749140549 Chr.12: 132141905-132141905 on Build GRCh38
rs778682017 Chr.12: 132141986-132141986 on Build GRCh38
rs754381528 Chr.12: 132141917-132141917 on Build GRCh38
rs747857727 Chr.12: 132141954-132141954 on Build GRCh38
rs1055693390 Chr.12: 132142063-132142063 on Build GRCh38
rs928758605 Chr.12: 132142039-132142039 on Build GRCh38
rs894490307 Chr.12: 132142070-132142070 on Build GRCh38
rs760041266 Chr.12: 132141918-132141918 on Build GRCh38
rs574441178 Chr.12: 132142018-132142018 on Build GRCh38
rs111718326 Chr.12: 132141907-132141907 on Build GRCh38
rs202048577 Chr.12: 132141963-132141963 on Build GRCh38
rs759368416 Chr.12: 132142056-132142056 on Build GRCh38
rs374298884 Chr.12: 132141947-132141947 on Build GRCh38
rs758112453 Chr.12: 132141992-132141992 on Build GRCh38
rs114651617 Chr.12: 132141920-132141920 on Build GRCh38
rs374721055 Chr.12: 132141898-132141898 on Build GRCh38
rs540285307 Chr.12: 132142046-132142046 on Build GRCh38
rs116281803 Chr.12: 132141882-132141882 on Build GRCh38
rs767039094 Chr.12: 132141916-132141916 on Build GRCh38
rs758364630 Chr.12: 132142044-132142044 on Build GRCh38
rs765888746 Chr.12: 132141981-132141981 on Build GRCh38
rs60927391 Chr.12: 132141880-132141880 on Build GRCh38
rs966088383 Chr.12: 132141926-132141926 on Build GRCh38
rs773370522 Chr.12: 132141909-132141909 on Build GRCh38
rs756805080 Chr.12: 132141884-132141884 on Build GRCh38
rs973594736 Chr.12: 132142008-132142008 on Build GRCh38
rs754939593 Chr.12: 132141984-132141984 on Build GRCh38
rs747237172 Chr.12: 132142006-132142006 on Build GRCh38
rs745332854 Chr.12: 132141893-132141893 on Build GRCh38
rs753711551 Chr.12: 132142054-132142054 on Build GRCh38
rs772377032 Chr.12: 132141956-132141956 on Build GRCh38
rs753295692 Chr.12: 132141983-132141983 on Build GRCh38
rs202047240 Chr.12: 132141919-132141919 on Build GRCh38
rs760129641 Chr.12: 132141967-132141967 on Build GRCh38
rs1044026748 Chr.12: 132142034-132142034 on Build GRCh38
rs114915755 Chr.12: 132142082-132142082 on Build GRCh38
rs201346110 Chr.12: 132142083-132142083 on Build GRCh38
rs767258596 Chr.12: 132141978-132141978 on Build GRCh38
rs753148115 Chr.12: 132141972-132141972 on Build GRCh38
rs200761682 Chr.12: 132141969-132141969 on Build GRCh38
rs749228331 Chr.12: 132141951-132141951 on Build GRCh38
rs115150746 Chr.12: 132141952-132141952 on Build GRCh38
rs866867846 Chr.12: 132141873-132141873 on Build GRCh38
rs750001522 Chr.12: 132141944-132141944 on Build GRCh38
rs552900844 Chr.12: 132141925-132141925 on Build GRCh38
rs374818022 Chr.12: 132142085-132142085 on Build GRCh38
rs761964646 Chr.12: 132141875-132141875 on Build GRCh38
rs908022011 Chr.12: 132142000-132142000 on Build GRCh38
rs773283835 Chr.12: 132141959-132141959 on Build GRCh38
rs766684366 Chr.12: 132142025-132142025 on Build GRCh38
rs760863787 Chr.12: 132141962-132141962 on Build GRCh38
rs769812971 Chr.12: 132141897-132141897 on Build GRCh38
rs764968350 Chr.12: 132141969-132141970 on Build GRCh38
rs754293715 Chr.12: 132141878-132141878 on Build GRCh38
rs367813909 Chr.12: 132142071-132142071 on Build GRCh38
rs760343231 Chr.12: 132142090-132142090 on Build GRCh38
rs774721533 Chr.12: 132142087-132142087 on Build GRCh38
rs780780937 Chr.12: 132141886-132141886 on Build GRCh38
rs745914025 Chr.12: 132142080-132142080 on Build GRCh38
rs376123326 Chr.12: 132141979-132141979 on Build GRCh38
rs760638825 Chr.12: 132141910-132141910 on Build GRCh38
rs574926162 Chr.12: 132141982-132141982 on Build GRCh38
rs199995098 Chr.12: 132141939-132141939 on Build GRCh38
rs983600829 Chr.12: 132141872-132141872 on Build GRCh38
rs776885124 Chr.12: 132142072-132142072 on Build GRCh38
rs559703354 Chr.12: 132142052-132142052 on Build GRCh38

View Full Product Details