Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81043295-81043536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATMIN
Entrez Gene ID
23300
Gene Name
ATM interactor
Gene Aliases
ASCIZ, ZNF822
UniGene
Hs.16349
Gene Chromosome Location
Chr.16: 81035853-81047346 on Build GRCh38
Transcripts
NM_001300728.1 NM_015251.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6606907 c.863C>T p.P288L Chr.16: 81043361-81043361 on Build GRCh38
4063059 c.929A>C p.Q310P Chr.16: 81043427-81043427 on Build GRCh38
127380 c.864G>A p.P288P Chr.16: 81043362-81043362 on Build GRCh38
5602865 c.921C>T p.P307P Chr.16: 81043419-81043419 on Build GRCh38
4510427 c.836C>T p.T279I Chr.16: 81043334-81043334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553978375 Chr.16: 81043435-81043435 on Build GRCh38
rs753748463 Chr.16: 81043456-81043456 on Build GRCh38
rs767720958 Chr.16: 81043405-81043405 on Build GRCh38
rs548482807 Chr.16: 81043333-81043333 on Build GRCh38
rs767819176 Chr.16: 81043369-81043369 on Build GRCh38
rs752528451 Chr.16: 81043451-81043451 on Build GRCh38
rs774929022 Chr.16: 81043401-81043401 on Build GRCh38
rs141724880 Chr.16: 81043462-81043462 on Build GRCh38
rs776210544 Chr.16: 81043516-81043516 on Build GRCh38
rs774940449 Chr.16: 81043471-81043471 on Build GRCh38
rs754899712 Chr.16: 81043348-81043348 on Build GRCh38
rs749866150 Chr.16: 81043371-81043371 on Build GRCh38
rs749380568 Chr.16: 81043466-81043466 on Build GRCh38
rs868332235 Chr.16: 81043360-81043360 on Build GRCh38
rs147897268 Chr.16: 81043354-81043354 on Build GRCh38
rs755667499 Chr.16: 81043329-81043329 on Build GRCh38
rs753090898 Chr.16: 81043406-81043407 on Build GRCh38
rs773439448 Chr.16: 81043494-81043494 on Build GRCh38
rs746037119 Chr.16: 81043362-81043362 on Build GRCh38
rs769279443 Chr.16: 81043376-81043376 on Build GRCh38
rs202062165 Chr.16: 81043340-81043340 on Build GRCh38
rs149138478 Chr.16: 81043420-81043420 on Build GRCh38
rs148070606 Chr.16: 81043461-81043461 on Build GRCh38
rs753345210 Chr.16: 81043419-81043419 on Build GRCh38
rs143600260 Chr.16: 81043444-81043444 on Build GRCh38
rs2278023 Chr.16: 81043411-81043411 on Build GRCh38
rs760445772 Chr.16: 81043495-81043495 on Build GRCh38
rs778748823 Chr.16: 81043350-81043350 on Build GRCh38
rs749981991 Chr.16: 81043322-81043322 on Build GRCh38
rs749231367 Chr.16: 81043396-81043396 on Build GRCh38
rs762248121 Chr.16: 81043402-81043402 on Build GRCh38
rs1014520209 Chr.16: 81043345-81043345 on Build GRCh38
rs866720462 Chr.16: 81043405-81043406 on Build GRCh38
rs766027738 Chr.16: 81043505-81043505 on Build GRCh38
rs748559297 Chr.16: 81043479-81043479 on Build GRCh38
rs922137973 Chr.16: 81043511-81043511 on Build GRCh38
rs768795012 Chr.16: 81043467-81043467 on Build GRCh38
rs747981765 Chr.16: 81043351-81043351 on Build GRCh38
rs777398323 Chr.16: 81043457-81043457 on Build GRCh38
rs17849653 Chr.16: 81043365-81043365 on Build GRCh38
rs539236397 Chr.16: 81043418-81043418 on Build GRCh38
rs772540932 Chr.16: 81043486-81043486 on Build GRCh38

View Full Product Details