Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 162383430-162383679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf226
Entrez Gene ID
400793
Gene Name
chromosome 1 open reading frame 226
Gene Aliases
-
UniGene
Hs.447011
Gene Chromosome Location
Chr.1: 162378906-162386818 on Build GRCh38
Transcripts
NM_001085375.1 NM_001135240.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1335736 c.791C>T p.A264V Chr.1: 162383526-162383526 on Build GRCh38
6122502 c.893G>T p.R298L Chr.1: 162383628-162383628 on Build GRCh38
4667377 c.794delC p.S266fs*14 Chr.1: 162383529-162383529 on Build GRCh38
4934148 c.890C>T p.A297V Chr.1: 162383625-162383625 on Build GRCh38
3477420 c.757G>A p.E253K Chr.1: 162383492-162383492 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043507790 Chr.1: 162383576-162383576 on Build GRCh38
rs200714502 Chr.1: 162383545-162383545 on Build GRCh38
rs748307397 Chr.1: 162383618-162383618 on Build GRCh38
rs956335898 Chr.1: 162383595-162383595 on Build GRCh38
rs183820691 Chr.1: 162383572-162383572 on Build GRCh38
rs769021007 Chr.1: 162383615-162383615 on Build GRCh38
rs964753877 Chr.1: 162383517-162383517 on Build GRCh38
rs952356905 Chr.1: 162383650-162383650 on Build GRCh38
rs376932252 Chr.1: 162383536-162383536 on Build GRCh38
rs199942554 Chr.1: 162383529-162383529 on Build GRCh38
rs781041784 Chr.1: 162383472-162383472 on Build GRCh38
rs905068861 Chr.1: 162383591-162383591 on Build GRCh38
rs751292437 Chr.1: 162383544-162383544 on Build GRCh38
rs752544365 Chr.1: 162383645-162383645 on Build GRCh38
rs200599520 Chr.1: 162383622-162383622 on Build GRCh38
rs767362145 Chr.1: 162383642-162383642 on Build GRCh38
rs534095245 Chr.1: 162383477-162383477 on Build GRCh38
rs753452827 Chr.1: 162383556-162383556 on Build GRCh38
rs770744623 Chr.1: 162383628-162383628 on Build GRCh38
rs367877155 Chr.1: 162383657-162383657 on Build GRCh38
rs754575231 Chr.1: 162383457-162383457 on Build GRCh38
rs766399642 Chr.1: 162383541-162383541 on Build GRCh38
rs747630045 Chr.1: 162383473-162383473 on Build GRCh38
rs772635250 Chr.1: 162383491-162383491 on Build GRCh38
rs777350627 Chr.1: 162383551-162383551 on Build GRCh38
rs780656454 Chr.1: 162383575-162383575 on Build GRCh38
rs371228999 Chr.1: 162383508-162383508 on Build GRCh38
rs577076806 Chr.1: 162383614-162383614 on Build GRCh38
rs568340772 Chr.1: 162383476-162383476 on Build GRCh38
rs760397357 Chr.1: 162383651-162383651 on Build GRCh38
rs766171442 Chr.1: 162383641-162383641 on Build GRCh38
rs747460918 Chr.1: 162383507-162383507 on Build GRCh38
rs374369490 Chr.1: 162383627-162383627 on Build GRCh38
rs768734941 Chr.1: 162383511-162383511 on Build GRCh38
rs775711895 Chr.1: 162383501-162383501 on Build GRCh38
rs774305081 Chr.1: 162383639-162383639 on Build GRCh38
rs776875683 Chr.1: 162383519-162383519 on Build GRCh38
rs762047926 Chr.1: 162383523-162383523 on Build GRCh38
rs755844997 Chr.1: 162383548-162383548 on Build GRCh38
rs752347885 Chr.1: 162383546-162383546 on Build GRCh38

View Full Product Details