Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89932500-89932741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACH2
Entrez Gene ID
60468
Gene Name
BTB domain and CNC homolog 2
Gene Aliases
BTBD25
UniGene
Hs.269764
Gene Chromosome Location
Chr.6: 89926528-90296912 on Build GRCh38
Transcripts
NM_001170794.1 NM_021813.3 XM_005248758.4 XM_005248759.4 XM_011536039.2 XM_011536040.2 XM_017011156.1 XM_017011157.1 XM_017011158.1 XM_017011159.1 XM_017011160.1 XM_017011161.1 XM_017011162.1 XM_017011163.1 XM_017011164.1 XM_017011165.1 XM_017011166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5871242 c.2311G>A p.E771K Chr.6: 89932623-89932623 on Build GRCh38
6495735 c.2272T>C p.S758P Chr.6: 89932662-89932662 on Build GRCh38
5424492 c.2264T>C p.I755T Chr.6: 89932670-89932670 on Build GRCh38
5886450 c.2338C>T p.P780S Chr.6: 89932596-89932596 on Build GRCh38
3630541 c.2326C>T p.P776S Chr.6: 89932608-89932608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368416831 Chr.6: 89932619-89932619 on Build GRCh38
rs754299207 Chr.6: 89932568-89932568 on Build GRCh38
rs776040980 Chr.6: 89932666-89932666 on Build GRCh38
rs140270440 Chr.6: 89932694-89932694 on Build GRCh38
rs750705861 Chr.6: 89932596-89932596 on Build GRCh38
rs1020109921 Chr.6: 89932656-89932656 on Build GRCh38
rs777216582 Chr.6: 89932616-89932616 on Build GRCh38
rs878885096 Chr.6: 89932715-89932715 on Build GRCh38
rs767049051 Chr.6: 89932541-89932541 on Build GRCh38
rs748305367 Chr.6: 89932690-89932690 on Build GRCh38
rs764721711 Chr.6: 89932550-89932550 on Build GRCh38
rs757652995 Chr.6: 89932572-89932572 on Build GRCh38
rs768610688 Chr.6: 89932602-89932602 on Build GRCh38
rs745724297 Chr.6: 89932651-89932651 on Build GRCh38
rs747141129 Chr.6: 89932601-89932601 on Build GRCh38
rs373437222 Chr.6: 89932531-89932531 on Build GRCh38
rs372285524 Chr.6: 89932624-89932624 on Build GRCh38
rs752773442 Chr.6: 89932705-89932705 on Build GRCh38
rs374620073 Chr.6: 89932653-89932653 on Build GRCh38
rs925306997 Chr.6: 89932536-89932536 on Build GRCh38
rs767748883 Chr.6: 89932699-89932699 on Build GRCh38
rs915321630 Chr.6: 89932657-89932657 on Build GRCh38
rs749282554 Chr.6: 89932645-89932645 on Build GRCh38
rs757186928 Chr.6: 89932648-89932648 on Build GRCh38
rs139170041 Chr.6: 89932635-89932635 on Build GRCh38
rs760300804 Chr.6: 89932614-89932614 on Build GRCh38
rs761363627 Chr.6: 89932667-89932667 on Build GRCh38
rs968647499 Chr.6: 89932523-89932523 on Build GRCh38
rs577012189 Chr.6: 89932561-89932561 on Build GRCh38
rs1042523147 Chr.6: 89932681-89932681 on Build GRCh38
rs763412318 Chr.6: 89932670-89932670 on Build GRCh38
rs141231678 Chr.6: 89932612-89932612 on Build GRCh38
rs759143305 Chr.6: 89932534-89932534 on Build GRCh38
rs368652670 Chr.6: 89932654-89932654 on Build GRCh38
rs371078442 Chr.6: 89932548-89932548 on Build GRCh38
rs147649326 Chr.6: 89932693-89932693 on Build GRCh38
rs755611605 Chr.6: 89932602-89932603 on Build GRCh38
rs188927020 Chr.6: 89932597-89932597 on Build GRCh38
rs769816700 Chr.6: 89932526-89932526 on Build GRCh38
rs768240230 Chr.6: 89932665-89932665 on Build GRCh38
rs766794727 Chr.6: 89932688-89932688 on Build GRCh38
rs879226832 Chr.6: 89932593-89932593 on Build GRCh38
rs199646582 Chr.6: 89932607-89932607 on Build GRCh38
rs750863282 Chr.6: 89932621-89932621 on Build GRCh38
rs201359587 Chr.6: 89932532-89932532 on Build GRCh38
rs139627462 Chr.6: 89932603-89932603 on Build GRCh38
rs150771521 Chr.6: 89932615-89932615 on Build GRCh38
rs965034801 Chr.6: 89932707-89932707 on Build GRCh38
rs3798789 Chr.6: 89932639-89932639 on Build GRCh38
rs773849871 Chr.6: 89932669-89932669 on Build GRCh38
rs755100835 Chr.6: 89932638-89932638 on Build GRCh38
rs375103380 Chr.6: 89932613-89932613 on Build GRCh38
rs773007535 Chr.6: 89932530-89932530 on Build GRCh38
rs200505776 Chr.6: 89932600-89932600 on Build GRCh38
rs867412754 Chr.6: 89932605-89932605 on Build GRCh38
rs766772225 Chr.6: 89932626-89932626 on Build GRCh38
rs765807351 Chr.6: 89932620-89932620 on Build GRCh38
rs147908828 Chr.6: 89932714-89932714 on Build GRCh38
rs78688236 Chr.6: 89932573-89932573 on Build GRCh38
rs200671083 Chr.6: 89932608-89932608 on Build GRCh38

View Full Product Details