Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99515111-99515370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC85C
Entrez Gene ID
317762
Gene Name
coiled-coil domain containing 85C
Gene Aliases
-
UniGene
Hs.709288
Gene Chromosome Location
Chr.14: 99511266-99604390 on Build GRCh38
Transcripts
NM_001144995.1 XM_011536705.2

SNP Detail

SNP ID Chromosome Location
rs762454138 Chr.14: 99515224-99515224 on Build GRCh38
rs773964458 Chr.14: 99515222-99515222 on Build GRCh38
rs756621787 Chr.14: 99515285-99515285 on Build GRCh38
rs761118332 Chr.14: 99515279-99515279 on Build GRCh38
rs983890738 Chr.14: 99515200-99515200 on Build GRCh38
rs893404592 Chr.14: 99515142-99515142 on Build GRCh38
rs772614849 Chr.14: 99515228-99515228 on Build GRCh38
rs902890101 Chr.14: 99515234-99515234 on Build GRCh38
rs766605931 Chr.14: 99515291-99515291 on Build GRCh38
rs765352190 Chr.14: 99515344-99515344 on Build GRCh38
rs372260518 Chr.14: 99515264-99515264 on Build GRCh38
rs1031737672 Chr.14: 99515139-99515139 on Build GRCh38
rs545417219 Chr.14: 99515152-99515153 on Build GRCh38
rs759725474 Chr.14: 99515302-99515302 on Build GRCh38
rs748899137 Chr.14: 99515207-99515207 on Build GRCh38
rs939913445 Chr.14: 99515212-99515212 on Build GRCh38
rs76063188 Chr.14: 99515136-99515136 on Build GRCh38
rs562759068 Chr.14: 99515211-99515211 on Build GRCh38
rs768309214 Chr.14: 99515213-99515213 on Build GRCh38
rs895199409 Chr.14: 99515287-99515287 on Build GRCh38
rs577159402 Chr.14: 99515164-99515164 on Build GRCh38
rs375793466 Chr.14: 99515176-99515176 on Build GRCh38
rs766385345 Chr.14: 99515254-99515254 on Build GRCh38
rs754124918 Chr.14: 99515296-99515296 on Build GRCh38
rs1012765149 Chr.14: 99515276-99515276 on Build GRCh38
rs958771631 Chr.14: 99515270-99515270 on Build GRCh38
rs566033676 Chr.14: 99515177-99515177 on Build GRCh38
rs757830225 Chr.14: 99515227-99515227 on Build GRCh38
rs754187800 Chr.14: 99515345-99515345 on Build GRCh38
rs953885950 Chr.14: 99515182-99515182 on Build GRCh38

View Full Product Details